Discover the power of biomarker testing on KRYPTOR for your laboratory
Benefits of B·R·A·H·M·S biomarkers for prenatal screening, including outstanding lot-to-lot stability, high precision with low interference, efficient workflow for short turnaround time, and more than 20 years of expertise.
Our B·R·A·H·M·S Biomarkers on the KRYPTOR platform offer cutting-edge and novel testing to enhance the care of preeclampsia and other life-threatening diseases. With our solution, you can improve clinical decision making while streamlining workflows and optimizing efficiencies in your laboratory.
Optimize your laboratory routine with biomarker testing on KRYPTOR for preeclampsia management
KRYPTOR compact PLUS analyzer
With Nobel Prize winning TRACE technology, Thermo Scientific B·R·A·H·M·S KRYPTOR compact PLUS and biotin-free assays provide results with low interference and high precision. This user-friendly bench-top instrument is easily integrated into any laboratory. Enhance biomarker testing in your lab with:
  • An easy and straightforward laboratory routine requiring less operator intervention
  • With the self-determining dilution factor, reduce human error with increased instrument autonomy
  • Cost reduction due to efficient sample processing
  • Low level of liquid and plastic waste for an environment friendly solution 
Get your sFlt-1/PlGF ratio result in 30 minutes and leverage the optimized workflow offered by the instrument.
The first and only FDA cleared biomarkers for the risk assessment and clinical management of preeclampsia
Thermo Scientific B·R·A·H·M·S sFlt-1/PlGF KRYPTOR Test System offers the highest level of compliance with FDA clearance on the B·R·A·H·M·S KRYPTOR compact PLUS.  
Intended use:
Chart illustrating the FDA approved clinical indication for B·R·A·H·M·S PlGF plus KRYPTOR and B·R·A·H·M·S sFlt-1 KRYPTOR. The action is to aid in the risk assessment of progression to preeclampsia with severe features. The patient type is for singleton pregnancy between gestation age 23+0 to 34+6/7 weeks as well as hospitalized for hypertensive disorders of pregnancy. In addition, this test can be used for patients hospitalized for hypertensive disorders of pregnancy. Timing is within next two weeks of presentation.
*As defined by American College of Obstetricians and Gynecologists (ACOG) guidelines
**Preeclampsia, chronic hypertension with or without superimposed preeclampsia, or gestaional hypertension
Learn from our customers
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S is a registered trademark of B·R·A·H·M·S GmbH.
KRYPTOR and TRACE are trademarks of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific. Nobel Prize is a registered trademark of the Nobel Foundation.
1x1 image pixel for data collection