c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Environmental scanning electron microscope
The Thermo Scientific Prisma E Scanning Electron Microscope (SEM) combines a wide array of imaging and analytical modalities with advanced automation to offer the most complete solution of any instrument in its class. It is ideal for industrial research and development, quality control, and failure analysis applications that require high resolution, sample flexibility and an easy-to-use operator interface. The Prisma E SEM succeeds the highly successful Thermo Scientific Quanta SEM.
ESEM with EDS
The unique combination of accessible all-round performance, a large set of accessories, and the most intuitive elemental analysis with Thermo Scientific ColorSEM make the Prisma E SEM the go-to SEM for micro-scale imaging and analysis in any industry or field.
Prisma E Scanning Electron Microscope features
Elemental information at your fingertips
Live composition-based image coloring for intuitive elemental analysis with optional ChemiSEM Technology and integrated energy-dispersive X-ray spectroscopy (EDS). Speed up your work and obtain the most complete sample information with always-on analysis.
Excellent image quality
Excellent image quality at low kV and low vacuum thanks to flexible vacuum modes, including through-the-lens differential pumping. Simultaneous secondary electron (SE) and backscattered electron (BSE) imaging in every mode of operation.
Minimize sample preparation time
Low vacuum and ESEM capability enable charge-free imaging and analysis of nonconductive and/or hydrated specimens.
In-situ study of materials in their natural state
With the Prisma E SEM's environmental SEM (ESEM) mode, samples can be imaged even if they are hot, dirty, outgassing or wet.
Excellent analytical capabilities
Excellent analytical capabilities with a chamber that allows 3 simultaneous EDS detectors, EDS ports that are 180° opposite, wavelength-dispersive spectroscopy (WDS), coplanar EDS/EBSD and high-quality charge-free EDS and EBSD in low vacuum.
Easy to use
Easy to use, intuitive software with user guidance and undo functionality makes highly effective operation possible for novice users, while enabling experts to do their work faster and with fewer mouse clicks.
Prisma E Scanning Electron Microscope specifications
Style Sheet for Products Table Specifications
Resolution
  • 3.0 nm at 30 kV in high vacuum, low vacuum and ESEM
  • 7.0 nm at 3 kV (BSED, beam decel.)
Standard detectors
  • ETD, low-vacuum SED (LVD), gaseous SED for ESEM mode (GSED), IR camera
Optional detectors
  • Thermo Scientific Nav-Cam+ Camera, DBS, DBS-GAD, ESEM-GAD, STEM 3+, WetSTEM, RGB-CLD, EDS, EBSD, WDS, Raman, EBIC, etc.
ChemiSEM Technology (optional)
  • Live quantitative SEM image coloring is available based on energy-dispersive X-ray spectroscopy (EDS). Point & ID, linescan, region, element maps and accurate Noran quantification are included.
Stage bias (beam deceleration, optional)
  • -4,000 V to +50 V
Low vacuum mode
  • Up to 2,600 Pa (H2O) or 4,000 Pa (N2)
Stage
  • 5-axis motorized eucentric stage, 110 x 110 mm2 with a 105° tilt range. Maximum sample weight: 5 kg in un-tilted position.
Standard sample holder
  • Standard multi-sample SEM holder uniquely mounts directly
  • onto the stage, hosts up to 18 standard stubs (⌀ 12 mm), does
  • not require tools to mount a sample
Chamber
  • 340 mm inside width, 12 ports, three simultaneous EDS detectors possible, two at 180°, coplanar EDS/EBSD orthogonal to the tilt axis of the stage
In situ accessories (optional)
  • Software controlled -20°C to +60°C Peltier cold stage
  • Software controlled 1,000°C low vacuum/ESEM heating stage
  • Software controlled 1,100°C high vacuum heating stage
  • Software controlled 1,400°C low vacuum/ESEM heating stage
  • Integrated gas injection: up to 2 units (other accessories may limit number of gas injection systems (GIS) available) for beam-induced deposition of the following materials:
    • Platinum
    • Tungsten
    • Carbon
  • Manipulators
  • Cryo-stage
  • Electrical probing / multi-probing stations
Software options
Style Sheet for Komodo Tabs
Resources
Webinar: Scanning electron microscopy: selecting the right technology for your needs
This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
  • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
  • How samples are characterized in their natural state without the need for sample preparation.
  • How new advanced automation allows researchers to save time and increase productivity.
Video Player is loading.
Current Time 0:00
Duration 1:13
Loaded: 13.60%
Stream Type LIVE
Remaining Time 1:13
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Goldrush. This mixture of particles contains gold (large area on the left), copper (smallest particles) and a Al/Si/Mg alloy (the bigger particles). The mixture is heated to higher and higher temperatures. At a certain point, gold starts diffusing across the metal surfaces, thereby completely changing their appearance. The experiment is performed using the High Vacuum Heating Stage mounted on a Prisma E SEM.
    Webinar: Scanning electron microscopy: selecting the right technology for your needs
    This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
    • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
    • How samples are characterized in their natural state without the need for sample preparation.
    • How new advanced automation allows researchers to save time and increase productivity.
    Video Player is loading.
    Current Time 0:00
    Duration 1:13
    Loaded: 13.60%
    Stream Type LIVE
    Remaining Time 1:13
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Goldrush. This mixture of particles contains gold (large area on the left), copper (smallest particles) and a Al/Si/Mg alloy (the bigger particles). The mixture is heated to higher and higher temperatures. At a certain point, gold starts diffusing across the metal surfaces, thereby completely changing their appearance. The experiment is performed using the High Vacuum Heating Stage mounted on a Prisma E SEM.
      Applications
      Process control using electron microscopy
      Process control using electron microscopy
      Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
      Quality control and failure analysis using electron microscopy
      Quality control and failure analysis
      Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
      Fundamental Materials Research
      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
      Semiconductor-RD_1920x1080
      Semiconductor research and development
      Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
      Semi-Metrology_1920x1080
      Semiconductor metrology
      Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
      Semiconductor Failure Analysis
      Semiconductor Failure Analysis
      Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
      Semi-Materials-Character_1920x1080
      Semiconductor materials characterization
      Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
      Techniques
      EDS Analysis with ChemiSEM Technology
      Energy dispersive X-ray spectroscopy for materials characterization.
      Learn more ›
      Imaging Hot Samples
      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
      Learn more ›
      Environmental SEM (ESEM)
      Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
      Learn more ›
      Particle analysis
      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
      Learn more ›
      Cathodoluminescence
      Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
      Learn more ›
      Semiconductor Analysis and Imaging
      Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
      Learn more ›
      EDS Analysis with ChemiSEM Technology
      Energy dispersive X-ray spectroscopy for materials characterization.
      Learn more ›
      Imaging Hot Samples
      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
      Learn more ›
      Environmental SEM (ESEM)
      Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
      Learn more ›
      Particle analysis
      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
      Learn more ›
      Cathodoluminescence
      Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
      Learn more ›
      Semiconductor Analysis and Imaging
      Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
      Learn more ›
      Style Sheet to change H2 style to p with em-h2-header class
      Contact us
      Electron microscopy services
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
      Learn more ›
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards