Core chemicals for oligonucleotides
For laboratories involved in custom oligonucleotide synthesis
We provide customized chemicals and chemical blends you need—on time, whenever you need it
Thermo Fisher Scientific has a long-standing history of manufacturing high quality chemicals for over 65 years—and the chemicals required for oligonucleotide synthesis are no exception. 
We offer custom chemical and chemical blends specifically designed for oligo synthesis. 
These include:
  • Extra-dry solvents
  • Deblocking agents
  • CAP A, CAP B
  • Activators
  • Oxidizers
We understand that every oligo customer is different.
Whether you're engaged in research, scale-up, or production, our customizable chemical and chemical blends are formulated to your precise requirements. Working with our team of experts, our service is tailored to meet your exact specifications.
Define your chemical needs, and we will manufacture the right products to enable you to synthesize oligonucleotides with high purity and yield to achieve the desired results for your oligonucleotide applications.
Decorative image highlighting Quality, Global supply, Partnership and Sustainable
Quality and compliance
Within our Thermo Scientific chemicals products and service, we offer compliant-ready specifications, with full traceability and complete documentation.
We adhere to stringent quality control processes in manufacturing – and custom QC is available upon request. Our products are manufactured in a world-class ISO-9001 facility.
As part of our service, we can provide you with validation samples from multiple lots. You will also have access to your own fleet of returnable containers: to ensure there is no potential for cross-contamination. 
Global supply
Our dedicated teams are committed to meeting your specific requirements across various geographies. This includes providing support for fleet management to ensure efficient logistics and transportation.
Our products are available from our global network of warehouses and can be delivered quickly to you: whenever and wherever you are located.
Partnership
Our teams have a proven track record of cultivating robust partnerships, offering technical expertise, providing valuable consultation, managing projects effectively, and maintaining clear and open lines of communication.
We recognize the significance of customizing our services to meet your specific requirements, and our dedicated teams are committed to supporting you throughout the entire process.
Sustainable
By utilizing the returnable container service within your fleet, the amount of glass and cardboard that needs to be disposed of can be reduced. For example, each 200 L returnable container eliminates almost 200 lbs./90 kgs of glass and cardboard.
As part of our service to you, we also take care of cleaning, maintaining, and refilling the empty drums/barrels, which not only reduces your hazardous disposal costs but also eliminates the need for rinsing at your site. 
A simplistic overview of the returnable container program that feeds into the fleet cycle we offer.
A simplistic overview of the returnable container program that feeds into the fleet cycle we offer, can be seen above.
Alongside these chemicals, we also have numerous phosphoramidites available within our product range as well as offer options for custom-synthesized oligos, primers, probes.
  • Shop for phosphoramidites here
  • Learn more about our custom-synthesized oligos, primers, probes here
We also offer solutions for the analysis of oligonucleotides, including the LC columns, vials and well-plates, instrumentation, and assay solutions.
Oligo synthesis basics
New to Oligo synthesis? Learn more about how chemicals support the process
Oligonucleotides (or “Oligos” for short) are short strands of nucleic acids, specifically DNA or RNA, that consist of a sequence of nucleotide building blocks. They are typically composed of around 10 to 100 nucleotides in length.
Oligonucleotides serve as essential tools in various areas of molecular biology research, diagnostics development, and therapeutics development. They are utilized for a wide range of applications, including DNA amplification, sequencing, gene expression analysis, gene editing, biomarker detection, nucleic acid-based assays, and drug development, among others.
What are the important factors when performing oligonucleotide synthesis?
There are several important factors to consider when performing oligonucleotide synthesis. These factors include the length and sequence of the oligonucleotide, the purity and yield of the product, the presence of any modifications or labeling, the turnaround time and cost of the service, and the quality and reliability of the supplier.
Additionally, you may also require specific technical support and expertise in designing and optimizing oligonucleotide synthesis protocols for their specific applications. By addressing these factors and providing high-quality, reliable services, suppliers can effectively meet the needs of their customers and establish a strong reputation in the industry.
How and why are chemicals used for oligonucleotide synthesis?
In oligonucleotide synthesis, a variety of chemicals are used to facilitate the synthesis process, and the process of producing the target oligomer sequence takes several cycles consisting of specific synthetic steps.
These chemicals typically include CAP A and CAP B, activators, oxidizers, deblocking reagents, solvents and phosphoramidites.
Activators and oxidizers are used to promote the coupling and oxidation steps, respectively, to help ensure efficient formation of phosphodiester bonds.
Deblocking reagents are employed to remove the protecting groups from the nucleotides, enabling subsequent addition of the next nucleotide in the sequence.
Solvents, such as acetonitrile or dichloromethane, are used to dissolve and mix the various reagents during the synthesis process.  Acetonitrile is particularly important as it is used in every step of the synthetic cycle as a washing solvent.
Phosphoramidites are the building blocks used to add individual nucleotides to the growing oligonucleotide chain. Amidites are used to protect the reactive groups on the nucleotide building blocks during synthesis.
As part of the synthesis cycle, capping reagents are used to convert unreacted OH groups to acetate groups to prevent them taking part in the synthetic cycle thereon. They can then be removed in the purification step. This prevents the formation of n-1 oligomers in the final product.
It is important to note that the specific chemicals and protocols used may vary depending on the synthesis platform and the desired application of the synthesized oligonucleotides.
Oligo C&B cycle
For Research Use Only. Not for use in diagnostic procedures.