Intuitive, user-programmable software
A user-friendly graphical user interface controls the CTS Rotea system. Create and optimize protocols for cell separation, washing, and concentration. The Rotea Protocol Builder software guides Rotea users through the protocol development process, while the simulation software helps ensure the protocols created will perform as intended before testing with samples. Additionally, there are software settings which can manage user control of process settings in a cGMP lab.
Video Player is loading.
Current Time 0:00
Duration 1:56
Loaded: 0%
Stream Type LIVE
Remaining Time 1:56
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Software to help enable 21 CFR part 11 compliance
    When scaling up to commercial manufacturing, we recommend upgrading one of the two software options available, the CTS Rotea SAE Software or the Gibco CTS Cellmation™ Software for DeltaV Systems to help enable 21 CFR part 11 compliance. Both software options help customers prepare for audit and regulatory filing processes through security, auditing, e-signature functionality while customers maintain the final responsibility for 21 CFR Part 11 compliance.
    Security, audit, and e-signature (SAE) software
    The Rotea SAE Software is designed for customers who want to upgrade, standalone instruments. The capabilities include password policies, user role designations, role permissions, tracking of actions and information via an audit log, and electronic signature configuration for specific functions. Customer preferences for these SAE settings are managed within the SAE Administrator Console (separate software application), which is recommended to be installed on the co-located Rotea system computer to enable uninterrupted communication between the system and the software for continuous tracking.
    Functionality of the SAE Administrator Console
    Cellmation software
    The Gibco CTS Cellmation Software is an off-the-shelf cell therapy manufacturing solution that provides digital connectivity to DeltaV for a range of modular cell therapy instruments offered by Thermo Fisher Scientific. The open platform allows for flexible management and integration of multiple instruments, streamlining cell therapy workflows and helping to ensure regulatory compliance. Powered by the Emerson DeltaV Distributed Control System (DCS), CTS Cellmation software helps ensure traceable and secure data storage as well as streamlined data management across processes.
    Schematic of CTS Cellmation Software connected to a range of modular cell therapy instruments
    Ordering information for available software upgrades
    The Rotea SAE Software or the Cellmation Software for DeltaV Systems can be purchased as part of an initial instrument purchase or as a separate, standalone purchase later. Each software option requires a system upgrade service by a member of the Global Service and Support team to enable the software.
    Rotea system software updates
    To review Release Notes and download the latest software and compatible firmware for the Rotea Counterflow Centrifugation System, visit the software download page.
    For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products.
    1x1 image pixel for data collection