plastic-packaging-bottles-vials-banner-470x200
Setting the standard for superior quality
Only the highest-quality resins that meet the most stringent standards are used in pharmaceutical, university research, food and consumer applications. Superior resins ensure low levels of extractables that can contaminate bottle contents.
Popular plastic packaging bottles and vials
nalgene-fep-bottles-bulk-pack-popular-products-210x100
nalgene-continuous-thread-lined-closures-serum-vials-sterile-bulk-pack-popular-products-210x100
nalgene-boston-round-narrow-mouth-hdpe-bottles-closure-bulk-pack-popular-products-210x100
Featured plastic packaging bottle and vial categories
Bulk Pack Laboratory Bottles
Square, wide mouth, general purpose bottles made from high-quality plastics in a variety of materials and shapes.
Bulk Pack Packaging Bottles
Bottles for storing, shipping, and packaging. Bulk pack packaging bottles are ideal as economical packaging containers for countless applications. They are available in multiple sizes and packaging formats.
Micro Packaging Vials
Containers for sampling, storage and shipping. Micro-packaging multiple closures options allows you to easily identify the reagents in your kits to minimize the chance for cross contamination.
Packaging Accessories
Securely seal your reagents. Accessories including a variety of colored closures and torque equipment for securely sealing your reagents. Closures are one-piece and linerless with integrally molded seal ring to ensure leakproof performance no one else can match. Torque wrench fittings ensure accurate torque readings.
Low Particulate Bottles
Accurately fill and pour your samples. Nalgene low particulate/low metal bottles provide excellent chemical resistance and are perfect for storing samples, bulk intermediates and active pharmaceutical ingredients.
Media Bottles
Our media bottles are biologically tested, leakproof, and sterile. Made from polyethylene terephthalate (PET) and polyethylene terephthalate copolyester glycol modified (PETG), their square shape design saves space.
Serum Vials
Protect against leakage, breakage and contamination. Our serum vials are lighter in weight than glass, are transparent for easy content viewing.
Diagnostic Bottles
Small volume bottles for sampling, storing, and shipping. Guaranteed leakproof, our diagnostic bottles are perfect for packaging your small volume reagents.
Dropper Bottles
Provide reliable and repeatable dispensing. Made of low-density polyethylene (LDPE) for packaging and dispensing your valuable reagents, Thermo Scientific™ Nalgene™ dropper bottles are an excellent alternative to pipetting and other dispensing devices. Our dropper bottles are available in 4-, 8-, and 15-mL sizes, white and natural with multiple colored closures.
rp-lab-bottles
Nalgene laboratory bottles are in a class of their own.
They're iconic in labs the world over for their guaranteed leakproof performance and rugged construction. What makes Nalgene so great? Thermo Scientific Nalgene bottles and their caps are carefully engineered to work perfectly together, providing dependable leakproof performance, every time, guaranteed.
Shop Laboratory Bottles ›