Guaranteed to deliver performance without particle release
Engineered for the analysis of both polar and nonpolar compounds, these Thermo Scientific GC columns incorporate a porous intermediate-polarity phase into the tubing walls with essentially no particle release thereby not only ensuring superior column performance and stability but also preventing damage to detection systems.
Contact us
Featured PLOT Columns
Common applications:
  • Alkanes up to C12
  • Unsaturated hydrocarbon isomers
  • Permanent and refinery or natural gases
  • Oxygenated compounds and solvents
Column
Recommended Application
TracePLOT™ TG-BOND Alumina GC Na2SO4 ColumnsUse these columns for linear and quantitative analyses of polar unsaturated hydrocarbons
TracePLOT™ TG-BOND Alumina GC KCl Deactivation ColumnsChoose these columns for linear and quantitative analyses of polar unsaturated hydrocarbons
TracePLOT™ TG-BOND Msieve 5A GC ColumnsSeparate AR/O2 and other permanent gases using these columns that incorporate the latest innovations in PLOT technology
TracePLOT™ TG-BOND Q GC ColumnsAchieve consistent, reproducible separations of CO2, methane and 02/N2/CO with these non-polar PLOT columns with essentially no particle release
TracePLOT™ TG-BOND Q+ GC ColumnsSeparate ethane, ethylene and acetylene consistently with these intermediate polarity PLOT columns with essentially no particle release
TracePLOT™ TG-BOND S GC ColumnsChoose these mid-polarity PLOT columns with essentially no particle release for consistent, reproducible analyses of non-polar and polar compounds
TracePLOT™ TG-BOND U GC ColumnsChoose these polar divinylbenzene ethylene glycol/dimethylacrylate PLOT columns for consistent, reproducible analyses of nonpolar and polar compounds with essentially no particle release
Chromatography Consumables Resources and Information
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Discover more resources
Need to find GC columns or accessories?
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
Get the GC columns catalog
CMD SchemaApp code