Ion Chromatography Detectors
Detection options for all the different kinds of analytes you need to measure
Choose the best conductivity (CD), electrochemical (ECD), variable wavelength (VWD), photodiode array (PDA) ion chromatography (IC) detector, or mass spectrometry (MS) detector for your IC application. Our detectors offer high sensitivity and minimum peak dispersion in an all-PEEK flow path, making the detectors compatible with eluents of pH 0–14.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Contact us
IC detectors
Conductivity IC Detectors
Conductivity IC detectors
Achieve high-performance conductivity detection for your ion chromatography separations.
Electrochemical IC Detectors
Electrochemical IC detectors
This detector is highly sensitive and specific to susceptible molecules, and is often used in tandem with conductivity detection as a general method that characterizes all ions in the sample.
PDA Photodiode Array IC Detector
PDA photodiode array IC detector
Maximize optical absorbance information with high-resolution, full UV-Vis spectra, highly flexible multiple wavelength signals and referencing capabilities. For use on all models of Dionex ICS systems.
VWD Variable Wavelength IC Detector
VWD variable wavelength IC detector
Get high-performance, multiple wavelength UV-Vis detection for routine ion chromatography with all models of Thermo Scientific Dionex ICS systems.
Single quadrupole mass spectrometer
Add the sensitivity and specificity of mass spectrometry (MS) to IC applications with this single quadrupole MS detector.
Need IC detector parts & accessories?
Find the right replacement lamps, detector cells, electrodes, and other parts and kits for all your Dionex IC Systems.
IC Detector parts and accessories
CMD SchemaApp code