Search Thermo Fisher Scientific
The answer depends on the type of ion exchange chromatography analysis you are running and the special features you might need. We offer a range of ion chromatography (IC) solutions from robust, basic systems for everyday analyses to high-throughput systems with electrolytic eluent generation and multiple detector options. All our latest IC systems provide high-pressure ion chromatography (HPIC) analysis capabilities. Explore the selection guide below to discover which IC system is right for your laboratory.
|
|
||
Description |
|||
Experience the benefits of continuous chemical suppression with high-pressure operation using the Thermo Scientific Dionex Inuvion Core IC system. This easily configurable platform allows you to upgrade as analysis requirements increase. | Simplify your ion analysis with the Thermo Scientific Dionex Inuvion IC system. This configurable IC system provides high-pressure operation, eluent degas and electrolytic suppression capabilities, with the option to add Reagent-Free IC eluent generation. |
The easy-to-use, reliable Thermo Scientific Dionex Integrion HPIC system performs versatile and efficient analyses. It offers flexible detector configurations, automated eluent generation, and high-pressure capabilities for faster analysis without compromising data quality. |
The Thermo Scientific Dionex ICS-6000 HPIC system is a modular IC system that comes in standard and microbore configurations and provide best-in-class resolution, speed and sensitivity, with 2D analysis capabilities. |
Chemical suppression |
|||
Automated sample preparation |
|||
|
|
|
|
High-pressure operation (up to 5,000 psi) |
|||
Electrolytically regenerated suppression |
|||
Electrolytic eluent generation |
|||
Gradient separations |
|||
|
|
||
Integrated electrochemical cell |
|||
|
|||
Modular configuration |
|||
|
|
||
Configurable as independent dual system |
|||
|
|||
Proportioned mechanical gradients | |||
2D chromatography | |||
* Indicates partial capability