Advanced technologies for monitoring and analysis to help deliver reliable results over a wide range of gases and particulate concentrations
Depending on the type of gas and particulates targeted for monitoring and the data output desired, various technologies have been proven to effectively analyze ambient air and detect its components.  We offer a wide range of products that utilize these technologies to help enable more reliable air quality monitoring for your operation. 
Beta attenuation
When beta rays strike a material, they can be absorbed, reflected or pass directly through. The attenuation of intensity in beta rays is proportional to the amount of material present. The attenuation through most materials is relatively consistent and is based on the electron density of the material (calculated by dividing the atomic number by the atomic mass). The attenuation for most materials is about 0.5, except for hydrogen and heavy metals.
The principle behind beta attenuation particulate sampling instruments (beta gauge) is that energy is absorbed from beta particles as they pass through particulate matter (PM) collected on a filter media. Beta gauge instruments have been designed to take advantage of this scientific principle to monitor/measure particulate matter (PM) concentrations. The attenuation due to only the PM is measurable if a baseline beta count through just the filter can be established prior to sampling. The difference between the baseline beta count and the beta count after sampling is directly proportional to the mass of PM in the sample.
Beta rays concept
The two main components of a beta attenuation measuring system are the beta source and the detector. The beta source must be selected so that: it has an energy level high enough for the beta particles to pass through the air volume, collection media (i.e., the filter tape) and the particulate; it has enough source material so that a high count rate is present; it is stable over long periods of time; and it does not present a danger to the health of personnel that come into contact with the instrument. The source of choice has been carbon-14 because: it has a safe yet high enough energy level; it has a half-life of 5,568 years; and it is relatively abundant. Many different types of detectors can quantify beta particle counts, but the ones most widely used are the Geiger Mueller counter or a photodiode detector. 
Thermo Scientific 5014i Beta Continuous Ambient Particulate Monitor
Measure PM-10, PM-2.5 or PM-1 mass concentrations with the Thermo Scientific 5014i Beta Continuous Ambient Particulate Monitor.
Thermo Scientific 5014i Beta Continuous Ambient Particulate Monitor
Thermo Scientific 5014iQ Beta Attenuation Monitor
The Thermo Scientific 5014iQ Beta Attenuation Monitor is an automated ambient particulate measurement instrument utilizing beta attenuation.
Thermo Scientific 5014iQ Beta Attenuation Monitor
Thermo Scientific Model 5028i Continuous Particulate Monitor
Conduct simultaneous, real-time measurement of PM10 and PM2.5 with this unit that combines dual channel sampling technology and beta attenuation analysis on the proven iSeries platform.
Model 5028i Continuous Particulate Monitor
Chemiluminescence
When combined, certain gases produce high energy chemical reactions that emit light energy (photons), known as chemiluminescence.  Specifically, light emission results when electronically excited molecules decay to lower energy states. These emissions are detected by photomultiplier tubes and, by measuring the intensity and characteristics of the light emitted, the presence and concentration of various gases can be accurately determined.  Our analyzers that operate using this principle employ advanced optical technology for high sensitivity and reliable readings.  Explore them here:
concept of luminescence of particles
Thermo Scientific Model 17i Ammonia Analyzer
The Thermo Scientific Model 17i Ammonia Analyzer uses the light producing reaction of nitric oxide (NO) with ozone (O3) as its basic principle. The instrument has three modes of operation, NO, NOx and Nₜ. Mode 1. While operating in the NO mode, the sample is mixed with ozone in the reaction chamber. This reaction produces a characteristic luminescence with intensity proportional to the concentration of NO.
Thermo Scientific Model 17i Ammonia Analyzer
Thermo Scientific Model 42i (NO-NO₂-NOₓ) Analyzer
Measure levels of nitrogen oxide (NO-NO2-NOx) in the emissions from a source using chemiluminescent technology with the Thermo Scientific Model 42i NO-NO2-NOx Analyzer.
Thermo Scientific Model 42i (NO-NO2-NOx) Analyzer
Thermo Scientific 42iQ NO-NO₂-NOₓ Analyzer
The Thermo Scientific 42iQ NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from sub-ppb levels up to 100ppm. This analyzer is a single chamber, single photomultiplier tube design that cycles between the NO and NOx modes.
Thermo Scientific 42iQ NO-NO2-NOx Analyzer
Thermo Scientific Model 42i-HL High Level NO-NO₂-NOₓ Analyzer
Measure levels of nitrogen oxide (NO-NO2-NOx) in the emissions from a source using chemiluminescent technology with the Thermo Scientific Model 42i NO-NO2-NOx Analyzer.
Thermo Scientific Model 42i (NO-NO2-NOx) Analyzer
Thermo Scientific 42iQ High Level NO-NO₂-NOₓAnalyzer
Get 24/7 access to your analyzer’s information. The iQ Series Gas Analyzers are engineered to get smarter over time and help minimize troubleshooting time with a highly precise, yet tough solution. The 42iQ High Level NO-NO2-NOx Analyzer measures the amount of nitrogen oxides in the air from 10 ppm up to 5000 ppm.
Thermo Scientific 42iQ High Level NO-NO2-NOx Analyzer
Thermo Scientific 42iQ Low Source NO-NO₂-NOₓ Analyzer
Get 24/7 access to your analyzer’s information. The iQ Series Gas Analyzers are engineered to get smarter over time and help minimize troubleshooting time with a highly precise, yet tough solution. The Thermo Scientific 42iQ Low Source NO-NO2-NOx Analyzer provides the same level of performance when measuring low to mid-levels of nitrogen oxides in the air using chemiluminescent technology.
Thermo Scientific 42iQ Low Source NO-NO2-NOx Analyzer
Thermo Scientific Model 42i-Y NOᵧ Analyzer
Measure the amount of nitrogen oxides in the air from sub-ppb levels up to 1000 ppb using chemiluminescence with the Thermo Scientific Model 42i-Y NOY Analyzer. The Model 42i-Y is a single chamber, single photomultiplier tube design that measures NOY which includes most oxides of nitrogen except for NH3 and N2O. The 42i-Y complies with all USEPA NCore Network requirements for the measurement of NOy.
Thermo Scientific Model 42i-Y NOY Analyzer
Thermo Scientific Model 42i-TL TRACE Level NOₓ Analyzer
Measure the amount of nitrogen oxides in the air from sub-ppb levels up to 1000 ppb using chemiluminescence with the Thermo Scientific Model 42i-TL TRACE Level NOx Analyzer. The Model 42i-TL is a single chamber, single photomultiplier tube design that cycles between the NO, NOx, and Zero modes. The addition of the Zero mode provides for excellent long-term stability and extremely low minimum detectable limits. The 42i-TL has independent outputs for NO, NO2, and NOx and each can be calibrated independently.
Thermo Scientific Model 42i-TL TRACE Level NOx Analyzer
Thermo Scientific 42iQTL Trace Level NO-NO₂-NOₓ Analyzer
The Thermo Scientific 42iQ Trace Level NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from sub-ppb levels up to 1000ppb. The 42iQTL is a single chamber, single photomultiplier tube design that cycles between the NO, NOx, and Zero modes.
 Thermo Scientific 42iQTL Trace Level NO-NO2-NOx Analyzer
Gas Chromatography (GC)
Gas chromatography (GC) is a proven analytical tool that was initially developed in the 1950s and is now a widely applied technique for separating and analyzing compounds that can be vaporized without decomposition.  Because GC is best used to measure volatile compounds and utilizes gas columns that are stable and long lasting, GC is ideal for certain gas measurement applications.  Explore instruments that use GC here.
gas chromatographic columns
Thermo Scientific Direct Methane & Non-Methane Hydrocarbon Analyzer Model 55i
Make automated measurements of methane and non-methane hydrocarbons with the Thermo Scientific Direct Methane & Non-Methane Hydrocarbon Analyzer, Model 55i. Unlike instruments that measure only methane and total hydrocarbons, this back-flush gas chromatography (GC) system provides a direct measurement of non-methane concentrations.
Thermo Scientific Direct Methane & Non-Methane Hydrocarbon Analyzer Model 55i
Gas Filter Correlation (GFC)
Gas Filter Correlation (GFC) is a form of Nondispersive Infrared detection. Like NDIR, GFC operates on the principle that carbon monoxide (CO) absorbs infrared radiation at a wavelength of 4.6 microns. Filtering isolates the detected wavelength range to that of the gas target. Because infrared absorption is a non-linear measurement technique, it is necessary to transform the basic analyzer signal into a linear output. The analyzer uses an exact calibration curve to accurately linearize the instrument output over any range up to a concentration of 10,000ppm.
concept of infrared technology
Thermo Scientific 48iQ Carbon Monoxide Analyzer
The Thermo Scientific 48iQ Carbon Monoxide (CO) Analyzer utilizes gas filter correlation technology to measure the amount of carbon monoxide in the air. The 48iQ Gas Analyzer uses an exact calibration curve to accurately linearize the instrument output over any range up to a concentration of 10,000 ppm.
48iQ
Thermo Scientific 48iQTL Trace Level Carbon Monoxide Analyzer
The Thermo Scientific 48iQ Trace Level Carbon Monoxide (CO) Analyzer utilizes gas filter correlation technology to measure the amount of carbon monoxide in the air. The instrument employs pressure compensation and maintains constant temperature over the operating temperature range for stability. Standard features also include an intelligent zero reference to optimize drift performance and dynamic filtering to maintain data stability and integrity in changing environmental conditions. User settable alarm levels for concentration and a wide variety of internal diagnostics are available from an easy-to-follow touch screen display.
48iQTL
Thermo Scientific Model 48i CO Analyzer
The Thermo Scientific Model 48i Carbon Monoxide (CO) Analyzer utilizes gas filter correlation technology to measure the amount of carbon monoxide in the air. It is approved to meet the following standards: US EPA, UK Environmental Agency, and the EU Environmental Agency, and has ethernet connectivity for efficient remote access.
48I
Thermo Scientific Model 48i-TLE Enhanced Trace Level CO Analyzer
The Thermo Scientific Model 48i-TLE Analyzer is a single chamber, single photomultiplier tube design that measures only CO. Dual range and auto range are standard features in this instrument. Because the instrument is operated continuously in the CO mode, the response time is less than 10 seconds.
Thermo Scientific Model 48i-TLE Enhanced Trace Level CO Analyzer
Gas scrubbing
Gas scrubbing technology combines filtration, catalytic conversion, and oxidation to produce pollutant free air (Zero Air) from ambient air.  Zero Air is then used for instrument calibration and as diluent air supply for spanning ambient air analyzers.  Gas scrubbing technology removes NO, NO2, O3, SO2, CO, and hydrocarbons. Our gas scrubbing technology passes pressurized air into a column of Purafil (potassium permanganate on alumina) which oxidizes NO to NO2. From there the air passes through a column of activated charcoal which removes NO2, SO2, O3 and hydrocarbons. Lastly, the air is moved into the reactor where it is heated to 350°C over a catalytic surface which converts CO to CO2 and any remaining hydrocarbons, including methane, to water and CO2. This process results in a pollutant free stream of air.
Activated Carbon Air Filter
Thermo Scientific 111iQ Zero Air Supply
The Thermo Scientific 111iQ Zero Air Supply supplies pollutant-free air (zero air) from ambient air to allow for proper zeroing, and to provide clean diluent air for spanning ambient air analyzers.
111IQ Series Zero Air Supply
Non-Dispersive Infrared (NDIR) technology
The relative simplicity of NDIR technology provides precise, long-term gas analysis while lowering operating cost throughout the life cycle of the instrument. NDIR analyzers operate on the principle that gases absorb radiation in specific infrared wavelength ranges. As infrared light passes through a container of gas, a non-dispersive infrared sensor detects how much of the filtered light wavelength the gas absorbs. A measurement of gas concentrations is obtained. Thermo Scientific analyzers combine this technology with advanced optical filters to enable even more precise measurements.
Non-dispersive infrared technology
Thermo Scientific 410iQ Carbon Dioxide Gas Analyzer
The Thermo Scientific 410iQ Carbon Dioxide (CO2) Analyzer utilizes advanced Non-Dispersive Infrared (NDIR) optical filter technology to measure concentration of CO2. The 410iQ Standard Analyzer uses an internally stored calibration curve to accurately linearize the instrument output over any range up to a concentration of 10,000 ppm.
Thermo Scientific 410iQ Carbon Monoxide Analyzer
Thermo Scientific 410iQ High Level Carbon Dioxide Gas Analyzer
The Thermo Scientific 410iQ High Level Carbon Dioxide (CO2) Analyzer utilizes advanced Non-Dispersive Infrared (NDIR) optical filter technology to measure concentration of CO2. The 410iQ High Level Analyzer uses an internally stored calibration curve to accurately linearize the instrument output over any range up to a concentration of 25% by volume.
Thermo Scientific 410iQ High Level Carbon Dioxide Gas Analyzer
Optically Enhanced Fourier Transform Infrared (OE-FTIR)
Optically Enhanced Fourier Transform Infrared (OE-FTIR) using breakthrough StarBoost Technology enables commercial FTIR gas analysis that dramatically increases sensitivity, linearity and dynamic range over narrow spectral bands of interest. It utilizes specialized optics, electronics and analysis algorithms to go beyond traditional FTIR gas analysis capabilities.
This enhancement technology, proven in demanding applications such as ethylene oxide and formaldehyde measurement, enables users to achieve single-digit ppb detection limits for many applications. It can be supplied as a turnkey add-on to the Thermo Scientific MAX-iR Gas Analyzer and is compliant with several regulatory methods including US Environmental Protection Agency (EPA) method 320 and American Society for Testing and Materials (ASTM) D6348.  
Optical light spectrum. Rainbow gradient background. Electromagnetic visible color spectrum for human eye. Color scheme from infrared to ultraviolet. Vector illustration isolated on white background.
Thermo Scientific MAX-iR FTIR Gas Analyzers
Re-invented with detection limits in single-digit parts per billion for most applications, the Thermo Scientific MAX-iR FTIR Gas Analyzer provides accurate gas detection limits even in the presence of a wide range of interferences and produces results in real time.  With Thermo Scientific StarBoost Technology, it can now drive into mid parts per trillion, challenging the use of traditional technologies that are slower and more costly to operate.
Thermo Scientific iQ series of air monitoring equipment
Thermo Scientific MAX-iAQ Continuous Ambient Air Monitoring System
The Thermo Scientific MAX-iAQ Continuous Ambient Air Monitoring System is a factory-ready, fully-automated ambient air monitoring solution for multipoint low-level detection, even in high humidity environments. The fully automated system can quantify 10-100+ compounds from up to 20 sample locations using FTIR spectral analysis.
Thermo Scientific™ MAX-iAQ Continuous Ambient Air Monitoring System
Gravimetric
Gravimetric technology is a quantitative method used to determine concentration or amount of a substance by measuring its mass. Our Partisol samplers uses this technique to accurately measure particulate matter in the ambient air for air quality assessment. Partisol samplers are specially designed instruments that capture airborne particles on filter media for subsequent analysis. This technique is widely used in environmental monitoring and regulatory compliance to measure concentrations of particulate matter, which can have significant health and environmental impacts.
Magnifying glass looking at power plant
Key components and functionality
  1. Sampler unit: The partisol sampler consists of a housing that contains the sampling components, including a pump, flow controller, and filter holder. The unit is typically placed in a fixed outdoor location to continuously sample ambient air.
  2. Filter media: Air is drawn through a filter, usually made of materials like Teflon, quartz, or glass fiber, which captures particulate matter. The filter can be designed to collect different sizes of particles, such as PM10 (particles with a diameter of 10 micrometers or less) or PM2.5 (particles with a diameter of 2.5 micrometers or less).
  3. Flow control: The sampler includes a flow control system to ensure a consistent and accurate flow rate of air through the filter. This is crucial for obtaining reliable data on particle concentrations.
  4. Data logging: Modern partisol samplers are equipped with data logging capabilities to record sampling conditions, such as flow rate, temperature, and humidity, ensuring accurate and comprehensive data collection.
Sampling process
The partisol sampler is set up at the monitoring site, with the filter installed in the filter holder. The sampler is programmed to operate at a specific flow rate and sampling schedule. Air is then drawn through the filter over a specified period, typically 24 hours, capturing particulate matter on the filter surface. After the sampling period, the filter is removed and sent to a laboratory for analysis. In the laboratory, the filter is weighed to determine the mass of particulate matter collected. Additional analyses, such as chemical composition or elemental analysis, can be performed to identify the types of particles present.
Thermo Scientific Partisol 2000i Air Sampler
Meet regulatory ambient air particulate sampling requirements for PM-2.5, PM-10 and TSP with the manual Thermo Scientific Partisol 2000 i-series Air Samplers. Partisol 2000i is a single channel manual sampler for PM10, PM2.5 and has Federal Reference Methods (FRM) designation. Featuring enhanced communication capabilities and long-term unattended operation, the 2000i sampler provides reliable and quiet operation, low maintenance requirements and a single-action filter exchange mechanism for added convenience.
Partisol 2000i-D Dichotomous Air Sampler
Thermo Scientific Partisol 2000i-D Dichotomous Air Sampler
Meet regulatory ambient air particulate sampling requirements for PM-2.5, PM-10 and TSP with the manual Thermo Scientific Partisol 2000 i-series Air Samplers. Partisol 2000iD (Dichotomous) is a dual channel manual sampler for simultaneous sampling of fine and coarse particulate matter.  This sampler features USB ports for improved data downloads, and exchange 47mm diameter sample filters automatically, which can be preset to a user-defined time interval.
Partisol 2000i-D Dichotomous Air Sampler
Thermo Scientific Partisol 2025i Sequential Air Sampler
Partisol 2025i is a single channel manual sampler for PM10, PM2.5. Advanced Thermo Scientific Partisol 2025i Sequential Air Samplers are designed to store 16 filter cassettes, allowing for two weeks of unattended daily-sampling of particulate matter.  This sampler features USB ports for improved data downloads, and exchanges 47mm diameter sample filters automatically, which can be preset to a user-defined time interval.
Partisol 2025i-D Dichotomous Sequential Air Sampler
Thermo Scientific Partisol 2025i-D Dichotomous Sequential Air Sampler
Partisol 2025iD (Dichotomous) is a dual channel manual sampler for simultaneous sampling of fine and coarse particulate matter.  Advanced Thermo Scientific Partisol 2025i-D Dichotomous Sequential Air Samplers are designed to store 16 filter cassettes, allowing for two weeks of unattended daily-sampling of particulate matter.  This sampler features USB ports for improved data downloads, and exchanges 47mm diameter sample filters automatically, which can be preset to a user-defined time interval.
Partisol 2025i-D Dichotomous Sequential Air Sampler
Pulsed Fluorescence technology
Our Pulsed Fluorescence analyzers operate on the principle that H2S can be converted to SO2. As the SO2 molecules absorb ultraviolet (UV) light and become excited at one wavelength, the molecules then decay to a lower energy state emitting UV light at a different wavelength. The pulsing of the UV source lamp serves to increase the optical intensity and a greater UV energy throughput and lower detectable SO2 concentrations are realized.
Because this technology uses reflective bandpass filters, which are less subject to photochemical degradation and are more selective in wavelength isolation than transmission filters, increased detection specificity and long-term stability are achieved.
ultraviolet pattern
Thermo Scientific Model 450i Hydrogen Sulfide - Sulfur Dioxide Analyzer
The Thermo Scientific Model 450i Hydrogen Sulfide and Sulfur Dioxide Analyzer measure the levels of H2S and SO2 in ambient air. This analyzer features pulsed fluorescence technology and performs simultaneous measurement readings to provide reliable data capture on a constant basis.
Thermo Scientific iQ series of air monitoring equipment
Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer
The Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer utilizes pulsed fluorescence technology. The 450iQ analyzer operates on the principle that H2S can be converted to SO2. As the SO2 molecules absorb ultraviolet (UV) light and become excited at one wavelength, the molecules then decay to a lower energy state emitting UV light at a different wavelength. Reflective bandpass filters, as compared to commonly used transmission filters, are less subject to photochemical degradation and are more selective in wavelength isolation. This results in both increased detection specificity and long-term stability.
450iQ
Thermo Scientific Model 43i-HL High Level SO2 Analyzer
Measure high levels of SO2 from a source up to 10,000 ppm, the first gas analyzer to utilize pulsed fluorescence technology to measure SO2.
43i
Thermo Scientific 43iQ Sulfur Dioxide Analyzer
The Thermo Scientific 43iQ Sulfur Dioxide (SO2) Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air. The pulsing of the UV source lamp serves to increase the optical intensity whereby a greater UV energy throughput and lower detectable SO2 concentration are realized.
Thermo Scientific Mercury Freedom System
Thermo Scientific Enhanced Trace Level SO₂ Analyzer Model 43i-TLE
Measure the levels of SO2 in ambient air down to 50 ppt with the Thermo Scientific Enhanced Trace Level SO2 Analyzer Model 43i-TLE, a pulsed fluorescence gas analyzer known for superior accuracy, sensitivity, and reliability in trace-level ambient applications.
Thermo Scientific Enhanced Trace Level SO2 Analyzer Model 43i-TLE
Thermo Scientific 43iQ Trace Level Sulfur Dioxide Analyzer
The Thermo Scientific 43iQ Trace Level SO2 Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air to .05ppb.
Thermo Scientific 43iQ Trace Level Sulfur Dioxide Analyzer
Thermo Scientific Model 43i-HL High Level SO₂ Analyzer
Measure high levels of SO2 from a source up to 10,000 ppm with the Thermo Scientific Model 43i-HL SO2 Analyzer, the first gas analyzer to utilize pulsed fluorescence technology to measure SO2.
Thermo Scientific Model 43i-HL High Level SO2 Analyzer
Thermo Scientific 43iQ High Level Sulfur Dioxide Analyzer
The Thermo Scientific 43iQ High Level SO2 Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air up to 1000ppm in standard range and up to 10000ppm in extended range.
Thermo Scientific 43iQ High Level Sulfur Dioxide Analyzer
SHARP technology
Sharp Technology
Thermo Fisher Scientific SHARP technology couples light scattering measurement with beta attenuation to provide precise and accurate data. It starts with the proven method of using beta attenuation to determine the mass of the collected sample and adds a nephelometer to increase the overall performance. ​
​The nephelometer uses light scattering that is fast and precise in measuring the particle count, and the nephelometer count readings are converted to a “raw” mass concentration based on an internal algorithm and initial factory calibration using a standard aerosol. ​While this calibration allows the nephelometer to provide mass concentration results that are accurate for the specific calibration aerosol at the time of manufacture, the calibration aerosol may not represent the actual sampled aerosol which can result in a measurement bias compared to the true mass readings. ​To compensate for this potential bias, the SHARP compares the nephelometer concentration with the beta measured concentration and adjusts the nephelometer concentration to match the concentration reported using the beta attenuation technique. The adjustment is performed continuously and reported in real-time. This results in a reported SHARP value that has high accuracy and high precision. ​
Thermo Scientific 5030iQ SHARP Monitor
The Thermo Scientific 5030iQ SHARP (synchronized hybrid ambient real-time particulate) Monitor is a real-time particulate monitor that utilizes beta attenuation and light scattering design. This instrument offers exceptional ease of use and provides long-term unattended operation.
Thermo Scientific 5030iQ SHARP Monitor
Thermo Scientific Model 5030i SHARP Monitor
Combine the precision of light-scattering nephelometry with the accuracy of beta attenuation technology with the Thermo Scientific Model 5030i SHARP Monitor.
Thermo Scientific Model 5030i SHARP Monitor
Tapered element oscillating microbalances (TEOM)
Particulate matter monitoring systems that employ tapered element oscillating microbalances (TEOM) technology are “gravimetric” instruments that draw (then heat) ambient air through a filter at a constant flow rate, continuously weighing the filter and calculating near real-time mass concentrations of particulate matter. 
The TEOM monitor technique relies upon an exchangeable filter cartridge seated on the end of a hollow tapered tube. The wider end of the tube is fixed. As the air passes through the filter, particulate is deposited. The filtered air then passes through the tapered tube to a flow controller. The tapered tube with the filter on its end is maintained in oscillation in a clamped-free mode. The frequency of oscillation is dependent upon the physical characteristics of the tapered tube and the mass on its free end.
TEOM technology concept
As particulate deposits land on the filter, the filter mass change is detected as a frequency change in the oscillation of the tube. The mass of the particulate matter is thus determined inertially, i.e. directly. When this mass change is combined with the flow rate through the system, the monitor yields an accurate measurement of the particulate concentration in real time. The major advantage of this method is that any changes in aerosol characteristics will not influence the accuracy of the mass measurement. 
Thermo Scientific 1405 TEOM Continuous Ambient Particulate Monitor
Take continuous direct mass measurements of particulates utilizing a tapered element oscillating microbalance (TEOM) with the Thermo Scientific 1405 TEOM Continuous Ambient Particulate Monitor.
Thermo Scientific 1405 TEOM Continuous Ambient Particulate Monitor
Thermo Scientific 1405-D TEOM Continuous Dichotomous Ambient Particulate Monitor
Simultaneously measure PM Fine (PM-2.5), PM Coarse, and PM-10 mass concentrations with the Thermo Scientific TEOM 1405-D Continuous Ambient Particulate Monitor.
1405 TEOM-D 900x900 1405 TEOM-D
Thermo Scientific 1405-D 1405-F TEOM Continuous Ambient Air Monitor
Take continuous direct mass measurements of PM-10, PM-2.5, and PM-1 particulates using the Thermo Scientific 1405-F TEOM Continuous Ambient Air Monitor.
Thermo Scientific 1405-D 1405-F TEOM Continuous Ambient Air Monitor
Thermo Scientific 1405-DF TEOM Continuous Dichotomous Ambient Air Monitor
Take continuous direct mass measurements of particulate with excellent short term precision using the Thermo Scientific 1405-DF TEOM Continuous Dichotomous Ambient Air Monitor.
Thermo Scientific 1405-DF TEOM Continuous Dichotomous Ambient Air Monitor
UV photometry
UV photometric gas analyzers take advantage of the fact that certain gases exhibit a pronounced absorption band in the spectral range of 200nm to 400nm. Because of the high absorption behavior of the gases in specific bands, analyzers that use UV photometry can reliably detect very low concentrations of target gases.  Additionally, results using this method are resistant to interference from the presence of water vapor and carbon dioxide. This technology is also advantageous in that additional optical spectrometers or filter elements are not required.
UV Photometry
Explore analyzers that use UV photometry:
Thermo Scientific Model 49i Ozone Analyzer
Measure the amount of ozone in ambient air with the dual-cell, UV photometric Thermo Scientific Model 49i Ozone Analyzer.
Thermo Scientific Model 49i Ozone Analyzer
Thermo Scientific 49iQ Ozone Analyzer
The Thermo Scientific 49iQ Ozone (O3) Analyzer utilizes UV Photometric technology to measure the amount of ozone in the air from ppb levels up to 200ppm. The Thermo Scientific 49iQ Analyzer is a dual cell photometer, the concept adopted by NIST for the national ozone standard. Because the instrument has both sample and reference flowing at the same time, a response time of 20 seconds can be achieved.
Thermo Scientific GS Omni Analyzer
Thermo Scientific Model 49i-PS Ozone Primary Standard
Produce ozone concentrations of up to 1000 ppb with the Thermo Scientific Model 49i-PS Ozone Primary Standard, a UV photometric primary standard suitable for calibration of ozone analyzers and transfer standards. The Model 49i-PS uses a dual-cell photometer, the concept adopted by NIST as the principle technology for the national ozone standard.
Thermo Scientific Model 49i-PS Ozone Primary Standard
Service and product availability may vary by country and are subject to varying regulatory requirements. Please contact your local sales representative for availability.