Cell culture red cells
Advance your research with proven cell culture instruments and equipment, including CO2 incubators, biological safety cabinets, general purpose centrifuges, lab refrigerators and freezers, automated cell counters, and cell imaging systems.
CO2 incubators provide optimal conditions for cell growth, controlling temperature, humidity and CO2 levels (O2 control optional). Uniformity and recovery of parameters and contamination control are important buying considerations.
Biological safety cabinets (BSC) precisely control the airflow of the work area and are essential in protecting your samples as well as for the safety of lab personnel. Safety and ergonomics are factors to consider when buying a BSC.
Centrifuges are used in the harvesting workflow step to separate or concentrate cells or cell components. Precise temperature control and accurate centrifugal speeds are critical for this process.
Our cell culture equipment helps you grow viable cell lines while preventing contamination. Learn more
Experience the convenience and power of our automated cell counters designed to meet the needs of any lab. Each compact instrument offers advanced algorithms to allow you to quickly and accurately count cells from the bench.
Thermo Scientific Precision General Purpose Water Baths offer enhanced temperature uniformity and stability and are easy to clean and maintain. The water baths can be used with water or thermal beads to prevent from contamination.
Thermo Scientific Cold Storage Solutions include refrigerators to store your medium, controlled-rate freezers to freeze down cells and ULT freezers as well as cryopreservation systems to store cells.
Thermo Scientific water purification systems offer a wide selection of purification methods, capabilities and options. For more than 130 years, Thermo Scientific lab water systems have been a trusted resource for science.
Powerful EVOS digital microscopes allow you to capture publication-quality images and data with just a few clicks. The EVOS line of products are versatile, compact and ideal for a broad range of imaging applications.
The Attune Nxt Flow Cytometer is compact, benchtop cell analyser that can be configured with up to 4 spatially separated lasers to flexibly design, run, and analyse multicolour panels of up to 14 colors.
Cell Culture Workflow
cell-culture-workflow
From sample preparation to storage, explore the steps and all products we can support you with for your cell culture application.
Resources
Find the information you need for successful cell culture—including application notes, videos & webinars, and the Gibco Education virtual labs—in the Cell Culture & Transfection Learning Center.
Support
Find tips, troubleshooting help, and resources for your cell culture workflow in the Cell Culture & Transfection Support Center
Style Sheet for Global Design System