Heat Stable Recombinant Proteins FGF2 and FGF10
Engineered for greater stability & efficiency
Heat-stable recombinant proteins have been engineered for greater stability in cell culture, exhibiting enhanced bioactivity at 37°C. Because Heat Stable proteins are bioactive longer than native proteins in standard cell culture conditions, researchers may be able to use less to maintain the same growth rates, therefore improving the efficiency of their cell culture applications.
  • Superior performance—retains bioactivity without the use of artificially high concentrations
  • Improved efficiency—the stable bioactivity means you won’t have to supplement with additional protein (a necessary step when using native proteins), and you’ll save money in reagent costs over the longer term
  • Easy to use—direct replacement for your current proteins allowing for easy substitution in protocols
  • Heat Stable Proteins availableHeat Stable bFGF and Heat Stable FGF-10.
Superior cell culture performance
Heat Stable FGFs maintain activity at 37°C
Graph of proliferation vs dose for HS bFGF
Heat Stable FGF10 proliferation response at 4 degrees C and 37 degrees C
Graph of proliferation vs dose for native bFGF
Native FGF10 proliferation response at 4 degrees C and 37 degrees C
Figure 1. Cell proliferation studies demonstrate Heat Stable FGFs exhibit superior activity compared to native FGFs. HS bFGF activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation (top panels). The 10 ng/mL HS bFGF (top left) and native bFGF (top right) solutions were stored at 4°C (unstressed) or 37°C (heat stressed) for 72 hours before being used to treat the Balb/3T3 cells in a dose-dependent manner. Likewise, HS FGF10 activity was measured with PrestoBlue HS Cell Viability reagent analysis of MCF7 cell proliferation (bottom panels). The 1 μg/mL HS FGF10 (bottom left) and native FGF10 (bottom right) solutions were stored at 4°C (unstressed) or 37°C (heat stressed) for 72 hours before being used to treat the MCF7 cells in a dose-dependent manner.
Over 80% sustained biological activity
bar chart of proliferation for HS bFGF and native bFGF
Heat Stable FGF10 vs Native FGF10 after 72 hours of heat stress
Figure 2. Heat Stable FGF growth factors demonstrate greater activity after 72 hours of heat stress than native proteins. (Left) HS bFGF activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation. (Right) HS FGF10 activity was measured with PrestoBlue HS Cell Viability reagent analysis of MCF7 cell proliferation. The percent activity is the amount of activity maintained after heat stress relative to the same solution stored at 4°C (unstressed). The 10 ng/mL HS bFGF and native bFGF, and 1 μg/mL HS FGF10 and native FGF10 solutions were stored at 4°C or 37°C for 72 hours before being used to treat Balb/3T3 or MCF7 cells, respectively.
line graph of 2 sources of bFGF following 2 treatments
Figure 3. Cell proliferation assay demonstrating how biological activity of tag-free heat stable PeproTech bFGF protein is maintained longer under cell culture conditions compared to the equivalent native bFGF protein.
HS bFGF offers increased efficiency
Bar chart of NSC doubling time of cultured NSCs when grown in native bFGF and Gibco heat stable bFGF
HS FGF10 performed equivalently to native FGF10
Microscopic images of cultured NSCs when grown in native bFGF and Gibco heat stable bFGF
Figure 4. Heat Stable FGFs allows normal proliferation of cells with growth factor.
(A) 5 ng/mL of bFGF leads to significantly slower neural stem cell (NSC) proliferations as compared with 5 ng/mL HS bFGF, which maintains a proliferation rate equivalent to that of 20 ng/mL native bFGF. Mean +/- SEM. ** = p < 0.01. (B) Additionally, while 5 ng/mL native bFGF leads to neurite outgrowth (denoted by red arrows), HS bFGF maintains multipotent NSC morphology. (C) Relative activity of FGF10 was measured following incubation of 1 μg/mL HS FGF10 and native FGF10 stock solutions at 4°C or 37°C for 72 hours before being used to treat MCF7 cells. Activity measured with PrestoBlue HS Cell Viability reagent demonstrated 10 ng/mL of HS FGF10 performed equivalently to 100 ng/mL of native FGF10. Mean +/- SEM. *** = p < 0.0001; n.s. = not significant.
HS bFGF maintains neural stem cell multipotency
Phase-contrast and immunostained images of NSCs cultured in Gibco HS bFGF
Figure 5. Heat Stable bFGF maintains the morphology and SOX1 expression of multipotent NSCs. Phase contrast (left) and immunostained (right) photomicrographs of NSCs cultured in the presence of HS bFGF (top row) and native bFGF (bottom row). Immunostaining for NSC multipotency marker SOX1 (red) with nuclear (DAPI) counterstain (blue).
HS bFGF does not interfere with undirected differentiation
Immunostained images of NSCs cultured in Gibco HS bFGF showing expected differentiation markers
Figure 6. Spontaneous differentiation supported by HS bFGF. Primary rat NSC isolated from Sprague Dawley E14 cortex cultured for 3 passages in DMEM/F12 + GlutaMAX supplemented with N2, NEAA, β-mercaptoethanol and 10 ng/mL HS bFGF. Media changes made every 48 hours without daily spiking of bFGF. At passage 3, cells were allowed to spontaneously differentiate by culturing in growth medium without HS bFGF. Cells showed equivalent potential to undergo undirected differentiation as compared to native bFGF, measured by markers MAP2 (neuron), GFAP (astrocytes) and GALC (oligodendrocytes).
HS bFGF enhances growth of cancer spheroids via a more streamlined protocol
Brightfield microscopy image of cancer spheroids grown with Gibco HS bFGF
Figure 7. HS bFGF enhances growth of cancer spheroids without the need for troublesome media changes. Spheroids were formed in Nunclon Sphera microplates in serum-free medium containing no bFGF, 10 ng/mL native bFGF, or 10 ng/mL HS bFGF. After 8 days in culture without media changes, the cells were stained via live (green)-dead (red) assay and DAPI counterstain (blue). All spheroids were viable, but the spheroid exposed to HS bFGF was the largest of the three experimental conditions.  For more information, please see our application note
Gene expression of cells treated with HS bFGF is comparable to that of cells treated with native bFGF
Figure 8. bFGF-target and off-target gene expression was comparable in cells treated with native bFGF and Heat Stable bFGF. The TaqMan Array Human Signal Transduction Pathways array (Cat. No. 4418775) and the TaqMan Array Human FGF Pathway (Cat. No. 4414136) were used to assess expression of genes in the androgen, calcium, CREB, estrogen, hedgehog, insulin, JAK-STAT, LDL, mitogenic, NFAT, NFkB, p53, phospholipase C, protein kinase C, retinoic acid, stress, survival, TGF-β, Wnt, and FGF-associated pathways, respectively. Expression was assessed using human induced pluripotent stem cells treated with 10 ng/mL native bFGF or Heat Stable bFGF. Genes from both arrays were separated into ‘bFGF-target’ or ‘off-target’ based on their downstream relationship to bFGF, as is currently indicated by the literature. The bFGF-target and off-target panels show the results from 43 and 71 genes, respectively.
HS bFGF outperforms top competitors
Bar chart of bFGF activity from Gibco HS bFGF and three competitor products
Figure 9. HS bFGF exhibits superior activity after extended 37°C incubation compared to competitor’s products. Activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation. The percent activity is amount of activity maintained after heat stress (37°C), relative to the same solution stored at 4°C (unstressed). The 10 ng/mL HS bFGF and competitor bFGF solutions were stored at 4°C or 37°C for 72 hours before being used to treat the Balb/3T3 cells, N≥3. Mean ± SEM.
HS bFGF facilitates stem cell culture workflows
4-panel microscopy image showing iPSCs cultured with Gibco HS bFGF
Figure 10. Comparison iPSCs cultured in the presence of Gibco HS bFGF or StemBeads reagent. Microscopic images of StemBeads reagent (10 ng/mL) captured at (A) 10x and (B) 20x magnification. Because StemBeads beads vary in size (arrows indicate different sized StemBeads particles), they can be difficult to distinguish from cells and also from debris. Twenty-four hours after seeding human iPSCs in the presence of either (C) 10 ng/mL Gibco HS bFGF or (D) 10 ng/mL StemBeads reagent, phase-contrast images were captured of the cultured cells. The image of cells cultured with HS bFGF is clear, containing minimal debris, whereas the StemBeads reagent–containing culture shows more apparent debris. In addition because the beads tend to sink relatively quickly in solution, accurately measuring StemBeads reagent concentration can present a challenge.
Heat Stable FGFs maintain activity at 37°C
Graph of proliferation vs dose for HS bFGF
Heat Stable FGF10 proliferation response at 4 degrees C and 37 degrees C
Graph of proliferation vs dose for native bFGF
Native FGF10 proliferation response at 4 degrees C and 37 degrees C
Figure 1. Cell proliferation studies demonstrate Heat Stable FGFs exhibit superior activity compared to native FGFs. HS bFGF activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation (top panels). The 10 ng/mL HS bFGF (top left) and native bFGF (top right) solutions were stored at 4°C (unstressed) or 37°C (heat stressed) for 72 hours before being used to treat the Balb/3T3 cells in a dose-dependent manner. Likewise, HS FGF10 activity was measured with PrestoBlue HS Cell Viability reagent analysis of MCF7 cell proliferation (bottom panels). The 1 μg/mL HS FGF10 (bottom left) and native FGF10 (bottom right) solutions were stored at 4°C (unstressed) or 37°C (heat stressed) for 72 hours before being used to treat the MCF7 cells in a dose-dependent manner.
Over 80% sustained biological activity
bar chart of proliferation for HS bFGF and native bFGF
Heat Stable FGF10 vs Native FGF10 after 72 hours of heat stress
Figure 2. Heat Stable FGF growth factors demonstrate greater activity after 72 hours of heat stress than native proteins. (Left) HS bFGF activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation. (Right) HS FGF10 activity was measured with PrestoBlue HS Cell Viability reagent analysis of MCF7 cell proliferation. The percent activity is the amount of activity maintained after heat stress relative to the same solution stored at 4°C (unstressed). The 10 ng/mL HS bFGF and native bFGF, and 1 μg/mL HS FGF10 and native FGF10 solutions were stored at 4°C or 37°C for 72 hours before being used to treat Balb/3T3 or MCF7 cells, respectively.
line graph of 2 sources of bFGF following 2 treatments
Figure 3. Cell proliferation assay demonstrating how biological activity of tag-free heat stable PeproTech bFGF protein is maintained longer under cell culture conditions compared to the equivalent native bFGF protein.
HS bFGF offers increased efficiency
Bar chart of NSC doubling time of cultured NSCs when grown in native bFGF and Gibco heat stable bFGF
HS FGF10 performed equivalently to native FGF10
Microscopic images of cultured NSCs when grown in native bFGF and Gibco heat stable bFGF
Figure 4. Heat Stable FGFs allows normal proliferation of cells with growth factor.
(A) 5 ng/mL of bFGF leads to significantly slower neural stem cell (NSC) proliferations as compared with 5 ng/mL HS bFGF, which maintains a proliferation rate equivalent to that of 20 ng/mL native bFGF. Mean +/- SEM. ** = p < 0.01. (B) Additionally, while 5 ng/mL native bFGF leads to neurite outgrowth (denoted by red arrows), HS bFGF maintains multipotent NSC morphology. (C) Relative activity of FGF10 was measured following incubation of 1 μg/mL HS FGF10 and native FGF10 stock solutions at 4°C or 37°C for 72 hours before being used to treat MCF7 cells. Activity measured with PrestoBlue HS Cell Viability reagent demonstrated 10 ng/mL of HS FGF10 performed equivalently to 100 ng/mL of native FGF10. Mean +/- SEM. *** = p < 0.0001; n.s. = not significant.
HS bFGF maintains neural stem cell multipotency
Phase-contrast and immunostained images of NSCs cultured in Gibco HS bFGF
Figure 5. Heat Stable bFGF maintains the morphology and SOX1 expression of multipotent NSCs. Phase contrast (left) and immunostained (right) photomicrographs of NSCs cultured in the presence of HS bFGF (top row) and native bFGF (bottom row). Immunostaining for NSC multipotency marker SOX1 (red) with nuclear (DAPI) counterstain (blue).
HS bFGF does not interfere with undirected differentiation
Immunostained images of NSCs cultured in Gibco HS bFGF showing expected differentiation markers
Figure 6. Spontaneous differentiation supported by HS bFGF. Primary rat NSC isolated from Sprague Dawley E14 cortex cultured for 3 passages in DMEM/F12 + GlutaMAX supplemented with N2, NEAA, β-mercaptoethanol and 10 ng/mL HS bFGF. Media changes made every 48 hours without daily spiking of bFGF. At passage 3, cells were allowed to spontaneously differentiate by culturing in growth medium without HS bFGF. Cells showed equivalent potential to undergo undirected differentiation as compared to native bFGF, measured by markers MAP2 (neuron), GFAP (astrocytes) and GALC (oligodendrocytes).
HS bFGF enhances growth of cancer spheroids via a more streamlined protocol
Brightfield microscopy image of cancer spheroids grown with Gibco HS bFGF
Figure 7. HS bFGF enhances growth of cancer spheroids without the need for troublesome media changes. Spheroids were formed in Nunclon Sphera microplates in serum-free medium containing no bFGF, 10 ng/mL native bFGF, or 10 ng/mL HS bFGF. After 8 days in culture without media changes, the cells were stained via live (green)-dead (red) assay and DAPI counterstain (blue). All spheroids were viable, but the spheroid exposed to HS bFGF was the largest of the three experimental conditions.  For more information, please see our application note
Gene expression of cells treated with HS bFGF is comparable to that of cells treated with native bFGF
Figure 8. bFGF-target and off-target gene expression was comparable in cells treated with native bFGF and Heat Stable bFGF. The TaqMan Array Human Signal Transduction Pathways array (Cat. No. 4418775) and the TaqMan Array Human FGF Pathway (Cat. No. 4414136) were used to assess expression of genes in the androgen, calcium, CREB, estrogen, hedgehog, insulin, JAK-STAT, LDL, mitogenic, NFAT, NFkB, p53, phospholipase C, protein kinase C, retinoic acid, stress, survival, TGF-β, Wnt, and FGF-associated pathways, respectively. Expression was assessed using human induced pluripotent stem cells treated with 10 ng/mL native bFGF or Heat Stable bFGF. Genes from both arrays were separated into ‘bFGF-target’ or ‘off-target’ based on their downstream relationship to bFGF, as is currently indicated by the literature. The bFGF-target and off-target panels show the results from 43 and 71 genes, respectively.
HS bFGF outperforms top competitors
Bar chart of bFGF activity from Gibco HS bFGF and three competitor products
Figure 9. HS bFGF exhibits superior activity after extended 37°C incubation compared to competitor’s products. Activity was measured with PrestoBlue dye analysis of Balb/3T3 cell proliferation. The percent activity is amount of activity maintained after heat stress (37°C), relative to the same solution stored at 4°C (unstressed). The 10 ng/mL HS bFGF and competitor bFGF solutions were stored at 4°C or 37°C for 72 hours before being used to treat the Balb/3T3 cells, N≥3. Mean ± SEM.
HS bFGF facilitates stem cell culture workflows
4-panel microscopy image showing iPSCs cultured with Gibco HS bFGF
Figure 10. Comparison iPSCs cultured in the presence of Gibco HS bFGF or StemBeads reagent. Microscopic images of StemBeads reagent (10 ng/mL) captured at (A) 10x and (B) 20x magnification. Because StemBeads beads vary in size (arrows indicate different sized StemBeads particles), they can be difficult to distinguish from cells and also from debris. Twenty-four hours after seeding human iPSCs in the presence of either (C) 10 ng/mL Gibco HS bFGF or (D) 10 ng/mL StemBeads reagent, phase-contrast images were captured of the cultured cells. The image of cells cultured with HS bFGF is clear, containing minimal debris, whereas the StemBeads reagent–containing culture shows more apparent debris. In addition because the beads tend to sink relatively quickly in solution, accurately measuring StemBeads reagent concentration can present a challenge.
Ordering information
Catalog # Name Size Price (USD) Qty
PHG0367 Human Heat Stable bFGF Recombinant Protein 5 µg
107.65
Online Exclusive
116.00
Save 8.35 (7%)
PHG0368 Human Heat Stable bFGF Recombinant Protein 50 µg
396.65
Online Exclusive
431.00
Save 34.35 (8%)
PHG0369 Human Heat Stable bFGF Recombinant Protein 100 µg
489.65
Online Exclusive
532.00
Save 42.35 (8%)
PHG0360 Human Heat Stable bFGF Recombinant Protein 500 µg
858.60
Special offer
Online exclusive
Ends: 16-Jun-2025
954.00
Save 95.40 (10%)
PHG0371 Heat Stable FGF-10 Recombinant Protein 5 µg
104.00
PHG0372 Heat Stable FGF-10 Recombinant Protein 50 µg
456.00
PHG0373 Heat Stable FGF-10 Recombinant Protein 100 µg
782.00
PHG0374 Heat Stable FGF-10 Recombinant Protein 500 µg
2,345.00
PHG0375 Heat Stable FGF-10 Recombinant Protein 1 mg
3,255.00
100-18BHS-100UG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 100 µg
457.20
Special offer
Online exclusive
Ends: 16-Jun-2025
508.00
Save 50.80 (10%)
100-18BHS-10UG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 10 µg
158.40
Special offer
Online exclusive
Ends: 16-Jun-2025
176.00
Save 17.60 (10%)
100-18BHS-1MG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 1 mg
1,466.00
100-18BHS-250UG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 250 µg
582.00
100-18BHS-500UG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 500 µg
916.00
100-18BHS-50UG Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech® 50 µg
371.70
Special offer
Online exclusive
Ends: 16-Jun-2025
413.00
Save 41.30 (10%)
FAQs
QuestionAnswer
What is the difference between the Heat Stable bFGF products?
A His tag is present on products with catalog numbers of PHG0367, PHG0368, PHG0369, and PHG0360.  
No His tag is present on products with catalog numbers of 100-18BHS-100UG, 100-18BHS-10UG, 100-18BHS-1MG, 100-18BHS-250UG, 100-18BHS-500UG, and 100-18BHS-50UG.
Is Heat Stable Recombinant Human bFGF full length?Yes, Heat Stable Recombinant Human bFGF is full-length at 155 amino acids, plus 20 amino acids for N-terminal tag for the his-tagged versions of the protein. See catalog numbers in the FAQ above.
Can Heat Stable Recombinant Human bFGF be used in place of native recombinant human bFGF?Yes, Heat Stable Recombinant Human bFGF can be used in place of native human bFGF. It is a direct replacement, offering greater stability in cell culture conditions.
Has protein engineering altered the activity of Heat Stable Recombinant Human bFGF?HS bFGF does not exhibit higher bioactivity, nor does it stimulate cell signaling differently than native bFGF.
Why should Heat Stable Recombinant Human bFGF be used instead of native recombinant human bFGF?Native human bFGF degrades at standard culture conditions (i.e., 37°C). This means that more frequent media changes and/or more protein is required to maintain biological activity. Heat Stable Recombinant Human bFGF retains bioactivity minimizing fluctuations to more closely mimic physiological conditions for cells.
See all FAQs for heat-stable recombinant proteins
Resources
Recombinant proteins learning center
Learn what recombinant proteins are, what they are used for, and what types are available
About PeproTech proteins
Learn about the PeproTech brand of recombinant proteins
Gibco cell culture basics
Learn about all aspects of cell culture
3D cell culture protocols
See which recombinant proteins are used to generate organoids
Western transfer protocols and recipes
Procedures and buffers for performing western blots
Cytokine and growth factor signaling pathways
Explore the role of proteins and cytokines in various signaling pathways
Support
Recombinant Proteins Support Center
Find Getting Started guides, protocols, troubleshooting, and other resources
Recombinant proteins FAQs
Search the database to find answers to your questions
Contact general support
Get technical or customer support for any recombinant protein; find SDS, CoA, and phone numbers based on a specific product
Contact GMP support
Get in touch with GMP technical and regulatory experts