Obtaining high quality, intact RNA is the first and often most critical step in performing gene expression analyses. To isolate high quality RNA, tissue has to be either processed immediately after harvest, snap-frozen, or stabilized in an intermediary solution to preserve RNA integrity and allow for storage. Several products designed specifically to stabilize and preserve the integrity of RNA are available from point of collection to PCR and analysis.
What are RNA stabilization reagents?
RNA stabilization reagents (also known as RNA stabilizers) are solutions that help stabilize and preserve the integrity of RNA. Because RNA is known to be readily degraded by free-floating RNases, RNA stabilization reagents are used to prolong RNA in samples. RNA stabilizers can be introduced to RNA samples from fresh cell culture, fresh tissue, frozen tissue, blood, or purified RNA.
Learn more about working with RNA
Learn more about RNase control
Select RNA stabilizers based on RNA sample type
RNA stabilizing solution: RNAlater solution for stabilizing fresh cells
RNAlater Stabilization Solution product photo
RNAlater solution is an aqueous, non-toxic fresh cell and tissue storage reagent that rapidly permeates tissue and cells to stabilize and protect cellular RNA. This reagent helps minimize the need to immediately freeze samples in order to preserve RNA.
How does RNAlater RNA stabilizing solution work? Once saturated in RNAlater solution, samples can be tested and kept for continued storage. Fresh tissue and cells treated with RNAlater solution and archived at –80°C can be subjected to ten freeze/thaw cycles without affecting the quality of subsequently isolated RNA.
Sample types RNAlater solution have been tested on include:
  • Animal tissue
  • Cells
  • Tissue culture cells
  • E. coli
  • Drosophila
  • White blood cells
  • Plant tissues
Learn more about RNAlater
Video: How to stabilize RNA in fresh specimens
Video Player is loading.
Current Time 0:00
Duration 1:51
Loaded: 8.93%
Stream Type LIVE
Remaining Time 1:51
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how to quickly and easily stabilize and protect RNA in fresh specimens. RNAlater Stabilization Solution eliminates the need to immediately process samples, freeze them in liquid nitrogen, or rush them to the freezer.
    Ordering information for RNAlater RNA stabilizing solutions
    Catalog # Name Size Price (USD) Qty
    AM7020 RNAlater™ Stabilization Solution, 100 mL Each
    206.65
    Online Exclusive
    215.00
    Save 8.35 (4%)
    AM7021 RNAlater™ Stabilization Solution, 500 mL Each
    641.65
    Online Exclusive
    660.00
    Save 18.35 (3%)
    AM7024 RNAlater™ Stabilization Solution, 1 x 250 mL Each
    458.65
    Online Exclusive
    481.00
    Save 22.35 (5%)
    AM7023 RNAlater™ Stabilization Solution, 20 x 5 mL Each
    388.65
    Online Exclusive
    407.00
    Save 18.35 (5%)
    AM7022 RNAlater™ Stabilization Solution, 50 x 1.5 mL Each
    375.65
    Online Exclusive
    387.00
    Save 11.35 (3%)
    RNA stabilizing solution: RNAlater-ICE for stabilizing frozen tissue
    RNAlater-ICE Frozen Tissue Transition Solution product photo
    RNAlater-ICE solution is an RNA stabilizing solution for frozen tissue. This reagent prevents RNA degradation during tissue thawing. Once treated with RNAlater-ICE solution, tissue can be safely stored at 4°C or at room temperature (for a limited period of time) and can be further processed while protecting RNA integrity.
    Sample types RNAlater-ICE solution have been tested on include:
    • Brain
    • Heart
    • Kidney
    • Spleen
    • Liver
    • Pancreas
    • Lungs
    • Thymus
    Ordering information for RNAlater-ICE solution
    Catalog # Name Size Price (USD) Qty
    AM7030 RNAlater™-ICE Frozen Tissue Transition Solution, 25 mL Each
    146.65
    Online Exclusive
    154.00
    Save 7.35 (5%)
    LeukoLOCK system and Tempus tube kits for blood stabilization
    Blood samples are notoriously difficult to deal with because of the blood cell lysis step and the subsequent high presence of globin mRNA in RNA prepared from whole blood. Globin mRNA interferes with downstream expression assays. We have options for stabilizing RNA in blood samples.
    LeukoLOCK Total RNA Isolation System product photo
    LeukoLOCK Total RNA Isolation System contains globin-free mRNA from RNAlater–stabilized leukocytes. Using this system, anticoagulated blood is filter-fractionated, keeping only the leukocytes, and stabilized with RNAlater solution for storage. By excluding red blood cells, this system isolates and purifies RNA from blood without the presence of globin mRNA.
    Tempus Blood RNA Tubes are available for the stabilization and purification of total RNA from whole blood at the point of collection for downstream assays. Each Tempus tube contains a stabilizing and lysing reagent that eliminates the need for subsequent red blood cell lysis or proteinase K treatment. The RNA stabilizing reagent inactivates RNases and selectively precipitates RNA. Genomic DNA can also be isolated from the supernatant of the blood samples stored in a Tempus tube once the RNA is pelleted.
    Ordering information for LeukoLOCK system and Tempus tube kits
    Catalog # Name Size Price (USD) Qty
    AM1923 LeukoLOCK™ Total RNA Isolation System Each
    1,140.00
    4342792 Tempus™ Blood RNA Tube Each
    503.65
    Online Exclusive
    532.00
    Save 28.35 (5%)
    The last step in many nucleic acid isolation protocols—and the first step of many downstream assays—is to resuspend the RNA pellet. After isolation and purification of RNA, it is essential to suspend and store RNA in an appropriate, nuclease-free solution.
    THE RNA Storage Solution is an RNA stabilizing solution that is compatible with many downstream assays, such as:
    • RT-PCR
    • In vitro transcription
    • Northern analysis
    • Nuclease protection assays
    It is a 1 mM sodium citrate, pH 6.5 ± 0.1 buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This solution is prepared from pure agents, under scrupulously monitored conditions in our state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to help ensure their purity and performance.
    Ordering information for THE RNA storage solution
    Catalog # Name Size Price (USD) Qty
    AM7000 THE RNA Storage Solution, 10 x 1 mL Each
    67.65
    Online Exclusive
    72.00
    Save 4.35 (6%)
    AM7001 THE RNA Storage Solution, 50 mL Each
    67.65
    Online Exclusive
    72.50
    Save 4.85 (7%)
    RNA stabilizing solution: RNAlater solution for stabilizing fresh cells
    RNAlater Stabilization Solution product photo
    RNAlater solution is an aqueous, non-toxic fresh cell and tissue storage reagent that rapidly permeates tissue and cells to stabilize and protect cellular RNA. This reagent helps minimize the need to immediately freeze samples in order to preserve RNA.
    How does RNAlater RNA stabilizing solution work? Once saturated in RNAlater solution, samples can be tested and kept for continued storage. Fresh tissue and cells treated with RNAlater solution and archived at –80°C can be subjected to ten freeze/thaw cycles without affecting the quality of subsequently isolated RNA.
    Sample types RNAlater solution have been tested on include:
    • Animal tissue
    • Cells
    • Tissue culture cells
    • E. coli
    • Drosophila
    • White blood cells
    • Plant tissues
    Learn more about RNAlater
    Video: How to stabilize RNA in fresh specimens
    Video Player is loading.
    Current Time 0:00
    Duration 1:51
    Loaded: 8.93%
    Stream Type LIVE
    Remaining Time 1:51
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Learn how to quickly and easily stabilize and protect RNA in fresh specimens. RNAlater Stabilization Solution eliminates the need to immediately process samples, freeze them in liquid nitrogen, or rush them to the freezer.
      Ordering information for RNAlater RNA stabilizing solutions
      Catalog # Name Size Price (USD) Qty
      AM7020 RNAlater™ Stabilization Solution, 100 mL Each
      206.65
      Online Exclusive
      215.00
      Save 8.35 (4%)
      AM7021 RNAlater™ Stabilization Solution, 500 mL Each
      641.65
      Online Exclusive
      660.00
      Save 18.35 (3%)
      AM7024 RNAlater™ Stabilization Solution, 1 x 250 mL Each
      458.65
      Online Exclusive
      481.00
      Save 22.35 (5%)
      AM7023 RNAlater™ Stabilization Solution, 20 x 5 mL Each
      388.65
      Online Exclusive
      407.00
      Save 18.35 (5%)
      AM7022 RNAlater™ Stabilization Solution, 50 x 1.5 mL Each
      375.65
      Online Exclusive
      387.00
      Save 11.35 (3%)
      RNA stabilizing solution: RNAlater-ICE for stabilizing frozen tissue
      RNAlater-ICE Frozen Tissue Transition Solution product photo
      RNAlater-ICE solution is an RNA stabilizing solution for frozen tissue. This reagent prevents RNA degradation during tissue thawing. Once treated with RNAlater-ICE solution, tissue can be safely stored at 4°C or at room temperature (for a limited period of time) and can be further processed while protecting RNA integrity.
      Sample types RNAlater-ICE solution have been tested on include:
      • Brain
      • Heart
      • Kidney
      • Spleen
      • Liver
      • Pancreas
      • Lungs
      • Thymus
      Ordering information for RNAlater-ICE solution
      Catalog # Name Size Price (USD) Qty
      AM7030 RNAlater™-ICE Frozen Tissue Transition Solution, 25 mL Each
      146.65
      Online Exclusive
      154.00
      Save 7.35 (5%)
      LeukoLOCK system and Tempus tube kits for blood stabilization
      Blood samples are notoriously difficult to deal with because of the blood cell lysis step and the subsequent high presence of globin mRNA in RNA prepared from whole blood. Globin mRNA interferes with downstream expression assays. We have options for stabilizing RNA in blood samples.
      LeukoLOCK Total RNA Isolation System product photo
      LeukoLOCK Total RNA Isolation System contains globin-free mRNA from RNAlater–stabilized leukocytes. Using this system, anticoagulated blood is filter-fractionated, keeping only the leukocytes, and stabilized with RNAlater solution for storage. By excluding red blood cells, this system isolates and purifies RNA from blood without the presence of globin mRNA.
      Tempus Blood RNA Tubes are available for the stabilization and purification of total RNA from whole blood at the point of collection for downstream assays. Each Tempus tube contains a stabilizing and lysing reagent that eliminates the need for subsequent red blood cell lysis or proteinase K treatment. The RNA stabilizing reagent inactivates RNases and selectively precipitates RNA. Genomic DNA can also be isolated from the supernatant of the blood samples stored in a Tempus tube once the RNA is pelleted.
      Ordering information for LeukoLOCK system and Tempus tube kits
      Catalog # Name Size Price (USD) Qty
      AM1923 LeukoLOCK™ Total RNA Isolation System Each
      1,140.00
      4342792 Tempus™ Blood RNA Tube Each
      503.65
      Online Exclusive
      532.00
      Save 28.35 (5%)
      The last step in many nucleic acid isolation protocols—and the first step of many downstream assays—is to resuspend the RNA pellet. After isolation and purification of RNA, it is essential to suspend and store RNA in an appropriate, nuclease-free solution.
      THE RNA Storage Solution is an RNA stabilizing solution that is compatible with many downstream assays, such as:
      • RT-PCR
      • In vitro transcription
      • Northern analysis
      • Nuclease protection assays
      It is a 1 mM sodium citrate, pH 6.5 ± 0.1 buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This solution is prepared from pure agents, under scrupulously monitored conditions in our state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to help ensure their purity and performance.
      Ordering information for THE RNA storage solution
      Catalog # Name Size Price (USD) Qty
      AM7000 THE RNA Storage Solution, 10 x 1 mL Each
      67.65
      Online Exclusive
      72.00
      Save 4.35 (6%)
      AM7001 THE RNA Storage Solution, 50 mL Each
      67.65
      Online Exclusive
      72.50
      Save 4.85 (7%)
      Resources for RNA stabilization
      Further reading
      Posters
      Tips from the bench
      Your data
      TechNotes articles
      Support
      For Research Use Only. Not for use in diagnostic procedures.
      Stylesheet for Classic Wide Template adjustments