One family of pipettes to meet any application challenges
Developed by Thermo Fisher Scientific, the Finnpipette systems are renowned for their accuracy, precision, and ease of use. With a wide selection of models and features, these pipettes are suitable for a variety of applications. With innovative features, such as adjustable volume setting, ergonomic design, and reliable performance, results and user comfort are optimized.
Key features
Ergonomically designed
Our lightweight designs to strain-free pipette features
Comprehensive model selection
Single-, 8-, 12- , 16- Channel
Manual and electronic pipettes
Volumes 0.2 µl to 10 mL
Quality you can depend on Certified to meet standards ISO 9001, ISO 14001 and ISO 13485
Industry-leading warranty
5-year warranty on F1 and F2 pipettes
Broad portfolio of tips Choose from standard or specialized varieties for specific application and improved ergonomics
Available Finnpipette systems
Exceptional performance with comfort
Effortless one-handed operation and security
Volume adjustment is now virtually effortless and enjoyably silent, also enabling one-handed volume changes. 
Enhanced Precision and comfort
Finnpipette F1 pipettes now come with a textured adjustment knob that provides a firmer feel of confidence when setting volumes. We even offer handle grip strips for your pipette, providing a more secure and comfortable grip. A wider tip ejector gives your thumb the room and respect it deserves.
finnpipette F1
Ergonomically designed to help reduce pipetting stress
Finnpipette F1 pipettes feature a lightweight design, now with lowered pipetting forces that help reduce Repetitive Stress Syndrome (RSI) without compromising results. That means maximum comfort with confidence you can count on each and every day.
finnpipette F1 multichannel plate
Customer testimonial
"We have always found Finnpipettes address a variety of ergonomic issues for our users; from the short form pipettes that assist individuals with a smaller hand size to the development of the index finger trigger pipetting action that can be used by left or right handed individuals and takes the strain away from the thumb.  We have been highly impressed by these innovations that aim to address RSI at the bench.”
Assistant Laboratory Manager, UK
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.
1x1 image pixel for data collection