Thermo Scientific Pierce Luciferase Reporter Assays allow you to investigate promoter activity by measuring light output from luciferase enzyme that is expressed under the control of your promoter of interest. Luciferase genes vary in stability, light output and spectral characteristics, and reporter kits are available with different luciferases for both single- and dual-spectral analyses.
The Thermo Scientific Pierce Luciferase reporter genes are human codon optimized and offered in a plasmid vector that is optimized for mammalian expression.
Key features of Thermo Scientific Luciferase Reporters
 Cypridina LuciferaseGaussia LuciferaseGaussia-Dura LuciferaseGreen Renilla LuciferaseRed Firefly Luciferase
SubstrateVargulinCoelenterazineD-luciferin
Sample TypeMediaLysate
Lysate
Relative Brightness in Flash Assay107 (media)107105
10(lysate)
Average Half-Life in Flash Assay30 min<5 minN/A5 min>30 min
Average Half-Life in Glow Assay1 hrN/A1 hr3 hr2 hr
The Gaussia, Gaussia-Dura (a mutant form of the Gaussia luciferase gene that confers better bioluminescent signal stability than native luciferase) and Cypridina luciferase proteins are secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The light signal produced by these three secreted luciferases is considerably greater than the signal from either native Firefly or Renilla luciferases. Although greater than 85% of the Gaussia, Gaussia Dura and Cypridina luciferase protein is secreted, the remaining intracellular portion can be robustly detected in lysate, facilitating multiplexing with other intracellular luciferases. 
The Green Renilla luciferase is an intracellular protein with a greater serum stability as well as higher light output than native Renilla and Firefly luciferases. Consequently, Green Renilla reporter assays are more sensitive than assays employing native Renilla luciferase. 
The Red Firefly luciferase is an intracellular protein with an emission peak shifted into the red spectrum compared to the native Firefly luciferase, which allows multiplexing with Gaussia, Cypridina and Green Renilla luciferases.
Featured luciferase assay categories
Luciferase reporter reagents, multiple cloning site vectors, and flash, glow and dual-spectral luciferase assay kits based on the naturally secreted Cypridina luciferase
Luciferase reporter reagents, multiple cloning site vectors, and flash and dual-spectral luciferase assay kits based on the naturally secreted Gaussia luciferase
Luciferase reporter reagents, multiple cloning site vectors, and flash, glow and dual-spectral luciferase assay kits based on the intracellularly expressed Renilla luciferase
Luciferase reporter reagents, multiple cloning site vectors, and flash, glow and dual-spectral luciferase assay kits based on the intracellularly expressed firefly luciferase
luciferase
Custom Cell-Based Assay Services
When your studies require specialized, reliable, high-quality solutions, count on our custom biology services. We offer all components necessary to develop the most appropriate assay for your research or discovery target of interest. Dedicated project managers will guide your project through every step of the process, with frequent progress updates.
Send us an inquiry
Supporting reagents for luciferase reporter assays
Videos of luciferase protocols
Video Player is loading.
Current Time 0:00
Duration 4:46
Loaded: 0.00%
Stream Type LIVE
Remaining Time 4:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video Player is loading.
    Current Time 0:00
    Duration 5:06
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 5:06
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Video Player is loading.
      Current Time 0:00
      Duration 4:46
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 4:46
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Video Player is loading.
        Current Time 0:00
        Duration 5:06
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 5:06