Wet Tank Transfer Systems
Wet transfer is the most common and traditional strategy for western blot transfer due to its efficiency and throughput. We offer three systems for tank transfer that minimize the amount of transfer buffer required.
Transfer membranes  Transfer buffers
Which wet tank transfer system is right for you?
 Invitrogen Mini Blot Module and Mini Gel TankInvitrogen XCell II Blot Module and SureLock TankInvitrogen SureLock Tandem Midi Gel Tank with Midi Blot Module
 Mini Blot ModuleXCell II Blot ModuleSureLock Tandem Midi Blot Module
Capacity1 mini gel per blot module; 1-2 blot modules per tank1-2 mini gels per blot module; 1 blot module per tank1 midi gel per blot module, 1-2 blot modules per tank
Transfer buffer requirements220 mL per blot800 mL300 mL per blot
Transfer time60 min60-120 min30 min
Blotting area9 x 9 cm9 x 9 cm9.2 x 14.4 cm
Compatible power supplyPowerEase Touch Power Supplies, Owl systems, for other systems use with Novex Power Supply Adapters (Cat. No. ZA10001)PowerEase Touch Power Supplies, Owl systems, for other systems use with Novex Power Supply Adapters (Cat. No. ZA10001)PowerEase Touch Power Supplies or Owl systems
Required equipmentMini Gel TankXCell SureLock Mini-Cell or XCell II Mini-CellSureLock Tandem Midi Gel Tank: capacity for up to 2 blot modules
Convenient, reliable western transfer
Two Mini Blot Modules fit into the Mini Gel Tank and are designed to make your western transfers simple and easy to perform.
  • Universal module design—allows modules to fit in either chamber of the tank, simplifying the transfer setup
  • Unique gasket seal—helps prevent buffer leakage so there is no mess during setup of your western transfer
  • 1/2 inch buffer chamber—requires half the typical volume of methanol-based transfer buffers
  • Standard 60 min transfer protocol—accelerates your western workflow so you can get results faster
  • Robust electrodes, sturdy steel plates—for highly efficient and reliable western transfers
Example transfer conditions
Gel typeMembraneVoltageStarting current (mA)Ending current (mA)Run time
Bolt Bis-Tris Plus 4-12% (MES)Nitrocellulose101606060
PVDF2034013060
NuPAGE 4-12% Bis-Tris (MES)Nitrocellulose101606060
PVDF2039013060
Novex 4-20% Tris-Glycine (denatured)Nitrocellulose10705060
PVDF2016010060
NuPAGE 3-8% Tris Acetate (denatured)Nitrocellulose101505060
PVDF2038013060
Novex 10-20% TricineNitrocellulose10706060
PVDF2018015060
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
How-to-videos
Video Player is loading.
Current Time 0:00
Duration 4:05
Loaded: 0%
Stream Type LIVE
Remaining Time 4:05
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    How to perform a western wet tank transfer using the Invitrogen Mini Blot Module
    Learn how to perform a western blot transfer using the Mini Blot module in a Mini Gel Tank.
    Video Player is loading.
    Current Time 0:00
    Duration 5:24
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 5:24
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      How to separate proteins using an Invitrogen precast SDS-PAGE gel
      Learn how to separate proteins by using a Invitrogen Bolt Bis-Tris Plus SDS-PAGE precast gel and Mini Gel Tank.
      Get started with Mini Bolt Module Welcome Packs
      Catalog # Name Size Price (USD) Qty
      B1000A Bolt™ Western Pack A (Nitrocellulose) Each
      1,054.00
      B1000B Bolt™ Western Pack B (PVDF) Each
      1,056.00
      The XCell II Blot Module allows you to easily transfer proteins or nucleic acids from mini-gels to membranes. It fits neatly into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for Western, Southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.
      Example transfer conditions
      Gel typeMembraneTransfer bufferTransfer conditionsExpected currentRun time
      Tris-GlycineNitrocellulose or PVDFTris-Glycine Transfer Buffer with 20% methanol.
      1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol.
      25V constant100 mA1-2 hr
      TricineNitrocellulose or PVDFTris-Glycine Transfer Buffer with 20% methanol.
      1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol.
      25V constantStart: 100 mA1-2 hr
      Bis-TrisNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 170 mA End: 100mA1 hour
      Tris-AcetateNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 200 mA End: 180mA60
      IEFNitrocellulose or PVDF0.7% acetic acid, pH 3.010 V constantStart: 65-85 mA60
      TBE, TBE-Urea and DAN RetardationNylon45 mM Tris, 45 mM boric acid, 1 mM EDTA30 V constantStart: 360 mA End: 270 mA1-2 hr
      The expected current listed in the table below is for transferring one gel. If you are transferring two gels in the blot module, the expected current will double.
      For overnight blotting, transfer at a constant voltage of 10-15 V.
      Video Player is loading.
      Current Time 0:00
      Duration 6:14
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 6:14
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Discover how to use the XCell II Blot Module
        Learn how to perform a wet tank transfer using the XCell II Blot Module in the SureLock Tank with this detailed step by step video.
        Video Player is loading.
        Current Time 0:00
        Duration 4:48
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 4:48
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Learn how to run a gel using the XCell SureLock System
          Watch this step by step video of how to run a NuPAGE precast get in the SureLock Tank.
          Convenient, reliable wet transfer of midi gels
          Invitrogen SureLock Tandem Midi Gel Tank with Midi Blot Module
          When your western blotting workflow requires higher throughput, Invitrogen midi gels will allow you to load up to 26 samples per gel. You can use the SureLock Tandem Midi Gel Tank to run up to 2 high-performance Invitrogen midi gels and transfer them to membranes in the same tank using the SureLock Tandem Midi Blot Module. Two SureLock Tandem Midi Blot Modules fit into the innovative, dual-purpose SureLock Tandem Midi Gel Tank and are designed for easy, room-temperature gel transfers. Only 300 mL of methanol-containing buffer is required per gel transfer, reducing buffer and hazardous disposal costs.
          • 2-in-1 SureLock Tandem Midi Gel Tank—no need to purchase a separate tank for western transfers
          • Room temperature transfer—save the time, planning, and mess required with other systems to pre-chill buffers, freeze ice packs, or prepare ice baths
          • 300 mL per gel transfer buffer volume—lower transfer buffer volume requirement reduces reagent consumption and hazardous disposal costs
          • Fast and efficient—30-minute transfer without compromising performance
          • Easy to use—convenient blot assembly instructions are printed on the accessory transfer tray
          • Robust electrode plates—for highly efficient and reliable western transfers
          Get started with a Midi Blot Module Welcome Pack
          SureLock Tandem Midi Welcome Pack, Nitrocellulose
          SureLock Tandem Midi Welcome Pack
          Includes:
          • SureLock Tandem Midi Gel Tank
          • Midi Blot Modules, 2 each
          • SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, Nitrocellulose
          • SureLock Tandem Transfer Tray
          • Blot Roller
          • Gel knife
          Consistent, reliable western blotting results
          When used with high-performance Invitrogen precast midi gels, you can depend on the SureLock Tandem Midi Gel Tank and Midi Blot Module to generate consistent, high-throughput western blots. The Invitrogen midi gels offer reproducible high-performance protein separation, resulting in sharp, well-resolved bands in straight lanes, and the SureLock Tandem Midi Blot Module provides dependable uniform transfer of proteins across the gel.
          To demonstrate the SureLock Tandem Midi Blot Module’s uniformity of transfer across a membrane, a NuPAGE 4-12% Bis-Tris Midi gel was loaded with recurring dilutions of E. coli lysate ranging from 2-0.25 µg and SeeBlue Plus2 Pre-stained Protein Standard. Electrophoresis was performed and the gel was transferred onto a 0.45 µm PVDF membrane. Results are shown in Figure 1.
          A
          Data illustrating uniform transfer is achieved using the SureLock Tandem Midi gel Tank
          B
          Densitometric Signal Linearity versus Protein Load
          Figure 1. Uniform transfer using SureLock Tandem Midi Gel Tank. Total protein normalization of transferred proteins shows uniform transfer across the blot. 
          A. No-Stain Reagent Labeled PVDF Blot. A NuPAGE 4-12% Bis-Tris Midi gel was loaded with recurring dilutions of E. coli Lysate ranging from 2 to 0.25 µg and SeeBlue Plus2 Pre-stained Protein Standard in lanes 1-2 and 19-20. The gel was electrophoresed using MES running buffer in the SureLock Tandem Midi Gel Tank. Proteins from the gel were transferred onto a 0.45 µm PVDF membranes at 25V constant for 30 minutes using the SureLock Tandem Midi Blot Module. After transfer, the proteins on the membrane were labeled following the No-Stain Protein Labeling Reagent protocol for midi-sized membranes. The image was captured using an iBright Imager with the No-Stain Membrane epi setting (455-485 nm excitation and 565-615 nm emission). As the SeeBlue Plus2 Pre-stained Protein Standard is already labeled, its protein bands are not all susceptible to labeling with No-Stain Reagent, and therefore are not all visible in this image. 
          B. Densitometric Signal Linearity versus Protein Load. The densitometric signal intensity was determined for each lane. Technical replicates (n=4) were averaged and plotted to determine the linear regression for the entire concentration range (R2=0.9994). Error bars represent standard deviation. 
          Recommended transfer conditions
          Recommended transfer conditions for all Invitrogen precast midi gels:
          Constant Voltage (V)Time (min)
          2530
          Mini Tank System
          Catalog # Name Size Price (USD) Qty
          A25977 Mini Gel Tank Each
          735.65
          Online Exclusive
          760.00
          Save 24.35 (3%)
          B1000 Mini Blot Module Each
          480.00
          B1000A Bolt™ Western Pack A (Nitrocellulose) Each
          1,054.00
          B1000B Bolt™ Western Pack B (PVDF) Each
          1,056.00
          NW2000 Mini Gel Tank and Blot Module Set Each
          1,314.65
          Online Exclusive
          1,368.00
          Save 53.35 (4%)
          XCell SureLock System
          Catalog # Name Size Price (USD) Qty
          EI9014 Buffer Core w/ Electrodes for XCell SureLock™ & XCell II™ Blot Module Each
          534.00
          EI9052 Sponge Pad for Blotting Each
          66.00
          EI9051 XCell II™ Blot Module Each
          704.00
          EI0001 XCell SureLock™ Mini-Cell Each
          895.65
          Online Exclusive
          938.00
          Save 42.35 (5%)
          EI0012 Buffer Dam for XCell SureLock™ and XCell II™ Blot Module Each
          58.75
          EI0002 XCell SureLock™ Mini-Cell and XCell II™ Blot Module Each
          1,481.65
          Online Exclusive
          1,538.00
          Save 56.35 (4%)
          SureLock Tandem Midi System
          Which wet tank transfer system is right for you?
           Invitrogen Mini Blot Module and Mini Gel TankInvitrogen XCell II Blot Module and SureLock TankInvitrogen SureLock Tandem Midi Gel Tank with Midi Blot Module
           Mini Blot ModuleXCell II Blot ModuleSureLock Tandem Midi Blot Module
          Capacity1 mini gel per blot module; 1-2 blot modules per tank1-2 mini gels per blot module; 1 blot module per tank1 midi gel per blot module, 1-2 blot modules per tank
          Transfer buffer requirements220 mL per blot800 mL300 mL per blot
          Transfer time60 min60-120 min30 min
          Blotting area9 x 9 cm9 x 9 cm9.2 x 14.4 cm
          Compatible power supplyPowerEase Touch Power Supplies, Owl systems, for other systems use with Novex Power Supply Adapters (Cat. No. ZA10001)PowerEase Touch Power Supplies, Owl systems, for other systems use with Novex Power Supply Adapters (Cat. No. ZA10001)PowerEase Touch Power Supplies or Owl systems
          Required equipmentMini Gel TankXCell SureLock Mini-Cell or XCell II Mini-CellSureLock Tandem Midi Gel Tank: capacity for up to 2 blot modules
          Convenient, reliable western transfer
          Two Mini Blot Modules fit into the Mini Gel Tank and are designed to make your western transfers simple and easy to perform.
          • Universal module design—allows modules to fit in either chamber of the tank, simplifying the transfer setup
          • Unique gasket seal—helps prevent buffer leakage so there is no mess during setup of your western transfer
          • 1/2 inch buffer chamber—requires half the typical volume of methanol-based transfer buffers
          • Standard 60 min transfer protocol—accelerates your western workflow so you can get results faster
          • Robust electrodes, sturdy steel plates—for highly efficient and reliable western transfers
          Example transfer conditions
          Gel typeMembraneVoltageStarting current (mA)Ending current (mA)Run time
          Bolt Bis-Tris Plus 4-12% (MES)Nitrocellulose101606060
          PVDF2034013060
          NuPAGE 4-12% Bis-Tris (MES)Nitrocellulose101606060
          PVDF2039013060
          Novex 4-20% Tris-Glycine (denatured)Nitrocellulose10705060
          PVDF2016010060
          NuPAGE 3-8% Tris Acetate (denatured)Nitrocellulose101505060
          PVDF2038013060
          Novex 10-20% TricineNitrocellulose10706060
          PVDF2018015060
          * Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
          How-to-videos
          Video Player is loading.
          Current Time 0:00
          Duration 4:05
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 4:05
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            How to perform a western wet tank transfer using the Invitrogen Mini Blot Module
            Learn how to perform a western blot transfer using the Mini Blot module in a Mini Gel Tank.
            Video Player is loading.
            Current Time 0:00
            Duration 5:24
            Loaded: 0%
            Stream Type LIVE
            Remaining Time 5:24
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              How to separate proteins using an Invitrogen precast SDS-PAGE gel
              Learn how to separate proteins by using a Invitrogen Bolt Bis-Tris Plus SDS-PAGE precast gel and Mini Gel Tank.
              Get started with Mini Bolt Module Welcome Packs
              Catalog # Name Size Price (USD) Qty
              B1000A Bolt™ Western Pack A (Nitrocellulose) Each
              1,054.00
              B1000B Bolt™ Western Pack B (PVDF) Each
              1,056.00
              The XCell II Blot Module allows you to easily transfer proteins or nucleic acids from mini-gels to membranes. It fits neatly into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for Western, Southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.
              Example transfer conditions
              Gel typeMembraneTransfer bufferTransfer conditionsExpected currentRun time
              Tris-GlycineNitrocellulose or PVDFTris-Glycine Transfer Buffer with 20% methanol.
              1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol.
              25V constant100 mA1-2 hr
              TricineNitrocellulose or PVDFTris-Glycine Transfer Buffer with 20% methanol.
              1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol.
              25V constantStart: 100 mA1-2 hr
              Bis-TrisNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 170 mA End: 100mA1 hour
              Tris-AcetateNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 200 mA End: 180mA60
              IEFNitrocellulose or PVDF0.7% acetic acid, pH 3.010 V constantStart: 65-85 mA60
              TBE, TBE-Urea and DAN RetardationNylon45 mM Tris, 45 mM boric acid, 1 mM EDTA30 V constantStart: 360 mA End: 270 mA1-2 hr
              The expected current listed in the table below is for transferring one gel. If you are transferring two gels in the blot module, the expected current will double.
              For overnight blotting, transfer at a constant voltage of 10-15 V.
              Video Player is loading.
              Current Time 0:00
              Duration 6:14
              Loaded: 0%
              Stream Type LIVE
              Remaining Time 6:14
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Discover how to use the XCell II Blot Module
                Learn how to perform a wet tank transfer using the XCell II Blot Module in the SureLock Tank with this detailed step by step video.
                Video Player is loading.
                Current Time 0:00
                Duration 4:48
                Loaded: 0%
                Stream Type LIVE
                Remaining Time 4:48
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  Learn how to run a gel using the XCell SureLock System
                  Watch this step by step video of how to run a NuPAGE precast get in the SureLock Tank.
                  Convenient, reliable wet transfer of midi gels
                  Invitrogen SureLock Tandem Midi Gel Tank with Midi Blot Module
                  When your western blotting workflow requires higher throughput, Invitrogen midi gels will allow you to load up to 26 samples per gel. You can use the SureLock Tandem Midi Gel Tank to run up to 2 high-performance Invitrogen midi gels and transfer them to membranes in the same tank using the SureLock Tandem Midi Blot Module. Two SureLock Tandem Midi Blot Modules fit into the innovative, dual-purpose SureLock Tandem Midi Gel Tank and are designed for easy, room-temperature gel transfers. Only 300 mL of methanol-containing buffer is required per gel transfer, reducing buffer and hazardous disposal costs.
                  • 2-in-1 SureLock Tandem Midi Gel Tank—no need to purchase a separate tank for western transfers
                  • Room temperature transfer—save the time, planning, and mess required with other systems to pre-chill buffers, freeze ice packs, or prepare ice baths
                  • 300 mL per gel transfer buffer volume—lower transfer buffer volume requirement reduces reagent consumption and hazardous disposal costs
                  • Fast and efficient—30-minute transfer without compromising performance
                  • Easy to use—convenient blot assembly instructions are printed on the accessory transfer tray
                  • Robust electrode plates—for highly efficient and reliable western transfers
                  Get started with a Midi Blot Module Welcome Pack
                  SureLock Tandem Midi Welcome Pack, Nitrocellulose
                  SureLock Tandem Midi Welcome Pack
                  Includes:
                  • SureLock Tandem Midi Gel Tank
                  • Midi Blot Modules, 2 each
                  • SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, Nitrocellulose
                  • SureLock Tandem Transfer Tray
                  • Blot Roller
                  • Gel knife
                  Consistent, reliable western blotting results
                  When used with high-performance Invitrogen precast midi gels, you can depend on the SureLock Tandem Midi Gel Tank and Midi Blot Module to generate consistent, high-throughput western blots. The Invitrogen midi gels offer reproducible high-performance protein separation, resulting in sharp, well-resolved bands in straight lanes, and the SureLock Tandem Midi Blot Module provides dependable uniform transfer of proteins across the gel.
                  To demonstrate the SureLock Tandem Midi Blot Module’s uniformity of transfer across a membrane, a NuPAGE 4-12% Bis-Tris Midi gel was loaded with recurring dilutions of E. coli lysate ranging from 2-0.25 µg and SeeBlue Plus2 Pre-stained Protein Standard. Electrophoresis was performed and the gel was transferred onto a 0.45 µm PVDF membrane. Results are shown in Figure 1.
                  A
                  Data illustrating uniform transfer is achieved using the SureLock Tandem Midi gel Tank
                  B
                  Densitometric Signal Linearity versus Protein Load
                  Figure 1. Uniform transfer using SureLock Tandem Midi Gel Tank. Total protein normalization of transferred proteins shows uniform transfer across the blot. 
                  A. No-Stain Reagent Labeled PVDF Blot. A NuPAGE 4-12% Bis-Tris Midi gel was loaded with recurring dilutions of E. coli Lysate ranging from 2 to 0.25 µg and SeeBlue Plus2 Pre-stained Protein Standard in lanes 1-2 and 19-20. The gel was electrophoresed using MES running buffer in the SureLock Tandem Midi Gel Tank. Proteins from the gel were transferred onto a 0.45 µm PVDF membranes at 25V constant for 30 minutes using the SureLock Tandem Midi Blot Module. After transfer, the proteins on the membrane were labeled following the No-Stain Protein Labeling Reagent protocol for midi-sized membranes. The image was captured using an iBright Imager with the No-Stain Membrane epi setting (455-485 nm excitation and 565-615 nm emission). As the SeeBlue Plus2 Pre-stained Protein Standard is already labeled, its protein bands are not all susceptible to labeling with No-Stain Reagent, and therefore are not all visible in this image. 
                  B. Densitometric Signal Linearity versus Protein Load. The densitometric signal intensity was determined for each lane. Technical replicates (n=4) were averaged and plotted to determine the linear regression for the entire concentration range (R2=0.9994). Error bars represent standard deviation. 
                  Recommended transfer conditions
                  Recommended transfer conditions for all Invitrogen precast midi gels:
                  Constant Voltage (V)Time (min)
                  2530
                  Mini Tank System
                  Catalog # Name Size Price (USD) Qty
                  A25977 Mini Gel Tank Each
                  735.65
                  Online Exclusive
                  760.00
                  Save 24.35 (3%)
                  B1000 Mini Blot Module Each
                  480.00
                  B1000A Bolt™ Western Pack A (Nitrocellulose) Each
                  1,054.00
                  B1000B Bolt™ Western Pack B (PVDF) Each
                  1,056.00
                  NW2000 Mini Gel Tank and Blot Module Set Each
                  1,314.65
                  Online Exclusive
                  1,368.00
                  Save 53.35 (4%)
                  XCell SureLock System
                  Catalog # Name Size Price (USD) Qty
                  EI9014 Buffer Core w/ Electrodes for XCell SureLock™ & XCell II™ Blot Module Each
                  534.00
                  EI9052 Sponge Pad for Blotting Each
                  66.00
                  EI9051 XCell II™ Blot Module Each
                  704.00
                  EI0001 XCell SureLock™ Mini-Cell Each
                  895.65
                  Online Exclusive
                  938.00
                  Save 42.35 (5%)
                  EI0012 Buffer Dam for XCell SureLock™ and XCell II™ Blot Module Each
                  58.75
                  EI0002 XCell SureLock™ Mini-Cell and XCell II™ Blot Module Each
                  1,481.65
                  Online Exclusive
                  1,538.00
                  Save 56.35 (4%)
                  SureLock Tandem Midi System
                  For Research Use Only. Not for use in diagnostic procedures.