yeast-showcase
Our PichiaPink Yeast Expression system delivers yields up 12 grams per liter of post-translationally modified recombinant protein.
  • Protein yields up to 12 g/L
  • Scalable production
  • Convenient kit formats
  • Posttranslational modifications via eukaryotic expression
PichiaPink Yeast Expression System for optimized expression
Our new PichiaPink Yeast Expression System contains both low and high copy plasmid backbones, 8 secretion signal sequences and 4 yeast strains—all to help you optimize for highest yield possible of your recombinant protein.
PichiaPink Yeast Expression System features:
  • Plasmid copy number: both high- and low-copy options enable optimization of toxic protein expression
  • Number of secretion signal leader sequences: 8 sequences
  • Number of strains: 4 strains
  • Number of protease-deficient host strains: 3 strains
  • Selection method: adenine selection
Obtain functional proteins
P. pastoris has many eukaryotic post-translation modification processes, offering a high probability of an active function recombinant protein. 
Yeast offer expression advantages
The advantages of P. Pastoris include rapid growth, a well-defined genetic background, simple media formulations and handling techniques--all of which make it an ideal host for producing large quantities of protein in a short period of time.
Scalable expression to meet your needs
Using the P. pastoris system for small scale-up to fermentation-scale production helps ensure you have the flexibility to produce the protein quantity you need for your application.
Ordering information
Catalog # Name Size Price (USD) Qty
A11150 PichiaPink™ Secretion Optimization Kit Each
6,140.00
A11151 PichiaPink™ Secreted Protein Kit Each
3,759.65
Online Exclusive
4,205.00
Save 445.35 (11%)
A11155 PichiaPink™ Secretion Signal Set Each
3,160.00
A11152 PichiaPink™ Vector Kit Each
2,055.00
A11153 PichiaPink™ Secreted Vector Kit Each
1,446.65
Online Exclusive
1,542.00
Save 95.35 (6%)
A11154 PichiaPink™ Expression Strain Set Each
2,195.00
A11156 PichiaPink™ Media Kit Each
1,196.00
For Research Use Only. Not for use in diagnostic procedures.