E-PAGE High-Throughput gel system
The Invitrogen E-PAGE High-Throughput (HTP) Precast Gel System is designed for fast, bufferless medium- and high-throughput protein analysis.
E-Editor 2.0 SoftwareAvailable E-PAGE Gels
Migration charts
The E-PAGE High-Throughput (HTP) Precast Gel System is designed for fast, bufferless medium- and high-throughput protein analysis. Invitrogen E-PAGE Gels are self-contained, precast gels that include a gel matrix and electrodes packaged inside a disposable cassette. E-PAGE Gels are available in 48-well or 96-well formats.
Features of the E-PAGE High-Throughput Precast Gel System include:
  • Ease-of-use—Quick setup and fast separations (23 minutes) for 1D protein analysis
  • Fast loading—Compatible with multichannel pipettors and robotic loading
  • Efficient western blotting and staining—Optimized protocols and reagents
Protein separation on an E-PAGE Gel requires an Invitrogen E-Gel Power Snap Plus Electrophoresis Systems, a compact and automated platform designed for medium- and high-throughput electrophoresis.
E-PAGE Gels
We offer two kinds of E-PAGE Gels:
  • Invitrogen E-PAGE 8% Protein Gels, 48-well—Each gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length, and can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system.
  • Invitrogen E-PAGE 6% Protein Gels, 96-well—Each gel contains 96 sample lanes and 8 marker lanes in a patented staggered-well format with a 1.6-cm run length; well openings are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid handling robotic device.
Both E-PAGE 48-well and 96-well cassettes are conveniently opened with a butterfly opener to remove the gel for downstream staining or blotting. In addition, each cassette is labeled with a unique barcode to facilitate identification of the gel using commercial barcode readers.
Available gel sizes 48-well (3.7 mm thick)
96-well (3.7 mm thick)
Storage conditionsRoom temperature 
Shelf life6–9 months
Available polyacrylamide concentrations6% (96 wells), 8% (48 wells) 
Separation range10–200 kDa
For use with (equipment) mini gelsE-Base integrated devices
Mode of separation Molecular weight
 E-PAGE 48-well 8% gelsE-PAGE 96-well 6% gels
Cassette size 13.5 (l) x 10.8 (h) x 0.67 cm (w)13.5 (l) x 10.8 (h) x 0.67 cm (w)
Gel thickness3.7 mm3.7 mm
Gel volume50 mL50 mL
Well depth3 mm3 mm
Number of wells48 + 4 markers96 + 8 markers
Well format2 x 26Staggered 8 x 13
Well opening3.6 mm x 2.2 mm3.8 mm x 1.8 mm
Running distance32 mm16 mm
Run time23 min14 min
Space between well centers4.5 mm9 mm
Sample volume15 µL20 µl
Maximum protein load20 µg20 µg
Catalog # Name Size Price (TWD) Qty
EP09606 E-PAGE™ Midi Protein Gels, 6%, 3.7 mm, 96-well, 8 gels (1 kit) Each
29,250.00
Online offer
Ends: 30-Jun-2025
33,300.00
Save 4,050.00 (12%)
EP04808 E-PAGE™ Midi Protein Gels, 8%, 3.7 mm, 48-well, 8 gels (1 kit) Each
16,240.00
Online offer
Ends: 30-Jun-2025
23,800.00
Save 7,560.00 (32%)
Catalog # Name Size Price (TWD) Qty
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
590.00
Online offer
Ends: 30-Jun-2025
850.00
Save 260.00 (31%)
EPBUF01 E-PAGE™ Loading Buffer 1 Each
2,210.00
Online offer
Ends: 30-Jun-2025
2,510.00
Save 300.00 (12%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
2,350.00
Online offer
Ends: 30-Jun-2025
3,440.00
Save 1,090.00 (32%)
migration-chart-e-page-gels
Migration pattern of Invitrogen E-PAGE MagicMark Unstained Protein Standard.
The E-PAGE High-Throughput (HTP) Precast Gel System is designed for fast, bufferless medium- and high-throughput protein analysis. Invitrogen E-PAGE Gels are self-contained, precast gels that include a gel matrix and electrodes packaged inside a disposable cassette. E-PAGE Gels are available in 48-well or 96-well formats.
Features of the E-PAGE High-Throughput Precast Gel System include:
  • Ease-of-use—Quick setup and fast separations (23 minutes) for 1D protein analysis
  • Fast loading—Compatible with multichannel pipettors and robotic loading
  • Efficient western blotting and staining—Optimized protocols and reagents
Protein separation on an E-PAGE Gel requires an Invitrogen E-Gel Power Snap Plus Electrophoresis Systems, a compact and automated platform designed for medium- and high-throughput electrophoresis.
E-PAGE Gels
We offer two kinds of E-PAGE Gels:
  • Invitrogen E-PAGE 8% Protein Gels, 48-well—Each gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length, and can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system.
  • Invitrogen E-PAGE 6% Protein Gels, 96-well—Each gel contains 96 sample lanes and 8 marker lanes in a patented staggered-well format with a 1.6-cm run length; well openings are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid handling robotic device.
Both E-PAGE 48-well and 96-well cassettes are conveniently opened with a butterfly opener to remove the gel for downstream staining or blotting. In addition, each cassette is labeled with a unique barcode to facilitate identification of the gel using commercial barcode readers.
Available gel sizes 48-well (3.7 mm thick)
96-well (3.7 mm thick)
Storage conditionsRoom temperature 
Shelf life6–9 months
Available polyacrylamide concentrations6% (96 wells), 8% (48 wells) 
Separation range10–200 kDa
For use with (equipment) mini gelsE-Base integrated devices
Mode of separation Molecular weight
 E-PAGE 48-well 8% gelsE-PAGE 96-well 6% gels
Cassette size 13.5 (l) x 10.8 (h) x 0.67 cm (w)13.5 (l) x 10.8 (h) x 0.67 cm (w)
Gel thickness3.7 mm3.7 mm
Gel volume50 mL50 mL
Well depth3 mm3 mm
Number of wells48 + 4 markers96 + 8 markers
Well format2 x 26Staggered 8 x 13
Well opening3.6 mm x 2.2 mm3.8 mm x 1.8 mm
Running distance32 mm16 mm
Run time23 min14 min
Space between well centers4.5 mm9 mm
Sample volume15 µL20 µl
Maximum protein load20 µg20 µg
Catalog # Name Size Price (TWD) Qty
EP09606 E-PAGE™ Midi Protein Gels, 6%, 3.7 mm, 96-well, 8 gels (1 kit) Each
29,250.00
Online offer
Ends: 30-Jun-2025
33,300.00
Save 4,050.00 (12%)
EP04808 E-PAGE™ Midi Protein Gels, 8%, 3.7 mm, 48-well, 8 gels (1 kit) Each
16,240.00
Online offer
Ends: 30-Jun-2025
23,800.00
Save 7,560.00 (32%)
Catalog # Name Size Price (TWD) Qty
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
590.00
Online offer
Ends: 30-Jun-2025
850.00
Save 260.00 (31%)
EPBUF01 E-PAGE™ Loading Buffer 1 Each
2,210.00
Online offer
Ends: 30-Jun-2025
2,510.00
Save 300.00 (12%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
2,350.00
Online offer
Ends: 30-Jun-2025
3,440.00
Save 1,090.00 (32%)
migration-chart-e-page-gels
Migration pattern of Invitrogen E-PAGE MagicMark Unstained Protein Standard.
For Research Use Only. Not for use in diagnostic procedures.