SnakeSkin Dialysis Tubing
Thermo Scientific SnakeSkin Dialysis Tubing is an easy, ready-to-use form of traditional dialysis membrane tubing. Composed of regenerated cellulose dialysis tubing, it is supplied as an open, pleated (telescoped) tube.
SnakeSkin dialysis tubing is supplied in 8-inch (20 cm) sticks that contain 35 feet of tubing of various diameters (16 mm, 22 mm, and 35 mm) and hold 2–10 mL of sample per centimeter of length. They can be used for desalting and buffer exchange of 10–100 mL sample volumes.
To use, simply pull out and cut off the required length of tubing, fold over one end of tubing, close this end with a dialysis clip, add sample at the open end, and use a second clip to close the remaining end.
Choose the right SnakeSkin dialysis tubing for your experiments
Circular internal diameter16 mm I.D.22 mm I.D.35 mm I.D.
Volume per cm2 mL3.8 mL9.6 mL
See all SnakeSkin Dialysis Tubing
Recommended accessories:
SnakeSkin Dialysis Clips — sturdy, non-floating tube closures suitable for dialysis tubing with flat tube diameter up to 50 mm
Catalog # Name Size Price (USD) Qty
88242 SnakeSkin™ Dialysis Tubing, 3.5K MWCO, 16 mm, 10.5 m Each
189.65
Online Exclusive
212.00
Save 22.35 (11%)
68035 SnakeSkin™ Dialysis Tubing, 3.5K MWCO, 22 mm, 10.5 m Each
203.65
Online Exclusive
234.00
Save 30.35 (13%)
88244 SnakeSkin™ Dialysis Tubing, 3.5K MWCO, 35 mm, 10.5 m Each
225.65
Online Exclusive
255.00
Save 29.35 (12%)
68700 SnakeSkin™ Dialysis Tubing, 7K MWCO, 22 mm, 10.5 m Each
207.65
Online Exclusive
234.00
Save 26.35 (11%)
88243 SnakeSkin™ Dialysis Tubing, 10K MWCO, 16 mm, 10.5 m Each
188.65
Online Exclusive
212.00
Save 23.35 (11%)
68100 SnakeSkin™ Dialysis Tubing, 10K MWCO, 22 mm, 10.5 m Each
204.65
Online Exclusive
234.00
Save 29.35 (13%)
88245 SnakeSkin™ Dialysis Tubing, 10K MWCO, 35 mm, 10.5 m Each
224.65
Online Exclusive
254.00
Save 29.35 (12%)
68011 SnakeSkin™ Dialysis Clips, 6 Clips Each
78.65
Online Exclusive
101.00
Save 22.35 (22%)
Featured data of SnakeSkin dialysis tubing
Influence of surface area to volume ratio on dialysis rate. Graph displays rates of removal of 1 M NaCl from 2 mL, 70 mL, 70 mL, and 200 mL samples dialyzed in 4 respective sizes of Thermo Scientific Dialysis Devices (see Table 1), each equipped with 3.5K MWCO membrane. Dialysis was conducted at room temperature against very large volumes (e.g., 4 L) of water (dialysate). At the indicated times (triangles), the dialysis buffer was changed, and the percentage of NaCl removal was determined by measuring the conductivity of the sample.
Video Player is loading.
Current Time 0:00
Duration 3:11
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:11
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Still using tubing for your dialysis?
    Try Thermo Scientific Slide-A-Lyzer G3 Dialysis Cassettes. These cassettes and flasks are a more convenient and secure approach to performing sample dialysis than traditional dialysis tubing. Slide-A-Lyzer cassettes and flasks offer a rigid, free-standing frame with a secure cap, allowing for easier sample loading/retrieval, and increased sample security.
    For Research Use Only. Not for use in diagnostic procedures.