The culture medium is the most important component of the culture environment, because it provides the necessary nutrients, growth factors, and hormones for cell growth, as well as regulating the pH and the osmotic pressure of the culture.
Although initial cell culture experiments were performed using natural media obtained from tissue extracts and body fluids, the need for standardization, media quality, and increased demand led to the development of defined media. The three basic classes of media are basal media, reduced-serum media, and serum-free media, which differ in their requirement for supplementation with serum.
Basal media
Gibco cell culture medias
The majority of cell lines grow well in basal media, which contain amino acids, vitamins, inorganic salts, and a carbon source such as glucose, but these basal media formulations must be further supplemented with serum.
View all Gibco Cell Culture Media
Reduced-serum media
Another strategy to reduce the undesired effects of serum in cell culture experiments is to use reduced-serum media. Reduced-serum media are basal media formulations enriched with nutrients and animal-derived factors, which reduce the amount of serum that is needed.
Learn more about Advanced Reduced-Serum Media
Serum-free media
Bottle containing Gibco Serum-Free Media
Serum-free media (SFM) circumvents issues with using animal sera by replacing the serum with appropriate nutritional and hormonal formulations. Serum-free media formulations exist for many primary cultures and cell lines, including recombinant protein producing lines of Chinese Hamster Ovary (CHO), various hybridoma cell lines, the insect lines Sf9 and Sf21 (Spodoptera frugiperda), and for cell lines that act as hosts for viral production (e.g., 293, VERO, MDCK, MDBK), and others. One of the major advantages of using serum-free media is the ability to make the medium selective for specific cell types by choosing the appropriate combination of growth factors. The table below lists the advantages and disadvantages of serum-free media.
Pros
  • Increased definition
  • More consistent performance
  • Easier purification and downstream processing
  • Precise evaluation of cellular functions
  • Increased productivity
  • Better control over physiological response
  • Enhanced detection of cellular mediators
Cons
  • Requirement for cell type-specific media formulations
  • Need for higher degree of reagent purity
  • Slower growth
Cell culture media best practices
Here are a few simple tips & tricks to help ensure your cell culture media are maintained for optimal performance.
icon of upward facing arrows
If refrigerated, warm your cell culture media before use. Your cells would be cold at 2–8°C, and you would be too!
lightbulb icon
Keep your cell culture media protected from light. Light exposure degrades the essential vitamins in media that your cells need to grow.
snowflake icon
Once you have supplemented the cell culture media with FBS, place the complete media into the refrigerator (2–8°C ) to maintain performance.
calendar icon
Use your supplemented media within 2–4 weeks to reduce the chances of contamination and the impact of pH drift.
Culture serum
Bottle containing Gibco FBS
Serum is vitally important as a source of growth and adhesion factors, hormones, lipids, and minerals for the culture of cells in basal media. In addition, serum also regulates cell membrane permeability and serves as a carrier for lipids, enzymes, micronutrients, and trace elements into the cell.  While other animal sera (e.g., horse, rabbit, goat, porcine, etc.) are available and utilized, fetal bovine serum (FBS) remains the most universally employed.
See all Gibco Fetal Bovine Serum 
Discover other serum products
FBS contains a sparse amount of gamma globulin, higher levels of growth factors, and fewer complement proteins than both calf and adult bovine serum.  This makes FBS ideal for propagating cell growth while also decreasing the possibility of mammalian cells binding or lysing in the culture, rationalizing the preference of FBS over other animal sera. However, using serum in media has some disadvantages, including high cost, problems with standardization, specificity, and variability, and unwanted effects such as stimulation or inhibition of growth and/or cellular function on certain cell cultures. If the serum is not obtained from reputable source, contamination can also pose a serious threat to successful cell culture experiments.
Our Gibco products, including sera, are tested for contamination and guaranteed for their quality, safety, consistency, and regulatory compliance. We offer a wide variety of serum for your specific cell culture needs—from basic research to specialty assays. Whether you need serum with the least viral risk, the lowest endotoxin levels, or sera qualified for specialty applications and assays, Gibco products offer superior value.
Fetal Bovine Serum (FBS) Basics
Fetal Bovine Serum (FBS) Basics
Discover the important factors to consider in FBS selection, the market scenarios that drive FBS availability and price, as well as the important terms you must know and use when searching for the right FBS for your research needs.
Explore now
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.