Zymographic detection of ß-D-glucuronidase activity.
Zymographic detection of ß-D-glucuronidase activity using ELF® 97 ß-D-glucuronidase substrate (Cat. No. E6587, bottom) after visualization of total-protein patterns using SYPRO® Tangerine protein gel stain (Cat. No. S12010, top).
Zymographic detection of ß-D-glucuronidase activity using ELF® 97 ß-D-glucuronidase substrate (Cat. No. E6587, bottom) after visualization of total-protein patterns using SYPRO® Tangerine protein gel stain (Cat. No. S12010, top).
Related Products
Catalog # Name Size Price (USD) Qty
S12010 SYPRO™ Tangerine Protein Gel Stain (5000X Concentrate in DMSO), 500 μL Each
330.65
Online Exclusive
340.00
Save 9.35 (3%)
Related Images