Word map for carbonyl compound reactions
A carbonyl group is a functional group consisting of a carbon atom joined to an oxygen atom by a double bond. The carbonyl group is present in many of the most synthetically important compounds, including aldehydes, ketones, esters, amides, and other carboxylic acid derivatives. The electronegativity imbalance between the carbon and the oxygen makes the carbon an electrophile that can be exploited synthetically in a number of different ways.
One of the earliest named reactions involving carbonyl group chemistry is the Aldol reaction, which involves the addition of the enol/enolate of a carbonyl compound to an aldehyde or ketone.
Featured reactions involving carbonyl compounds in this series are:
Other well-known named reactions that feature carbonyl groups include:
  • Barbier coupling reaction
  • Baylis–Hillman reaction
  • Corey–Chaykovsky epoxidation
  • Corey–Fuchs alkyne synthesis
  • Dakin oxidation
  • Eschweiler–Clarke methylation
  • Evans aldol reaction
  • Hantzsch dihydropyridine synthesis
  • Pictet–Spengler tetrahydroisoquinoline synthesis
  • Stetter reaction
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!  
Vote now