Word map of Reduction Reactions in Organic Chemistry
Historically, chemical transformations in which hydrogen was gained or oxygen was lost were termed "reduction reactions".
The modern chemical definition of reduction is when a compound gains one or more electrons. It is therefore the opposite of oxidation, where a compound loses one or more electrons.
One of the earliest named reactions featuring reduction is the Tishchenko reaction, named after the Russian organic chemist Vyacheslav Evgen'evich Tishchenko. This reaction is still industrially relevant today as it is used to convert acetaldehyde into the commercially important solvent ethyl acetate.
In this series, we feature the following reduction reactions:
Other named reduction reactions include:
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers.  Do you have a favorite reaction?  Let us know!  
Vote now