Iliad Scanning Transmission Electron Microscope overview
For those seeking to push the boundaries of analytical microscopy, the Thermo Scientific Iliad (S)TEM is an excellent solution for advanced materials characterization. The Iliad EELS Spectrometer and Energy Filter in combination with one of the several EDX detector systems and the new NanoPulser electrostatic beam blanker for electron dose optimization and time-resolved experiments ensure versatility, helping you tackle a diverse range of advanced experiments.
Iliad Scanning Transmission Electron Microscope models
Ultimately, whether you choose the Thermo Scientific Iliad Ultra (S)TEM or the Thermo Scientific Iliad 300 (S)TEM, you can be sure that you are investing in a state-of-the-art system that will elevate your analytical experiments to new heights. Select the model that aligns with your specific needs and embark on a journey of scientific discovery and innovation.
Iliad Ultra (S)TEM
The Iliad Ultra (S)TEM is an excellent solution for characterizing the most complex and challenging materials. By seamlessly providing multimodal, high-speed EDS and EELS with a combination of Ultra-X EDS Detectors, the Iliad EELS Spectrometer and Energy Filter and NanoPulser, the Iliad Ultra (S)TEM enables investigations of the most complex modern materials. The live HT switch lets you choose the optimal voltage to mitigate the beam damage for the most advanced beam-sensitive materials. Plus, when choosing the optimal experimental conditions for each technique, you can freely switch within the wide range of high tension to help ensure optimal results.
  • Seamless multimodal spectroscopy ensured by the Iliad EELS Spectrometer and Energy Filter and Ultra-X EDS Detector
  • NanoPulser for dose optimization and time-resolved experiments
  • Live HT switch in the range of operating voltages from 30 kV to 300 kV
Iliad Ultra (S)TEM
Iliad 300 (S)TEM
The Iliad 300 (S)TEM provides a combination of high flexibility and high productivity to ensure accurate characterization of modern complex materials. The Iliad EELS Spectrometer and Energy Filter combined with two EDS detector system options provide seamless spectroscopy. Operation voltages ranging from 30 to 300 kV give you the choice of the most suitable experiment parameters. And the NanoPulser electrostatic beam blanker delivers dose optimization and provides the base for time-resolved experiments. With the Iliad 300 (S)TEM, you get multimodal, flexible investigation of a wide range of materials.
  • Seamless spectroscopy ensured by the Iliad EELS Spectrometer and Energy Filter combined with Super-X or Dual-X EDS detectors
  • NanoPulser for dose optimization and time-resolved experiments
  • High flexibility to accommodate a broad range of applications
Iliad 300 (S)TEM
For Research Use Only. Not for use in diagnostic procedures.