KingFisher Presto System
The Thermo Scientific KingFisher Presto is a versatile benchtop sample preparation instrument that integrates with a compatible robotic liquid handler (required) for hands-free automation. It offers consistent, ultrahigh-throughput extraction and purification of DNA, RNA, proteins, and cells.
Features
In addition to the common features of all KingFisher Purification Systems, the Presto offers:
Automated ultrahigh-throughput purification
24–96 samples per run
Compact size
Easy fitting to automation systems
Designed for integration
Compatible with selected liquid handling instruments
Choose from two plate formats
Wide 20 µL–5 mL volume range
Heating capability
For applications requiring elevated temperatures
Demo videos
Video Player is loading.
Current Time 0:00
Duration 3:01
Loaded: 0%
Stream Type LIVE
Remaining Time 3:01
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Protocols and software
    Thermo Scientific BindIt software is included with KingFisher Duo Prime, Flex, and Presto instruments. The software makes it easy to create, download, run, modify, and store protocols for your KingFisher applications. The software allows you to define and edit steps, parameters, plates, and reagents for every part of your protocol—bind, wash, and elute.
    For more information about BindIt, see the Features tab.
    Applications
    The KingFisher Presto can gently and efficiently isolate and purify nucleic acids, proteins, and cells from a range of sample types for a wide variety of downstream applications. For a selection of application kits, reagents, and protocols that run on the KingFisher Presto, see the Kits, Reagents, and Protocols tab.
    Robotic liquid handlers
    Many commercial robotic liquid handlers are compatible with KingFisher Presto. Contact your KingFisher sales representative or your preferred vendor to see if they have developed and tested a KingFisher interface.
    Plates, plastics, and setup
    For information about plates, plastics, and plate setup products designed to work optimally with the KingFisher Presto, see the Accessories tab.
    Specifications
    Attribute
    Description
    Instrument size
    Benchtop: integrates with robotic liquid handler
    Throughput level
    Ultrahigh
    Processing volume range
    50–150 µL: KingFisher 96-well plate
    50–1,000 µL: 96 deep-well plate
    200–5,000 µL: 24 deep-well plate
    Samples per run
    24 or 96
    Software included
    BindIt
    Customizable protocols
    Yes (with BindIt software)
    Heating/cooling
    From 5ºC above ambient temperature to 115ºC
    Ultraviolet lamp
    No
    Bar code reader
    Included
    User-selectable magnetic heads
    2
    Standalone use
    No; must be integrated with liquid handler
    Protocol test run capability through BindIt software
    Size
    36 x 46.5 x 40 cm; 14.2 x 18.3 x 15.7 in. (W x D x H)
    Weight
    24.0 kg; 52.9 lbs
    Ordering information