Leverage your numerical simulations using Meshing for FEA/CFD for 3D Visualization and Analysis Software
Computer aided engineering (CAE) is used in many fields, such as automotive, aerospace, and biomechanics. Expensive experiments are being replaced by numerical simulations, allowing faster time to market while reducing costs. Simulations also help to advance understanding in many research themes. CAE areas include stress analysis using finite element analysis (FEA) or thermal and fluid flow analysis using computational fluid dynamics (CFD). Those analyses are combined with imaging techniques, such as microCT, to better understand and predict properties and to aid in creation of geometries for finite element models (simulation inputs).
Thermo Scientific Amira and Avizo Software provide a fast and robust meshing engine to automatically create simulation inputs from 3D labeled images. Amira and Avizo Software generate high-quality mesh with smoothed conforming interfaces from large multi-material segmented images. You have fine control over size element distribution, while Amira and Avizo Software preserve thin structures and topology. Meshes can be exported to major solver’s format, along with boundary conditions. Amira and Avizo Software also provide import of the solver’s output for post-processing the results with advanced visualization tools.
Use cases
The XWind extension for Amira-Avizo Software provides dedicated meshing capabilities for advanced simulation (FEA/CFD):
  • Generate high-quality mesh with smoothed conforming interfaces and topology preservation directly from a labeled (segmented) image
  • Dedicated, easy-to-use meshing workroom allows for the creation of mesh and defining boundary conditions for CFD with advanced 3D visualization and inspection tools
  • Mesh any topology with any number of materials
  • Fast robust meshing engine for very large data sets (e.g., 2,000-cube data set can be meshed in couple of hours depending on topology)
  • Fine control over distribution of element size to mix refined and coarser areas with global optimization of mesh quality
  • Boundary layer (high quality within thickness)
  • Export to major solver’s format
  • Export DVC displacements calculated with the Amira-Avizo Software XDigitalVolumeCorrelation extension at the mesh boundaries to link experiment with simulation
Services
Amira-Avizo-Introductory-training_1160x600
Introductory training
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-advanced-training_1160x600
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-Custom-Dev_1160x600
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.
Resources
Video Player is loading.
Current Time 0:00
Duration 0:36
Loaded: 27.17%
Stream Type LIVE
Remaining Time 0:36
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Amira Meshing for FEA/CFD - Upper airway flow analysis in corrective rhinosurgery research.
    Video Player is loading.
    Current Time 0:00
    Duration 0:54
    Loaded: 18.21%
    Stream Type LIVE
    Remaining Time 0:54
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Avizo software provides a virtual lab for characterizing materials. Avizo’s fast and robust meshing capability allows for collaboration with simulation solvers for FEA analysis.
      Video Player is loading.
      Current Time 0:00
      Duration 1:20
      Loaded: 12.38%
      Stream Type LIVE
      Remaining Time 1:20
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Discover how Amira-Avizo Software enables the whole 3D printing workflow, from 2D image stack import, to 3D reconstruction and 3D meshing, right through to 3D printer compatible formats export.
        Data courtesy of National University of Singapore.
        Video Player is loading.
        Current Time 0:00
        Duration 0:36
        Loaded: 27.17%
        Stream Type LIVE
        Remaining Time 0:36
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Amira Meshing for FEA/CFD - Upper airway flow analysis in corrective rhinosurgery research.
          Video Player is loading.
          Current Time 0:00
          Duration 0:54
          Loaded: 2.40%
          Stream Type LIVE
          Remaining Time 0:54
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Avizo software provides a virtual lab for characterizing materials. Avizo’s fast and robust meshing capability allows for collaboration with simulation solvers for FEA analysis.
            Video Player is loading.
            Current Time 0:00
            Duration 1:20
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 1:20
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Discover how Amira-Avizo Software enables the whole 3D printing workflow, from 2D image stack import, to 3D reconstruction and 3D meshing, right through to 3D printer compatible formats export.
              Data courtesy of National University of Singapore.
              Style Sheet for Products Table Specifications
              Contact us
              Style Sheet for Global Design System
              Style Sheet for Komodo Tabs
              Style Sheet for Support and Service footer
              Style Sheet for Fonts
              Style Sheet for Cards