The Grignard Reaction is a prominent process to form carbon-carbon bonds
Wittig reaction overview
Named Reactions in Organic Synthesis
Grignard Reaction Mechanism
Watch our video to learn more about the Grignard Reaction and its mechanisms. 
Video Player is loading.
Current Time 0:00
Duration 2:45
Loaded: 5.93%
Stream Type LIVE
Remaining Time 2:45
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Grignard Reaction?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Grignard Reaction. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Grignard Reaction
    In 1900, French chemist Victor Grignard discovered that when treating an alkyl halide with magnesium metal in diethyl ether, a cloudy solution of an organomagnesium compound was formed. This substance would subsequently react with aldehydes and ketones to produce secondary and tertiary alcohols, respectively.
    These organomagnesium compounds became known as Grignard reagents, and their addition across carbon–heteroatom multiple bonds is now called the Grignard reaction. Very shortly after this discovery, the Grignard reaction became one of the best known and most versatile carbon–carbon bond forming reactions.
    Grignard reagents are typically prepared by reacting alkyl, aryl, or vinyl halides with magnesium metal in aprotic nucleophilic solvents such as ethers. The carbon magnesium bond is highly polar, making Grignard reagents excellent carbon nucleophiles. As a result, the subsequent carbon–carbon bond-forming step is straightforward.
    Grignard reagents have been used in the synthesis of several natural products, including the total synthesis of (±)-lepadiformine and several natural and modified cyclotetrapeptide trapoxins.
    Other Carbonyl Compound Reactions
    Other Carbonyl Compound Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 4.09%
    Stream Type LIVE
    Remaining Time 4:00