If you’re looking to culture human primary amniotic fluid cells or chorionic villus cells, AminoMAX™ media is the right product for you.  Our formulations are designed to increase cell attachment, growth rates and provide high metaphase yields.
Product Information
AmnioMAX™ C-100 medium was developed specifically for the in vitro diagnostic testing of human amniotic fluid specimens and chorionic villus sampling.  Designed to help minimize diagnostic turnaround time, AmnioMAX™ medium maximizes colony growth while maintaining the cytogenetic analyzability of resulting colonies.
AmnioMAX™-II Complete Medium is a second-generation formulation, further optimized for cell attachment and increased pH stability.  Supplied in a single bottle, it is complete with antibiotics and growth promoting agents, and requires no further supplementation.
AmnioMAX™ medium supports the growth of amniocytes (Figure 1) and delivers superior plating density.  These results demonstrate that human amniocytes cultured in AmnioMAX™ medium give rise to approximately 22% to 25% more colonies and 44% to 80% more cytogenetically analyzable colonies as compared to other amniocyte media.  In addition, as shown in Figure 2, the mean mitotic index rate of cultured primary amniocytes grown in AmnioMAX™ medium was significantly greater than that achieved with Chang medium.
Figure 1 - Growth kinetics of aminocytes cultured with AmnioMAX™ medium.  A representative growth curve of secondary aminocytes cultured in either AminoMAX™ medium or MEM-alpha supplemented with 20% FBS is shown.  Cell counts were taken at various intervals.  Results shown are the mean of duplicate samples.
Figure 2 - Mitotic rate comparison of AmnioMAX™ medium and Chang medium.  The relative mitotic index rate for primary aminocytes cultured in either AminoMAX™ medium or Chang medium in situ for 6 days was determined.  The number of mitotic events per 5,000 cells was scored for each culture medium.
Ordering information
Catalog # Name Size Price (USD) Qty
11269016 AmnioMAX™-II Complete Medium Each
143.00
12558011 AmnioMAX™ C-100 Complete Medium Each
146.00
17001082 AmnioMAX™ C-100 Basal Medium, 90 mL Each
40.73
17001074 AmnioMAX™ C-100 Basal Medium, 450 mL Each
159.00
12556015 AmnioMAX™ C-100 Supplement, 15 mL Each
89.65
Online Exclusive
98.00
Save 8.35 (9%)
12556023 AmnioMAX™ C-100 Supplement, 75 mL Each
361.65
Online Exclusive
396.00
Save 34.35 (9%)
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions