Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®.
Collage of images of cyanobacteria stained with various blue- or green-fluorescent probes to complement the natural red autofluorescence from chlorophyll and phycobilisomes. The round cells are Synechocystis sp. (strain PCC 6803), and their membranes were labeled with green-fluorescent BODIPY® FL propionic acid (Cat. no. D2183). The cylindrical cells are Synechococcus sp. (strain PCC 7942), stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490). The filamentous cyanobacteria, Anabaena cylindrica, were labeled with either the green-fluorescent cystosolic stain, CellTracker™ Green BODIPY® (Cat. no. C2102), or with the lipophilic membrane stain BODIPY® FL (Cat. no. D2183). The image was contributed by Mary Sarcina, University College, London.
Collage of images of cyanobacteria stained with various blue- or green-fluorescent probes to complement the natural red autofluorescence from chlorophyll and phycobilisomes.  The round cells are <i>Synechocystis</i> sp. (strain PCC 6803), and their membranes were labeled with green-fluorescent BODIPY® FL propionic acid (Cat. no. D2183). The cylindrical cells are <i>Synechococcus</i> sp. (strain PCC 7942), stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490).  The filamentous cyanobacteria, <i>Anabaena cylindrica</i>, were labeled with either the green-fluorescent cystosolic stain, CellTracker™ Green BODIPY® (Cat. no. C2102), or with the lipophilic membrane stain BODIPY® FL (Cat. no. D2183). The image was contributed by Mary Sarcina, University College, London.
Related Products
Catalog # Name Size Price (USD) Qty
C2102 CellTracker™ Green BODIPY™ Dye, 5 mg Each
361.65
Online Exclusive
372.00
Save 10.35 (3%)
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D2183 BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid) Each
423.00
Related Images