Centrifuge Best Practices
Best practices for laboratory centrifuges
Welcome to our comprehensive resource hub on best practices for lab centrifuges. Whether you're a novice or an experienced professional, our site provides essential information to help ensure you get the most out of your centrifugation processes. Explore centrifugation basics and understand the fundamental principles, learn how to properly balance a centrifuge, and discover vital care and maintenance tips to extend the life of your equipment. 
Additionally, our collection of installation videos and step-by-step tutorials will guide you through setup and use, helping support optimal centrifuge efficiency.
Proper care and maintenance
On-demand lab talk: Centrifuge best practice
On-demand lab talk: Centrifuge rotor safety
Rotor care guide
RPM, RCF and k-factor
Converting RPM (revolutions per minute) to RCF (relative centrifugal force) is a common task in laboratory settings when using centrifuges. It involves a straightforward calculation using the rotor radius, while the k-factor provides a means to evaluate and compare the efficiency of different centrifuge rotors. Understanding both concepts is crucial for optimizing centrifugation protocols in the laboratory.
Relative Centrifugal Force (RCF) is a measure of the force exerted on a sample in a centrifuge, relative to the force of gravity. The formula to convert RPM to RCF is:
g = (1.118 x 10-5) x r x RPM2
  • g is the relative centrifugal force (RCF).
  • r is the radius of the rotor in centimeters (cm).
  • RPM is the speed of the centrifuge in revolutions per minute.
The k-factor is a value that describes the efficiency of a centrifuge rotor in pelleting particles. It is used to compare the relative performance of different rotors and is particularly useful when switching between rotors or centrifuges.
The k-factor is particularly important for:
  • Comparing the performance of different rotors.
  • Estimating the time required for pelleting particles of a given size and density.
  • Standardizing protocols across different centrifuges and rotors.
RPM vs RCF Calculator for Fiberlite rotors
Application note
Effortlessly transfer centrifugation protocols: simplifying your workflow (k-factor)
How-to videos
Box to bench videos
Box to Bench
Thermo Scientific Megafuge ST4F Plus general purpose centrifuge unboxing and installation
Box to Bench
Thermo Scientific Sorvall X1 Pro benchtop centrifuge unboxing and installation
Box to Bench and Setup
Thermo Scientific CentriPAK BioProcess Container (BPC) System
Tutorials
Touchscreen tutorial for Thermo Scientific X Pro general purpose centrifuges
How to correctly handle windshield rotors
ClickSeal O-Ring Installation
How to use the Ultracrimp Sealing tool for Ultracrimp Tubes
How to use the CentriPAK BPC
How to set up and use the TZ-32 Rotor in a WX+ Ultracentrifuge
How to set up and use the TCF-20 Rotor in a LYNX 6000 Superspeed Centrifuge
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.
1x1 image pixel for data collection