Unleashing the power of Prompt Gamma Neutron Activation Analysis (PGNAA) for industrial processing
PGNAA technology involves bombarding materials with neutrons and measuring the secondary or prompt gamma rays that are subsequently emitted. When a thermal, or relatively low energy neutron (<0.025 eV) closely approaches or collides with the nucleus of an atom there is a transfer of energy that triggers the sequence of events shown below.
PGNAA nucleus transition
The energy of the gamma-ray released as the nucleus returns to its stable state is associated with the atom from which it was released giving each element an identifying signature or fingerprint. Threshold of detection values for different elements are a function of their tendency to interact with neutrons and determine the suitability of PGNAA for alternative applications. That said, the applicability of PGNAA methods extends right across the processing industries as the following resources illustrate.
Useful resources
See here for several pieces of information that can help, please visit the resources section to find more in depth information.
App note: Bulk Ore Sorting in Base and Precious Metals
Find out more about the benefits of bulk ore sorting and how best to use online elemental analysis to implement it.
Bulk Ore Sorting Mine
Success story: Improving sinter feed analysis
Read how Hüttenwerke Krupp Mannesmann GmbH (HKM, Duisburg, Germany), a subsidiary of Thyssenkrupp Steel Europe AG achieved a payback time of less than one year following an investment in PGNAA for sinter feed analysis.
Sinter Feed Analysis
Video: CQM Flex Coal Analysis
Take a closer look at how PGNAA delivers the information coal processors need to meet increasingly stringent customer requirements, profitably and with minimal environmental impact.
CQM Flex Video
Animation: Mining Tool Kit
Zoom in to the supply chain link of interest to see how PGNAA and PFTNA is used from mine to product quality control.
Mining animation
Neutron supply
There are two main options for neutron supply for PGNAA technology.  One is a radioisotope, Californium 252 (252Cf), and the other is a neutron generator system. Such systems deploy PFTNA, using isotopes of hydrogen, deuterium (2H) and tritium (3H,) to produce neutrons via a fusion reaction. One distinctive difference when using a neutron generator versus a traditional Cf-252 source is therefore the ability to turn off the neutron generator when not in use.
Chart of energy
System design
In addition to the neutron source, the performance and ease of use of PGNAA online analyzers is defined by detector quality and configuration and the extent to which system hardware and software support integration within the process.
Scintillation detectors with a high purity crystalline structure are the technology of choice for gamma-ray detection. These produce photons, proportional in energy to the energy of the detected gamma-rays, that are amplified and converted into electrical signals to generate a composite energy spectrum for the sample as shown below.
When it comes to system configuration, cross-belt analyzers such as the Thermo Scientific CB Omni Agile, and Thermo Scientific ECA-3 Elemental Cross-belt Analyzer are installed directly on the conveyor belt to provide minute-by-minute, uniform measurement of the entire material stream. This approach eliminates sampling errors and delivers data at a high frequency thereby helping to minimize variability in material quality.  The Thermo Scientific CQM Flex Coal Analyzer is designed to fit in line with a mechanical sampling system to offer higher precision and accuracy. PGNAA technology is equally suitable for slurries, with systems such as the Thermo Scientific GS Omni Analyzer enabling the measurement of multiple process streams depending on measurement frequency requirements.
Data presentation and use
The high measurement frequency of online PGNAA systems makes effective data handling and presentation an essential element of any project. Optimizing data use is the key to achieving a good rate of return on any investment in PGNAA technology and powerful, industry-relevant software can be critical in this regard.  
Webcast: Realize the vision of clean energy through accurate and effective representative analysis in bulk ore sorting
Find out more about the criticality of uniform, representative PGNAA measurements when it comes to effective bulk ore sorting, and the system features that deliver it.
Realize the vision of clean energy through accurate and effective representative analysis in bulk ore sorting
Webcast: The value behind integration and digitalization of processing plants data
Learn about the untapped potential for digitalization and enhanced control in mineral processing, the associated benefits and how PGNAA can help.
Digitalization Process Plants
For Research Use Only. Not for use in diagnostic procedures.