For researchers pursuing the next scientific breakthrough, the Thermo Scientific Vanquish Neo UHPLC system delivers maximum performance 24/7 for reproducible nano-, capillary-, and micro-flow LC-MS. Combining a high degree of innovation with the best-in-class Vanquish platform enhances productivity for novice and expert LC-MS users alike.
Highlights
All-in-one nano-, capillary-, and micro-flow LC
Thermo Scientific ProFlow™ XR active flow control pump technology delivers excellent retention time precision from nano up to 100 μL/min at pressures up to 1500 bar. Vial bottom detection enables injections from volume-limited samples.
Accelerated productivity
Fast sample loading, column equilibration and ultra-low system gradient delay volume reduce method overhead time, improving sample throughput while enhancing MS utilization. The tandem direct injection workflow accelerates productivity further by eliminating sample loading, gradient delay, column washing, and column equilibration from method cycle time.
Maximized performance
Unique low-flow split-loop autosampler design limits sample dispersion, reduces system carryover, and maximizes sample usage. The metering device improves quantitation through high injection volume linearity, precision, and accuracy.
Long-term, trouble-free analysis
Next-generation Thermo Scientific SmartInject technology improves column lifetime and retention time precision. Maintenance-free 1500 bar compatible ceramic valves and near-zero dead volume Thermo Scientific Viper and nanoViper fittings minimize leaks with robust long-term operation.
Intelligent operation
The Thermo Scientific Vanquish User Interface provides walk-up and remote system monitoring and control with automated procedures and guided instrument maintenance and troubleshooting scripts, improving productivity for all users. The intelligent method editor wizard ensures compatibility of method parameters with installed consumables.
Seamless LC-MS integration
The Vanquish Neo UHPLC system excels at high-sensitivity LC-MS applications and seamlessly integrates with the Thermo Scientific mass spectrometry portfolio through a comprehensive suite of ESI sources. Harness speed, robustness and sensitivity with triple quadrupole MS systems or resolution, mass accuracy, and versatility with Thermo Scientific Orbitrap MS systems.
Videos
Explore a new dimension in proteomics quantitation
Abstract graphic for LFQ DIA Platform, 1400x200
System specifications
The table below provides an overview of the Vanquish Neo UHPLC system specifications for a standard set up. Not all specifications are listed. For additional information, download the Vanquish Neo UHPLC System specifications sheet.
Category
Vanquish Neo UHPLC System
System pressure, maximum
1500 bar
Flow range
1 nL/min – 100 μL/min, in 1 nL increments 
(100 nL/min – 100 μL/min, recommended)
Pump options (default gradient delay volume)
Binary (< 25 nL)
Biocompatible
Yes
pH range
2 – 10 
System design
Integrated, with slide-in modules
Touchscreen user interface display compatible
Yes
Active flow control for maximum retention time precision
Yes, Thermo Scientific ProFlow XR pump technology
SmartInject technology for maximum injection precision and improved column lifetime
Yes
Injection volume range
Standard: 0.01–25 μL
Optional: 0.01–100 μL
High volume trapping by multiple draw-and-trap cycles up to 500 μL 
Sample cooling
Yes, 4 – 40°C 
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available 
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available (optional)
No
Column compartment thermostatting
Yes (optional) 
Minimum 5 K above room temperature to 80 ⁰C
Forced air
Column compartment pre-heater
Passive (optional)
Column compartment post-column cooler
No
Workflows Direct injection
Tandem direct injection
Trap-and-elute
Heated trap-and-elute
Detector choices
Mass spectrometry
Variable wavelength detector F
Optional add-on components Column Compartment N
Binary Pump N
Contact us to get a quote or discuss an HPLC/UHPLC solution