TAPwithRFID-showcase
One simple step gets you to your data faster
Applied Biosystems TaqMan Array Plates with RFID and QuantStudio 6/7 Pro Real-Time PCR System create an improved user experience with “smart” features that eliminate manual steps currently necessary to upload plate layout and assay information into your experiment file.
Data are seamlessly transferred from the RFID tag to the QuantStudio 6/7 Pro system; TaqMan file download and transfer are no longer necessary.
Get TaqMan gold-standard performance and reproducibility from pre-spotted assays, while leveraging the innovative, time-saving features to improve traceability and reduce manual errors.
Related product
QuantStudio 6/7 Pro Real-Time PCR System
TaqMan Array Plates with RFID can only be used on the QuantStudio 6/7 Pro Real-Time PCR System. Don’t have a QuantStudio 6/7 Pro system?
Learn more ›
QuantStudio7Pro
Features of TaqMan Array Plates with RFID
  • Smart workflow—plate layout and assay information are stored on an RFID tag, which is read by the QuantStudio 6/7 Pro instrument and transferred to the EDS file
  • Efficient data analysis—no need to download TaqMan files and upload a plate layout.  Once your experiment is complete, you can start analyzing your data.
  • Flexible formats—nine different formats are available depending on how many genes you are studying
  • Convenient setup—just add master mix and cDNA
  • Gold-standard performance—high-quality TaqMan Assays are pre-spotted on the plate for superior reproducibility
Workflow benefits
Uploading the plate layout and assay information to your QuantStudio 6/7 Pro system:
RFID-workflow-illustration
There are two methods for ordering TaqMan Array Plates with RFID.
Method 1: order plates online
  1. Configure your TaqMan Array Plate online using the configurator tool.
  2. Once all assays have been added to the plate layout, click "Complete Your Design".
  3. On the subsequent "Custom Array Summary" screen, enter the quantity of plates needed, and check the box to "Add RFID tag(s) for QuantStudio 6/7 Pro" (shown at right).
  4. Click "Add to Cart".
  5. TaqMan Array plates and RFID tags are added to the cart together.
  6. Check out as usual.
Method 2: order plates offline
  1. Ask your Sales Representative to add part number A43823 to your purchase order. (There is no additional charge.)
  2. Email your purchase order, list of Assay IDs, TaqMan Array Plate part number, and quantity to genomicorders@thermofisher.com.
If you don’t have a purchase order prior to contacting Genomic Orders, then be sure to let them know that you want A43823 added to your order.
List of Custom TaqMan Array Plate part numbers currently available with RFID tags
FormatPart number No. of assays + controlsNo. of samples / replicatesMinimum order qty
RFID tagA43823n/an/an/a
TaqMan Array, Standard, 96-well plate; Format 844132667 + 1126
TaqMan Array, Standard, 96-well plate; Format 16441326415 + 166
TaqMan Array, Standard, 96-well plate; Format 16 Plus441326512 + 466
TaqMan Array, Standard, 96-well plate; Format 32439152831 + 136
TaqMan Array, Standard, 96-well plate; Format 32 Plus439152928 + 436
TaqMan Array, Standard, 96-well plate; Format 48439152647 + 126
TaqMan Array, Standard, 96-well plate; Format 48 Plus439152744 + 426
TaqMan Array, Standard, 96-well plate;  Format 96439152495 + 116
TaqMan Array, Standard, 96-well plate; Format 96 Plus439152592 + 416
For Research Use Only. Not for use in diagnostic procedures.