Velox Software for Transmission Electron Microscopy
Multimodal TEM and STEM data acquisition and analysis
All-in-one Thermo Scientific Velox Software was meticulously designed to offer comprehensive and advanced capabilities for both (scanning) transmission electron microscopy and transmission electron microscopy. It provides a fast and efficient means to acquire and analyze multimodal data, ensuring superior experimental control, high reproducibility, and high yield to support quantitative materials analysis.
Velox Software benefits
Learning curve icon
Reduced learning curve
The mode-dependent user interface features graphical layouts that make the software easier to learn and more intuitive.
All data icon
All your data in one program
Collect images and analyze data with popular acquisition workflows optimized for productivity.
Support icon
Professionally developed, maintained, and supported
Get new features and overall improvements through regular software updates.
Advanced energy-dispersive X-ray spectroscopy capabilities
Velox Software is optimized for energy-dispersive X-ray spectroscopy with a robust mapping engine that combines multiple techniques for transmission electron microscopy. It offers:
  • On-the-fly drift correction that ensures accurate data acquisition by correcting for any drift in real time
  • Recursive mapping that enhances the quality and resolution of mapping results
  • Independent channel readout that allows simultaneous data collection from multiple channels
  • Unique absorption correction with holder compensation that ensures precise quantification by compensating for absorption effects
These features help you collect best-in-class EDS spectrum images with high yield and precise quantification. Velox Software is specifically tailored for use with Thermo Scientific Super-X, Dual-X, and Ultra-X Detector Systems, enhancing its EDS capabilities.
Extensive support for electron energy loss spectroscopy
In addition to EDS, Velox Software offers extensive support for electron energy loss spectroscopy. The software facilitates easy and accurate data acquisition and analysis on the Thermo Scientific Iliad (S)TEM’s spectrometer and energy filter, so you can perform detailed EELS analysis with confidence.
Streamlined workflows with an intuitive user interface
Velox Software significantly streamlines complex workflows through its intuitive user interface. The user-friendly design reduces the learning curve associated with advanced STEM and TEM techniques, integrating multiple data acquisition workflows into a single package. This integration enhances the efficiency of scientific research, so you can focus more on your experiments and less on software navigation.
Advanced drift compensation and interactive detector layout
Velox Software incorporates advanced drift compensation methods to ensure high-quality imaging and accurate compositional maps. The interactive detector layout allows for reproducibility and control over experimental parameters, providing flexibility and precision in scientific experiments. Additionally, the software supports flexible STEM and TEM movie recording capabilities, enabling dynamic studies and detailed analysis.
Velox Software features
Velox Software is a powerful tool designed to enhance all materials science experiments. It provides precise and accurate data acquisition, high-quality imaging, and efficient workflows, making it a powerful asset for anyone working with advanced electron microscopy techniques. Velox Software helps you achieve reliable results and democratize your TEM.
Seamless data integration
With Velox Software, all your data is consolidated into one software platform. This integration streamlines your workflow, allowing for efficient data acquisition, analysis, and interpretation. Experience the convenience of having all your research data in one place.
Drift compensation
Velox Software’s advanced drift compensation methods — like drift corrected frame integration (DCFI) and cross correlations in fast recursive mapping — deliver high quality for image and compositional mapping.
Drift compensation Sample courtesy: TU Delft, The Netherlands
Integrated ergonomic user interface
Velox Software’s intuitive, user-friendly interface allows for quick mastery without extensive training. All acquisition controls are grouped together and designed to make every mode and workflow more straightforward. Plus, it provides guidance for challenging imaging conditions and standard live camera overexposure indication.
Integrated ergonomic user interface
Interactive live graphical detector layout
The interactive detector interface and automatic metadata storage deliver greater reproducibility, experimental control, and documentation. You can also easily export your data.
Interactive live graphical detector layout
Time-resolved EDS
Velox Software records and saves every STEM and EDS frame in one experiment dataset. After acquisition, you can use frame removal tools to modify the acquired data cube to minimize artifacts from sample damage and improve data quality for all your imaging and analytical experiments.
Spectrum image data as acquired in Velox Software
EDS quantification software
When performing EDS quantification, Velox Software  compensates for holder shadowing at sample tilt and offers independent detector readouts to help improve accuracy. It also delivers live feedback during acquisition and fast post-processing of EDS data, so you’re always up to date.
EDS quantification software
High-contrast imaging for all elements
With its unique integrated differential phase contrast (iDPC) mode, Velox Software enables simultaneous high-contrast atomic imaging of light and heavy elements at low dose, making it possible to image beam-sensitive materials in their natural state.
High-contrast imaging for all elements
Differential phase contrast on magnetic materials
The differential phase contrast (DPC) mode in Velox Software enables high-contrast imaging of magnetic materials. This mode also makes it possible to directly image complex materials used in data storage and electronic devices and identify a variety of features, including domain wall type, tail-to-tail and head-to-head Bloch lines, wall thickness, and defects.
Differential phase contrast on magnetic materials Courtesy H. Nakajima and S. Mori, Osaka Prefecture University
Live TEM image rotation
Within Velox Software, you can use the mouse to quickly and easily rotate your sample image to get a better view without removing it from the microscope. It also enables automatic stage direction adjustment while keeping all raw data. Both features make it a powerful setup tool for multimodal data collection.
Live TEM image rotation
4D STEM data acquisition
4D STEM acquires all data simultaneously in one experiment, hence data mining to extract desired information. With 4D STEM, you can acquire large datasets of diffraction patterns, enabling detailed spatial and temporal analysis of materials at high resolution.
4D STEM data acquisition
Automated autofocus, astigmatism, auto center and auto tilt
Velox Software automates autofocus, auto astigmatism correction, click auto center, and click auto tilt to help you get the most out of your microscope time. It autosaves data to help prevent loss, exports to common files types, and supports batch exports.
Automated autofocus, astigmatism, auto center and auto tilt
Simultaneous acquisition of EELS and EDX analytical information
Velox Software can acquire EDX and EELS signals simultaneously at a high rate to minimize sample damage and deliver all analytical information in one experiment.
Simultaneous acquisition of EELS and EDX
Stay up to date with Velox Software
Quick and easy installation
Velox Software can be easily installed in just a minute, so you always have access to the latest features and advancements.
No interruption to your workflow
There’s no need to stop the TEM server or restart your PC when installing Velox Software, so you can update your software without any disruption to your ongoing work.
Timely email notifications
You’ll get timely email notifications about new updates and features.
Long-term compatibility
Velox Software is compatible with server versions for approximately a year and a half, providing long-term stability and reliability for your research needs.
Unified installer for online and offline use
The same installer can be used for both online and offline installations, making it convenient and flexible for various installation environments.
Detailed release notes
Each update comes with detailed release notes that describe the new features and improvements so you know exactly what’s changed in Velox Software.
TEM and STEM brochures, datasheets, and eBooks
Velox Software datasheet
Atomic-resolution STEM image of resin-embedded tungsten oxide nanowire imaged using the Thermo Scientific Talos F200X TEM with X-CFEG. Lamella prepared using Thermo Scientific Helios DualBeam FIB-SEM by Kristýna Bukvišová. Bulk material courtesy of Dr. Vojtech Kundrat, Weizmann Institute of Science.
For Research Use Only. Not for use in diagnostic procedures.