Search Thermo Fisher Scientific
Glow discharge mass spectrometers enable you to define new standards in the direct analysis of solid samples, providing you with the solution of choice for high throughput determination of trace elements in high purity conductive and semi-conductive materials.
Providing complete material characterization for high purity metals and alloys, semicon and battery applications, the Thermo Scientific Element GD Plus GD-MS system features a fast glow discharge ion source in a high-resolution mass spectrometer. The ideal tool for the direct analysis of high purity conductive and semi-conductive materials, the system can detect and routinely quantify almost all elements present in a solid sample at or below the ppb range.
Redefine the analysis of advanced high purity materials directly in solid state with the Thermo Scientific Element GD Plus GD-MS. For high through-put and extra low ppb level detection limits, the Element GD Plus GD-MS is the most convenient and powerful tool for bulk metal analysis and depth profiling in routine and research applications.
The fast flow source offers uniquely high sensitivity and low levels of interferences. Additionally, the fast flow source enables rapid pump down and sample acquisition times, dramatically improving sample through-put and lab productivity.
The simplicity of high-resolution ICP-MS allows the most advanced performance, without compromising on straightforward and reliable method development. The Element GD Plus GD-MS provides interference free measurements, resulting in simple linear calibration curves for quantification. Three fixed resolution settings (300, 4,000, 10,000), with switching times of <1 s, ensure optimum conditions to reliably remove spectral interferences. The fixed-slit design guarantees maximum stability and reproducibility.
Typical applications involve the simultaneous analysis of matrix elements (%), traces (ppm) and ultra-traces (ppb). To achieve this, the detection system offers a fully automatic detector with >12 orders of linear dynamic range with automatic cross calibration between different detection modes.
The quantification of gas phase elements like trace carbon, nitrogen and oxygen within materials has traditionally been accomplished through combustion techniques, requiring a second analytical instrument. The CNO option of the Element GD Plus GD-MS cleans the argon discharge gas through purification units adsorbing moisture, carbon dioxide and nitrogen traces from the argon gas supplied. This allows sub-ppm limits of detection to be achieved for carbon, nitrogen and oxygen.
After finishing the sample run, it takes just one push to open the Glow Discharge source. No tools are required.
Even the most sophisticated elemental analysis is made more straightforward with the right GD-MS accessories. Improve performance, convenience and flexibility with accessories designed specifically for your glow discharge mass spectrometer.
The CNO option cleans the argon discharge gas through purification units adsorbing moisture, carbon dioxide and nitrogen traces from the argon gas supplied, allowing sub-ppm limits of detection to be achieved for C, N, and O.
High through-put analysis of ultra-trace impurities in high purity copper made simple with the Element GD Plus GD-MS.
Learn about the capabilities of the Element GD Plus µs-Fast-Flow Glow Discharge Mass Spectrometer for high throughput trace metal determination in high purity alumina powders with minimum sample preparation.
The Element GD Plus GD-MS features a fast flow glow discharge source that can be operated in continuous or pulsed mode. Samples with a lower melting point can be analyzed in pulsed mode to avoid melting.
Element GD Plus GD-MS provides highly precise trace metal detection in nickel super alloys. Its lower sputter rate than non-pulsed mode ensures excellent spot-to-spot and per-spot accuracy.
Trace contaminations in 7N high purity gallium phosphide can be determined reliably and routinely with the Element GD PLUS GD-MS without prior digestion.
Use isotopic fingerprints in food, fiber, liquid, or stone to track origins, authenticity, and identity. IRMS identifies isotopic signatures for geographic, botanical, soil, and fertilization information.
*Required field