Search Thermo Fisher Scientific
See it fast and identify it faster with the Nicolet RaptIR +/RaptIR FTIR Microscope
In analytical labs where details matter most—when productivity is measured in microns and minutes—turn to the Thermo Scientific Nicolet RaptIR or RaptIR + FTIR Microscope for detailed answers.
These research-grade microscopes help you rapidly locate and identify trace materials, inclusions, impurities, microparticles, and determine the distribution of materials within a sample with incredible clarity and precision. Its clean, adaptive user interface provides increased flexibility and usability for all users, from specialists to novices. The Nicolet RaptIR and RaptIR + FTIR Microscopes provide answers quickly, regardless of industry or application.
Expanding your options, the RaptIR + adds the versatility of user-swappable detectors offering extended-range operation or liquid-nitrogen-free operation. When adapted to interface with a Nicolet iS50 FTIR Spectrometer, the RaptIR + enables a wide range of new analysis possibilities, including TGA-IR, GC-IR, NIR, FT-Raman, and much more. For a more cost-conscious choice, the RaptIR + can also adapt to the Nicolet Apex FTIR Spectrometer.
Nicolet RaptIR + FTIR Microscope | Nicolet RaptIR FTIR Microscope | |
---|---|---|
Visible resolution | 1 micron | 1 micron |
Infrared resolution | 5 micron | 5 micron |
Multiple detectors | Swappable, MCT-A, MCT-B, InGaAs | Fixed, MCT-A, MCT-B |
Spectrometer option | Nicolet iS50 FTIR Spectrometer (right or left side) Nicolet Apex FTIR Spectrometer (right side) |
Nicolet iS50 FTIR Spectrometer (right side) |
Objectives | 4X or 40X visible, 15X or grazing angle infrared | 4X or 40X visible, 15X or grazing angle infrared |
Polarizers | Motorized, visible and IR | Motorized, visible and IR |
From the mosaic feature to infrared capability and a seamless user experience, the Nicolet RaptIR and RaptIR + FTIR Microscopes are full of technological innovations that help specialists and non-specialists alike.
The Nicolet RaptIR and RaptIR + FTIR Microscopes are always coupled to a Thermo Scientific Nicolet FTIR Spectrometer. Using it with the Thermo Scientific Nicolet iS50 FTIR Spectrometer provides a broader array of capabilities—including FT-Raman, TGA-IR, NIR, far-infrared, and diamond ATR—for addressing complex analytical challenges. This combination will help you find answers more quickly, whether you are studying powders, liquids, gases, or micron-sized particulates.
The Nicolet RaptIR +/RaptIR FTIR Microscope automatically ensures you can use it anywhere, anytime.
The versatility of the RaptIR +/RaptIR allows you to analyze across a wide range of applications:
The software’s clean, adaptive user interface allows specialists and novice users alike to obtain the data they’re looking for. Workflow automation, combined with the ability to select multiple regions of interest in a sample for collection in one pass, helps save time and reduce errors. Autofocus and automated illumination allow you to get to data collection quickly and easily. Fully customizable microparticles analysis workflow allows users of varying experience to click, get images, and move forward, choosing what they want to show in tailored reports.
See for yourself in these short videos how easy it is to use the Nicolet RaptIR +/RaptIR FTIR Microscope to find a sample, collect a mosaic, or collect a background and map:
For Research Use Only. Not for use in diagnostic procedures.