Search Thermo Fisher Scientific
As a leader in PCR innovation, we offer you the gold standard in real-time PCR instrumentation. With our Applied Biosystems real-time PCR platforms, you get true value with excellent performance, reliability, and world-class support. Designed for compatibility with Applied Biosystems TaqMan Assays and the flexibility of unlimited real-time chemistry choices, we make it easy for you to get started with your experiments. The Applied Biosystems QuantStudio family of instruments enables you to obtain the results you need, connect and collaborate with colleagues, and achieve your research goals.
Request a quote Download brochure Financing Programs
Looking to commercialize? Contact our OEM services team today ›
Simple to use for all levels of experience | Easy controls for optimized experiments | |
Request a quote › | Request a quote › | |
Reactions per Run | 96 | 96 or 384 |
Block Format | 96-Well 0.2-mL or 96-Well Fast 0.1-mL | 96-Well 0.2-mL, 96-Well Fast 0.1-mL, or 384-Well |
Color Combinations (Multiplexing) | Up to 4 | 96-well up to 21 384-well up to 5 |
Run Time | As fast as 30 min | As fast as 30 min |
Programmable Volumes | ||
Validated volumes | 10-100 µl (96-well 0.2mL) 10-30 µl (96-well 0.1mL) | 10-100 µl (96-well 0.2mL) 10-30 µl (96-well 0.1mL) 5-20 µl (384-well) |
Automation Compatible | No | No |
Remote Monitoring | ||
On-Board Memory | ||
Email Notification | ||
Assist with 21 CFR Part 11 Compliance | Security, auditing, e-signature package included | |
Sensitivity | Down to 1 copy | |
Dynamic Range | Up to 10 logs** | |
Chemistry | All real-time PCR-based chemistries. Flexibility for chemistries with or without passive reference dye | |
Applications Available: | ||
GeneExpression | ||
Genotyping | ||
Copy Number Variation | ||
Protein Detection | ||
MicroRNA | ||
Pathogen Detection | ||
HRM | ||
IVD Platform | No | Available in certain countries |
Temperature Uniformity | 0.4 "degree symbol"C | |
Excitation Source | Bright white LED | |
Filters/Colors | 4 excitation filters (450–600 nm) 4 emission filters (500–640 nm) | 96-well: 6 excitation filters (450–670 nm) 6 emission filters (500–720 nm) 384-well: 5 excitation filters (450–650 nm) 5 emission filters (500–700 nm) |
Cloud-enabled system | Yes | |
Wi-Fi Connectivity | Yes | |
Size | 27 x 50 x 40 cm | |
Request a quote › | Request a quote › |
High flexibility and intuitive design | Smart and connected | High versatility and intuitive design | Smarter productivity | Maximum throughput, flexibility, and scalability | |
Request a quote › | Request a quote › | Request a quote › | Request a quote › | Request a quote › | |
Colors | 5 colors with OptiFlex | 5 colors with OptiFlex | 6 colors with OptiFlex (21 filter combinations) | 6 colors with OptiFlex (21 filter combinations) | 6 colors with OptiFlex (21 filter combinations) |
Reactions per Run | 96 or 384 | 96 or 384 | 96 or 384 | 96 or 384 | 96, 384, or 12,228 |
Interchangeable block formats | 96-well (0.1 and 0.2 mL), 384-well | 96-well (0.2 mL), 384-well | 96-well (0.1 and 0.2 mL), 384-well, TaqMan Array | 96-well (0.2 mL), 384-well | 96-well (0.1 and 0.2 mL), 384-well, TaqMan Array, OpenArray |
Throughput | High | High | High | High | Very High |
Dimensions (H x W x D) | 73.8 x 50.1 x 66.1 cm | 52.5 x 33.8 x 54.7 cm | 73.8 x 50.1 x 66.1 cm | 52.5 x 33.8 x 54.7 cm | 73.8 x 50.1 x 66.1 cm |
VeriFlex temperature control | N/A | Yes, 3 zones | N/A | Yes, 6 zones | N/A |
Excitation Source | Halogen lamp | Bright white LED | Halogen lamp | Bright white LED | White LED |
Sensitivity | Down to 1 copy | ||||
Dynamic Range | Up to 10 logs** | ||||
21 CFR Part 11-enablement | Optional security, auditing, e-signature package | No | Optional security, auditing, e-signature package | Security, auditing, e-signature package † | Optional security, auditing, e-signature package |
Touch screen | Yes | Yes, interactive | Yes | Yes, interactive | Yes |
Cloud-enabled system | No | Yes | No | Yes | No |
Run Time | As fast as 30 min | ||||
Chemistry | All real-time PCR-based chemistries. Flexibility for chemistries with or without passive reference dye. | ||||
Applications Available: | |||||
GeneExpression | |||||
Genotyping | |||||
Copy Number Variation | |||||
Protein Detection | |||||
MicroRNA | |||||
Pathogen Detection | |||||
HRM | |||||
IVD Platform | No | No | Available in certain countries | No | No |
Request a quote › | Request a quote › | Request a quote › | Request a quote › | Request a quote › | |
*Duplex capabilities with OpenArray and multiplex capabilities for other formats. |
Powerfully Simple Digital PCR | |
Request a quote › | |
Reactions per run | 16 per plate |
Format | 20,000 microchambers per sample |
Color combinations (multiplexing) | 4 (endpoint detection) |
Run time | ~90 minutes per run |
Validated volumes | 9 µL per reaction |
Assists with 21 CFR Part 11 compliance? | Yes |
Sensitivity | Down to 1 copy |
Dynamic range | 5 logs |
Chemistry | TaqMan |
Applications available | Rare mutation detection, quantification of molecular standards, copy number variation, absolute quantification, pathogen detection, digital PCR |
IVD platform | No |
Size | W 60 cm D 62 cm H 54 cm |
Request a quote › |
Warranty information | ||
1-year standard warranty | ||
Service plans available post warranty | ||
AB Complete | ||
AB Assurance | ||
AB Maintenance | ||
Planned Maintenance | ||
Compliance services | ||
IQ/OQ/PQ or IPV | ||
OQ/PQ or IPV |
Warranty information | |||||
1-year standard warranty | |||||
Service plans available post warranty | |||||
AB Complete | |||||
AB Assurance | |||||
AB Maintenance | |||||
Planned Maintenance | |||||
Compliance services | |||||
IQ/OQ/PQ or IPV | |||||
OQ/PQ or IPV | |||||
CSV |
Warranty information | |
1-year standard warranty | |
Service plans available post warranty | |
AB Assurance | |
Smart Start orientation | |
Customer Concierge services | |
Rapid Exchange | |
Compliance services | |
IQ/OQ/IPV | |
OQ/IPV | |
SAE IQ/OQ |
QuantStudio 1*, 3 & 5 systems | QuantStudio 6 & 7 Pro systems | |
Instrument status | ||
Monitor run over smart phone (ICMA) | ||
Experiment Design App (DA) | ||
Data analysis modules (SC, RQ, PA, GT, HRM) | ||
Voice commands | ||
Smart Help | ||
Smart remote support | ||
Instrument scheduler |
*QuantStudio 1 system is not available in the US and Canada.
Learn more about connected instruments and how they can help to accelerate your science.
Find the parts you need as well as part numbers for our legacy real-time PCR systems.
A wide range of software applications to help you get the most from your Applied Biosystems real-time PCR system.
Select your real-time PCR (qPCR) instrument from the drop-down menu, and easily find the plates, optical caps, and optical seals compatible with your instrument.
Applied Biosystems Spectral Calibration Kits and TaqMan® RNase P Instrument Verification Plates to help calibrate and verify the performance of your real-time PCR system.
Looking for a thermal cycler for end-point PCR?
For Research Use Only. Not for use in diagnostic procedures.