Reproductive Health Research Using Next-Generation Sequencing
Understanding risk factors for inherited genetic disorders is an important step in evaluating reproductive options. Expanded carrier screening (ECS) research and preimplantation genetic testing (PGT) offer critical insights that may increase probability of success for future pregnancies and healthier future generations.
Next-generation sequencing (NGS) has revolutionized reproductive health research with a high-throughput technology for rapidly sequencing DNA, with high accuracy. NGS has expanded both our menu of known genetic variants and our capacity to identify them. Ion Torrent NGS solutions deliver scalable, precise sequencing to better support reproductive health outcomes.
Take a tour of a reproductive health genetics research lab: solutions for every step in the workflow.
Video Player is loading.
Current Time 0:00
Duration 1:59
Loaded: 8.22%
Stream Type LIVE
Remaining Time 1:59
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Note: Each workflow may require different instruments. Contact your local representative to build the right solution for your needs.
    Expanded carrier screening research
    Uncovering genetic risk and carrier status data has proven helpful in providing scientific insights for reproductive options. While traditional carrier screening only targets single gene disorders according to ancestry or family history, ECS by NGS screens for genetic disorders that occur at increasing frequency in broader populations.
    Ion Torrent CarrierSeq ECS Kit
    This 420-gene panel enables the analysis of >36,000 non-benign ClinVar variants for single-nucleotide variants (SNVs), insertions and deletions (indels), and copy number variants (CNVs)—all with a single, consolidated NGS assay.
    Preimplantation genetic testing
    When in-vitro fertilization (IVF) is identified as a likely path to successful pregnancy, Preimplantation genetic testing (PGT) provides testing labs with genetic information in the research of embryo prioritization.
    Ion ReproSeq PGS kit + PGD-SEQ kit
    The Ion ReproSeq PGS kit for PGT-A and PGD-SEQ kit for PGT-M enable simultaneous research of chromosomal abnormalities and monogenic disorders for IVF with a single sample and one convenient NGS workflow.
    These comprehensive, end-to-end genetic analysis solutions include reagents for library preparation, template preparation, and sequencing, plus simple, integrated data analysis and reporting tools, making results accessible, regardless of NGS or bioinformatics expertise.
    Ion Torrent Genexus System
    oca-plus-page-genexus
    The Genexus Integrated Sequencer simplifies templating, sequencing, analyzing, and reporting for PGT-A samples. Perform premium PGT-A with Ion ReproSeq™ and Ion AmpliSeq Polyploidy Panel kits, now available for 192 or 384 samples. With workflow automation, sequencing performance, and automated data analysis and reporting, the Genexus System makes NGS accessible to labs without the need for extensive expertise in sequencing or bioinformatics. From embryo biopsy samples or spent cell culture media, you can obtain analyzed data in under 11 hours for up to 48 samples.
    Ion GeneStudio S5 System
    ion-genestudio-5
    Ion GeneStudio S5 systems provide simple, sample-to-data workflows for targeted sequencing, with industry-leading speed and scalability. Five Ion S5 chips enable a sequencing throughput range of 2M to 130M reads per run. Combined with the Ion Chef System, this complete solution provides a walk-away workflow that includes automated Ion AmpliSeq library preparation and reproducible template preparation and chip loading, with less than 45 minutes of hands-on time. All Ion Torrent NGS workflows are backed by application-specific analysis and reporting solutions.
    For Research Use Only. Not for use in diagnostic procedures.
    CN: 64186