This section presents a selected set of primary antibodies for detecting expression tags, including Green Fluorescent Protein (GFP) and Red Fluorescent Protein (RFP) as well as oligohistidine, hemagglutinin and c-myc. The full range of Invitrogen primary antibodies can be surveyed using our online primary antibody search utility at www.invitrogen.com/handbook/antibodies.
Direct labeling of primary antibodies can be accomplished via amine-reactive chemistry implemented in our comprehensive selection of convenient and easy-to-use protein labeling kits (Kits for Labeling Proteins and Nucleic Acids—Section 1.2). Alternatively, Zenon immunolabeling technology (Zenon Technology: Versatile Reagents for Immunolabeling—Section 7.3) can be used to prepare fluorophore-, biotin- or enzyme-labeled primary antibodies but is less demanding than chemical labeling methods in terms of antibody quantity and purity requirements.
Antibodies Specific for Reporter Gene Products
Anti–Green Fluorescent Protein (GFP) Antibodies
Expression of the intrinsically fluorescent Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria has become a popular method for following gene expression and protein localization.ref We offer a rabbit polyclonal antibody that is raised against GFP purified directly from A. victoria. This anti-GFP antibody, which is available as a complete serum (A6455) or as an IgG fraction (A11122) purified by ion-exchange chromatography, facilitates the detection of native GFP, recombinant GFP and GFP fusion proteins by immunofluorescence ref (photo), western blot analysis ref and immunoprecipitation. Several direct conjugates derived from the IgG fraction are also available:
  • Green-fluorescent Alexa Fluor 488 rabbit anti-GFP antibody (A21311)
  • Orange-fluorescent Alexa Fluor 555 rabbit anti-GFP antibody (A31851)
  • Red-fluorescent Alexa Fluor 594 rabbit anti-GFP antibody (A21312)
  • Far-red–fluorescent Alexa Fluor 647 rabbit anti-GFP antibody (A31852)
  • Biotin-XX rabbit anti-GFP antibody (A10259)
  • Horseradish peroxidase (HRP) rabbit anti-GFP antibody (A10260)
In addition, we offer an affinity-purified IgY fraction of chicken anti-GFP antibody, both unlabeled (A10262) and as the biotin-XX conjugate (A10263). Chicken IgY lacks a classic Fc domain and therefore does not bind to mammalian IgG Fc receptors, resulting in lower backgrounds during immunostaining protocols. The chicken IgY is also antigenically different from the mammalian IgG, allowing antibodies from both species to be used in a single immunostaining experiment.
Lastly, we offer a rabbit monoclonal anti-GFP antibody (G10362), as well as two mouse monoclonal anti-GFP antibodies. The rabbit monoclonal anti-GFP antibody is raised against full-length GFP and is suitable for applications such as ELISAs, immunohistochemistry, immunocytochemistry, immunoprecipitation and western blotting. Mouse monoclonal 11E5 (anti-GFP, clone 11E5; A11121) is optimized for western analysis, allowing colorimetric detection of as little as 10 ng of GFP or GFP-fusion proteins. Mouse monoclonal 3E6 (anti-GFP, clone 3E6; A11120) is useful for immunoprecipitation, immunocytochemical localization and immunosorbent assays (ELISAs).
Anti–Red Fluorescent Protein (RFP) Antibody
We offer an affinity-purified rabbit polyclonal anti–Red Fluorescent Protein antibody (anti-RFP, R10367) for western blot and immunocytochemistry applications. Full-length recombinant denatured and nondenatured TagRFP was used as the immunogen, making the antibody suitable for immunocytochemical detection of the expression products from our CellLight RFP BacMam vectors (CellLight reagents and their targeting sequences—Table 11.1, BacMam Gene Delivery and Expression Technology—Note 11.1).
Anti–Glutathione S-Transferase Antibody
One common partner in protein fusions is glutathione S-transferase (GST), a protein with natural binding specificity for glutathione that can be exploited to facilitate its purification.ref We prepare a highly purified rabbit polyclonal anti-GST antibody (A5800) that can be used for western blot analysis, immunodetection and immunoprecipitation of GST fusion proteins.ref This highly specific and high-titer antibody, which was generated against a 260–amino acid N-terminal fragment of the Schistosoma japonica enzyme expressed in Escherichia coli, is particularly useful for detecting GST distribution in cells.ref To facilitate the localization of GST and GST-fusion proteins using immunofluorescence techniques, we prepare the anti-GST antibody labeled with our green-fluorescent Alexa Fluor 488 dye (A11131).
Because the GST portion of the fusion protein retains its affinity and selectivity for glutathione, the fusion protein can be conveniently purified from the cell lysate in a single step by affinity chromatography on glutathione agarose ref (Figure 7.5.1). For the purification of GST fusion proteins, we offer glutathione linked via the sulfur atom to crosslinked beaded agarose, which is available as 10 mL of a sedimented bead suspension (G2879, Detecting Protein Modifications—Section 9.4).
polyacrylamide gel 
Figure 7.5.1 Coomassie brilliant blue–stained SDS-polyacrylamide gel, demonstrating the purification of a glutathione S-transferase (GST) fusion protein using glutathione agarose (G2879). Lane 1 contains crude supernatant from an Escherichia coli lysate and lane 2 contains the affinity-purified GST fusion protein.
Anti–β-Glucuronidase Antibody
The E. coli β-glucuronidase (GUS) gene (uidA) is a popular reporter gene in plants.ref For western blot and immunohistochemical analysis of transformed plant tissue ref and transfected animal cells,ref we offer unlabeled rabbit anti–β-glucuronidase antibody (A5790) raised against E. coli type X-A β-glucuronidase. Our fluorogenic and chromogenic β-glucuronidase substrates are described in Detecting Glycosidases—Section 10.2.
Anti–β-Galactosidase Antibody
We offer a polyclonal antibody to the widely used reporter gene product β-galactosidase. Our rabbit anti–β-galactosidase antibody (A11132) is raised against E. coli–derived β-galactosidase and demonstrates high specificity for the enzyme. Whether the enzyme is used as a reporter or as a fusion protein, this antibody provides an easy tool for detecting the β-galactosidase protein. The antibody is suited to a variety of techniques, including immunoblotting, ELISA and immunoprecipitation. Our fluorogenic and chromogenic β-galactosidase substrates are described in Detecting Glycosidases—Section 10.2.
Antibodies Specific for Epitope Tags
Epitope tagging is a powerful and versatile strategy for detecting and purifying proteins expressed by cloned genes. Protein expression vectors are often engineered with a nucleotide sequence that encodes a peptide epitope tag. Typically, a gene is cloned in-frame relative to the epitope tag such that, upon expression, a fusion protein containing the epitope tag is synthesized. Detection and purification of the epitope-tagged fusion protein can be mediated by antibodies to the engineered peptide, thus eliminating the need for antibodies to proteins from each newly cloned gene. Here we highlight a series of antibodies to common epitopes, including oligohistidine, hemagglutinin (HA) and c-myc, but a more comprehensive list can be found using our online primary antibody search utility at www.invitrogen.com/handbook/antibodies.
Penta·His Antibody
The oligohistidine domain is a Ni2+-binding peptide sequence comprising a sequence of four to six histidine residues.ref When the DNA sequence corresponding to an oligohistidine domain is fused in-frame with a gene of interest, the resulting recombinant fusion protein can be easily purified using a nickel-chelating resin.ref
In collaboration with QIAGEN, we offer a highly selective antibody to oligohistidine fusion proteins (Penta·His mouse monoclonal antibody, P21315). The antibody is useful for detecting sequences of five or six histidine residues, whether present at the C-terminus, the N-terminus or an internal position in a protein, in both western blots and in immunohistochemistry.
The Penta·His antibody can detect as little as 1–2 ng in western blot applications using fluorescent or chemiluminescent development techniques (photo); the supplied amount (100 µg) is sufficient for 50–100 minigel blots. This antibody does not recognize tetrahistidine domains or domains in which the histidine string is interrupted by another amino acid. The antibody binds to the oligohistidine domain regardless of the surrounding amino acid context and even when the group is partially hidden, although subtle differences in the amino acid context may change the sensitivity limit for a particular fusion protein. The Penta·His antibody is directly useful for immunoprecipitation, ELISAs and immunohistochemistry.
Anti-HA Antibody
Our mouse monoclonal anti-hemagglutinin (HA) antibody was raised against the 12–amino acid peptide CYPYDVPDYASL. It recognizes the influenza hemagglutinin epitope YPYDVPDYA, which has been used extensively as a general epitope tag in expression vectors.ref The extreme specificity of this antibody permits unambiguous identification and quantitative analysis of the tagged protein. This antibody is effective in immunoblotting, immunofluorescence and immunoprecipitation of tagged proteins. We offer Alexa Fluor 488 (A21287) and Alexa Fluor 594 (A21288) conjugates of the anti-HA antibody for those applications that require a directly conjugated antibody.
Anti–c-myc Antibody
We offer two antibodies to c-myc, which is commonly used in epitope tagging.ref Our mouse monoclonal anti–c-myc antibody (IgG1,κ, clone 238-19510; A21280) was raised against the peptide AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA, which corresponds to amino acids 408–439 of the human c-myc protein. The chicken anti–c-myc antibody (IgY, A21281) was raised against the peptide EQKLISEEDL. These antibodies specifically react with the C-terminal epitope (AEEQKLISEEDL) ref of human c-myc protein encoded in many expression vectors.
Ordering Information
Catalog # Name Size Price (USD) Qty
A11133 Bovine Serum Albumin Polyclonal Antibody 500 µL
358.00
A6428 Carboxypeptidase Y Monoclonal Antibody (10A5B5) 250 µg
664.00
A21281 Myc Tag Polyclonal Antibody 100 µL
610.00
A21280 c-Myc Monoclonal Antibody (289-19510) 100 µL -
A6429 DPM1 Monoclonal Antibody (5C5A7) 250 µg
658.00
A11132 beta Galactosidase Polyclonal Antibody 500 µL
255.00
A21282 GFAP Monoclonal Antibody (131-17719) 100 µL
516.00
A21294 GFAP Monoclonal Antibody (131-17719), Alexa Fluor™ 488 50 µL
520.00
A21295 GFAP Monoclonal Antibody (131-17719), Alexa Fluor™ 594 50 µL
516.00
A5790 GUSB Polyclonal Antibody 500 µL
440.00
A5800 GST Tag Polyclonal Antibody 500 µL
492.00
A11131 GST Tag Polyclonal Antibody, Alexa Fluor™ 488 500 µL
520.00
A21270 Golgin-97 Monoclonal Antibody (CDF4) 100 µg -
A10262 GFP Polyclonal Antibody 100 µL
556.00
A10263 GFP Tag Polyclonal Antibody, Biotin 100 µL -
A11120 GFP Monoclonal Antibody (3E6) 100 µg
514.00
A11121 GFP Monoclonal Antibody (11E5) 100 µg -
A11122 GFP Polyclonal Antibody 100 µL
556.00
A21311 GFP Polyclonal Antibody, Alexa Fluor™ 488 100 µL
564.00
A31851 GFP Polyclonal Antibody, Alexa Fluor™ 555 100 µL
564.00
A21312 GFP Polyclonal Antibody, Alexa Fluor™ 594 100 µL
520.00
A31852 GFP Polyclonal Antibody, Alexa Fluor™ 647 100 µL
446.00
A10259 GFP Polyclonal Antibody, Biotin 100 µL
492.00
A10260 GFP Polyclonal Antibody, HRP 200 µg
508.00
A6455 GFP Polyclonal Antibody 100 µL
424.00
A6427 VMA2 Monoclonal Antibody (13D11B2) 250 µg
658.00
A21287 HA Tag Monoclonal Antibody (16B12), Alexa Fluor™ 488 100 µL
642.00
A21288 HA Tag Monoclonal Antibody (16B12), Alexa Fluor™ 594 100 µL
632.00
A21271 HuC/HuD Monoclonal Antibody (16A11) 100 µg
488.00
A21272 HuC/HuD Monoclonal Antibody (16A11), Biotin 100 µg
546.00
A21277 HuR Monoclonal Antibody (19F12) 100 µg
474.00
A6473 NMDAR2A Polyclonal Antibody 10 µg
626.00
A6474 NMDAR2B Polyclonal Antibody 10 µg
722.00
A21359 NDUFB6 Monoclonal Antibody (21C11BC11) 100 µg -
A6404 MTCO2 Monoclonal Antibody (12C4F12) 100 µg
504.00
A21347 OxPhos Complex IV subunit IV Monoclonal Antibody (10G8D12C12) 100 µg
610.00
A21348 OxPhos Complex IV subunit IV Monoclonal Antibody (20E8C12) 100 µL
582.00
A21355 ATPIF1 Monoclonal Antibody (5E2D7) 100 µg
495.00
A21351 ATP Synthase beta Monoclonal Antibody (3D5AB1) 100 µg
632.00
A21273 PEP12 Monoclonal Antibody (2C3G4) 50 µg -
A21327 Phosphatidylinositol 4,5-bisphosphate Monoclonal Antibody (2C11) 100 µL -
A21328 Phosphatidylinositol 3,4,5-trisphosphate Monoclonal Antibody (RC6F8) 100 µL
598.00
A21326 PDH subunit E2 / E3bp Monoclonal Antibody (13G2AE2BH5) 100 µg -
A6442 Synapsin 1 Polyclonal Antibody 10 µg
570.00
A11130 Transferrin Receptor Monoclonal Antibody (236-15375) 50 µg -
A11126 alpha Tubulin Monoclonal Antibody (236-10501) 50 µg
424.00
A21371 alpha Tubulin Monoclonal Antibody (236-10501), Biotin 50 µg
481.00
A23210 APO-BrdU™ TUNEL Assay Kit, with Alexa Fluor™ 488 Anti-BrdU Each
814.00
Q10013 CD14 Monoclonal Antibody (TüK4), Qdot™ 605 100 µL
756.00
Q10056 CD14 Monoclonal Antibody (TuK4), Qdot™ 655 100 µL
758.00
Q10064 CD14 Monoclonal Antibody (TuK4), Qdot™ 800 100 µL
766.00
Q10065 CD27 Monoclonal Antibody (CLB-27/1), Qdot™ 605 100 µL
754.00
Q10066 CD27 Monoclonal Antibody (CLB-27/1), Qdot™ 655 100 µL
728.00
Q10012 CD3 Monoclonal Antibody (S4.1), Qdot™ 655 100 µL
742.00
Q10053 CD38 Monoclonal Antibody (HIT2), Qdot™ 605 100 µL -
Q10051 CD45 Monoclonal Antibody (HI30), Qdot™ 605 100 µL
752.00
Q10062 CD45 Monoclonal Antibody (HI30), Qdot™ 705 100 µL -
Q10060 CD4 Monoclonal Antibody (S3.5), Qdot™ 705 100 µL
752.00
Q10008 CD4 Monoclonal Antibody (S3.5), Qdot™ 605 100 µL
772.00
Q10007 CD4 Monoclonal Antibody (S3.5), Qdot™ 655 100 µL
772.00
Q10009 CD8 Monoclonal Antibody (3B5), Qdot™ 605 100 µL
758.00
Q10055 CD8 Monoclonal Antibody (3B5), Qdot™ 655 100 µL
754.00
Q10059 CD8 Monoclonal Antibody (3B5), Qdot™ 705 100 µL
770.00
Q10156 CD45 Monoclonal Antibody (HI30), Qdot™ 800 100 µL
772.00
Q10153 CD10 Monoclonal Antibody (MEM-78), Qdot™ 605 100 µL -
Q10172 CD2 Monoclonal Antibody (S5.5), Qdot™ 605 100 µL
746.00
Q10179 CD19 Monoclonal Antibody (SJ25-C1), Qdot™ 655 100 µL
764.00
Q10069 CD45RA Monoclonal Antibody (MEM-56), Qdot™ 655 100 µL
766.00
Q10047 CD45RA Monoclonal Antibody (MEM-56), Qdot™ 605 100 µL
742.00
Q10092 CD4 Monoclonal Antibody (RM4-5), Qdot™ 605 100 µL
624.00
Q10176 CD45R Monoclonal Antibody (RA3-6B2), Qdot™ 655 100 µL
584.00
Q10054 CD3e Monoclonal Antibody (UCHT1), Qdot™ 605 100 µL
752.00
Q10052 HLA-DR Monoclonal Antibody (TU36), Qdot™ 605 100 µL
738.00
Q10063 HLA-DR Monoclonal Antibody (Tü36), Qdot™ 800 100 µL -
Q10076 Mouse IgG2a Isotype Control, Qdot™ 705 100 µL -
Q10014 Mouse IgG2a Isotype Control, Qdot™ 605 100 µL
363.00
Q10015 Mouse IgG2a Isotype Control, Qdot™ 655 100 µL
350.00
P21315 Penta-His Tag Monoclonal Antibody 100 µg
1,002.00
For Research Use Only. Not for use in diagnostic procedures.