Images of an NIH 3T3 cell showing colocalization of the CoroNa™ Red sodium indicator with MitoTracker® Green FM.
Images of an NIH 3T3 cell showing colocalization of the CoroNa™ Red sodium indicator (left panel; Cat. no. C24430, C24431) with the MitoTracker® Green FM mitochondrial marker (right panel, M7514). A cell loaded with both dyes was imaged consecutively using Omega Optical bandpass filter set XF41 for CoroNa™ Red sodium indicator and set XF23 for MitoTracker® Green FM.
Images of an NIH 3T3 cell showing colocalization of the CoroNa™ Red sodium indicator (left panel; Cat. no. C24430, C24431) with the MitoTracker® Green FM mitochondrial marker (right panel, M7514). A cell loaded with both dyes was imaged consecutively using Omega Optical bandpass filter set XF41 for CoroNa™ Red sodium indicator and set XF23 for MitoTracker® Green FM.
Related Products
Catalog # Name Size Price (USD) Qty
M7514 MitoTracker™ Green FM - Special Packaging, 20 x 50 μg, 20 x 50 μg Each
504.65
Online Exclusive
520.00
Save 15.35 (3%)
Related Images