End-to-end solution for your MS-based Protein Footprinting higher order structural analysis
Our entire workflow has been meticulously optimized to provide you with the highest level of insight, eliminating any uncertainty when it comes to selecting the most suitable configuration for your experiments. We offer a comprehensive solution that encompasses all necessary components, along with a wide range of resources to support you throughout the entire workflow.
Separations optimized for MS-based Protein Footprinting
Vanquish Neo UHPLC System
For experiments that require high reproducibility and sensitivity with low carryover. The Thermo Scientific Vanquish Neo UHPLC System enables high-sensitivity LC-MS applications. It flawlessly integrates with the industry-leading Thermo Scientific mass spectrometry portfolio and ESI sources.
Vanquish Neo UHPLC System
EASY-Spray PepMap Neo UHPLC Columns
Thermo Scientific EASY-Spray PepMap Neo UHPLC Columns feature easy connectivity, high reproducibility, and excellent separations. Neo columns provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The click-and-spray format enables perfect connections to our mass spectrometers every time.
EASY-Spray PepMap Neo UHPLC Columns
FAIMS Pro Duo interface - For experiments that require extended dynamic range
The Thermo Scientific FAIMS Pro Duo interface offers improved selectivity and sensitivity, even at very low picogram loading levels. This enables the generation of deeper sequence coverage, allowing for the detection of subtle differences in low abundance relevant modification sites. The advanced high field asymmetric ion mobility technology employed by this interface enables precise targeting of peptides within the ion cloud, while effectively maintaining the cleanliness of the mass spectrometer and ensuring the purity of the spectra.
FAIMS Pro Duo interface
Mass spectrometers for Protein Footprinting
The Thermo Scientific Orbitrap family of mass spectrometers offers reliable, proven technology to support MS-based Footprinting analysis with accuracy and precision.
Differentiating features of all Orbitrap instruments
  • Automatic gain control to enable consistent spectral quality and quantitative accuracy from picogram to microgram loading levels without user intervention
  • Normalized collision energy to enable reproducible fragmentation efficiency of all precursors without optimization within and across sample cohorts
  • Complete mass range (m/z) of MS/MS scans to enable unambiguous sequence annotation
  • Exceptional spectral quality and intra-spectrum dynamic range enabled by the Orbitrap mass analyzer
Orbitrap Astral Mass Spectrometer - For experiments that require unprecedented depth
The Thermo Scientific Orbitrap Astral Mass Spectrometer offers a new level of depth in characterization of the proteins, with industry leading sensitivity of analysis, while maintaining the ability to analyze samples of high dynamic range. The platform supports samples of low amount (<pg) and high amount (>ug) without compromise or saturation of analysis.
Orbitrap Astral Mass Spectrometer
Orbitrap Ascend Structural Biology Mass Spectrometer for experiments that require ultimate characterization and versatility
The Thermo Scientific Orbitrap Ascend Tribrid Mass Spectrometer leverages exceptional versatility of analysis to enable ultimate characterization of labeled samples.
Flexible fragmentation modes with CID, HCD, ETD, EtHCD and UVPD enable complementary spectra to provide unambiguous identification of modified amino acids by bottom-up or top-down approaches.
Orbitrap Ascend Structural Biology MS
Orbitrap Exploris 480 Mass Spectrometer for experiments that require Orbitrap accuracy and enhanced selectivity for accurate quantitation
The Thermo Scientific Orbitrap Exploris 480 Mass Spectrometer offers proven reliability and spectral quality to support accurate identification of modification sites. The platform offers versatile operation to support data-dependent or data-independent acquisitions for discovery and quantitation with high-resolution accurate mass.
Orbitrap Exploris 480 Mass Spectrometer
Orbitrap Exploris 240 Mass Spectrometer for experiments that require application flexibility and quantitative precision and accuracy
The Thermo Scientific Orbitrap Exploris 240 Mass Spectrometer offers proven reliability and spectral quality to support accurate identification and quantification of modification sites.
High-quality Orbitrap DDA spectra in conjunction with Thermo Scientific BioPharma Finder Software or Proteome Discoverer Software ensures confident and rapid identification of all peptides.
Orbitrap Exploris 240 Mass Spectrometer
Data analysis for Protein Footprinting
Proteome Discoverer Software - For experiments that require flexible data analysis
Thermo Scientific Proteome Discoverer Software offers predefined workflows to support both LFQ and TMT quantification, PTMs.
Proteome Discoverer Software
BioPharma Finder Software
Thermo Scientific BioPharma Finder Software supports all MS-based Protein Footprinting data analysis, including peptide identification, PTM analysis and quantification.
BioPharma Finder Software
For Research Use Only. Not for use in diagnostic procedures.