Madin-Darby canine kidney cells. LysoSensor Yellow/Blue DND-160.
Dual-emission ratiometric measurement of lysosomal pH using LysoSensor Yellow/Blue DND-160 (L7545). Madin-Darby canine kidney cells were exposed to pH-calibration buffers (pH 4.5, top panel; or pH 7.0, bottom panel) in the presence of nigericin (N1495) and monensin. These pseudocolored images were constructed from two emission images at 450 +/- 33 nm and 510 +/- 20 nm, both excited at 365 +/- 8 nm. (In these images, fluorescence near pH 4 has been pseudocolored blue and fluorescence near pH 7 pseudocolored yellow-green.)
Dual-emission ratiometric measurement of lysosomal pH using LysoSensor Yellow/Blue DND-160 (L7545). Madin-Darby canine kidney cells were exposed to pH-calibration buffers (pH 4.5, top panel; or pH 7.0, bottom panel) in the presence of nigericin (N1495) and monensin. These pseudocolored images were constructed from two emission images at 450 +/- 33 nm and 510 +/- 20 nm, both excited at 365 +/- 8 nm. (In these images, fluorescence near pH 4 has been pseudocolored blue and fluorescence near pH 7 pseudocolored yellow-green.)
Related Products
Catalog # Name Size Price (USD) Qty
L7545 LysoSensor™ Yellow/Blue DND-160 - Special Packaging Each
516.00
Related Images