Blood Screening and Typing
Supporting the screening needs of blood banks and medical laboratory professionals
Our assays are designed to facilitate blood genotyping and antibody screening, with products that allow for an efficient workflow, including fast and simple data analysis and reporting.
LinkSēq ABO typing kits
LABType Assay
LinkSēq HPA typing kits
LinkSeq Assay
LABScreen Multi multiplex assay
AllType NGS Assay
Blood screening and typing categories
Blood genotyping
Our LinkSēq blood products offer rapid genotyping of ABO and HPA to predict comprehensive phenotype profiles.
Blood screening
Our LABScreen Multi assay allows for simultaneous detection of HLA and HNA antibodies in a single well to efficiently stratify the risk of Transfusion-Related Acute Lung Injury (TRALI).
Product support and software files
Access One Lambda software, product documentation, and support files                                                   
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.