Applied Biosystems™
GeneChip™ System 3000
Catalog number: 00-0218
Have Questions?
Applied Biosystems™
GeneChip™ System 3000
Catalog number: 00-0218
Have Questions?
Catalog Number
00-0218
Unit Size
1 each
Price (USD)
Catalog NumberUnit SizePrice (USD)
00-02181 each
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip instrument system is a fully integrated platform for conducting your research using GeneChip brand probe arrays. The GeneChip Scanner 3000 7G System includes the GeneChip Scanner 3000 7G pre-assembled with the Autoloader, Fluidics Station 450, and Hybridization Oven 645i. This system can be purchased with or without a powerful computer workstation with quad-core Intel Core i9-12900 processor loaded with GeneChip Data Collection (GCDC) software. The instrument is used for multiple application areas, such as whole-genome association and targeted genotyping..
The purchase price includes a one-year service agreement.
Related Links
GeneChip Scanner 3000 7G, enabled for High-Resolution Scanning*
GeneChip Operating Software (GCOS) v1.1.1 (includes the GeneChip Scanner 3000 High-Resolution Scanning Patch
*GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, such as serial number series 501, will require the GeneChip Scanner 3000 High-Resolution Update, Catalog Number 00-0110, to be installed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions
22.5 x 31 x 44.5 in. (GeneChip Scanner 3000 with Autoloader)
28 x 16.1 x 15.8 in. (GeneChip Fluidic Station 450)
Product Line
GeneChip™
For Use With (Application)
GeneChip Array Scanning
Weight
105 lb. (GeneChip Scanner 3000 with Autoloader), 80 lb. (GeneChip Fluidic Station 450)