Invitrogen™
eBioscience™ TMB Solution (1X)
Catalog number: 00-4201-56
Have Questions?
Invitrogen™
eBioscience™ TMB Solution (1X)
Catalog number: 00-4201-56
Have Questions?
Catalog Number
00-4201-56
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
00-4201-56100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The 1x eBioscience TMB Solution is a sensitive chromogenic substrate supplied as single ready-to-use solution. TMB (3,3’,5,5’-Tetramethylbenzidine) is a substrate for horseradish peroxidase (HRP) and is recommended for Ready-SET-Go! ELISA kits.
During the enzymatic reaction the TMB substrate will result in a blue color that can be read on a spectrophotometer at 650 nm. This kinetic reaction can be stopped using 1M phosphoric acid resulting in a color change to yellow that can be read at 450 nm.
Reported Application
ELISA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
TMB Solution
Detection Method
Colorimetric
Product Line
eBioscience™
Substrate Properties
Chemical Substrate
Substrate Type
HRP (Horseradish Peroxidase)