Invitrogen™
eBioscience™ 1X RBC Lysis Buffer
Catalog number: 00-4333-57
Have Questions?
Invitrogen™
eBioscience™ 1X RBC Lysis Buffer
Catalog number: 00-4333-57
Have Questions?
Catalog Number
00-4333-57
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
00-4333-57200 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This 1X Red Blood Cell (RBC) Lysis Buffer is formulated for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood. This buffer contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes when used as instructed. Nucleated red cells are not effectively lysed with ammonium chloride.
Reported Application
Flow Cytometric Analysis, Cell Culture
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
eBioscience™
Quantity
200 mL
Format
Liquid
Product Type
Lysis Buffer
Volume (Metric)
200 mL