eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
Product Image
Invitrogen™
eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer. PHA is a mixtureRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4977-03Promo Image4 x 100 μL
00-4977-93Promo Image100 μL
Catalog number 00-4977-03
Price (USD)
254.00
Each
Add to cart
Quantity:
4 x 100 μL
Price (USD)
254.00
Each
Add to cart
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer. PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris. PHA-L is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePhytohemagglutinin-L (PHA-L) Solution
Quantity4 x 100 μL
Product LineeBioscience™
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link