1x1 image pixel for data collection
eBioscience™ 1-step Fix/Lyse Solution (10X)
Product Image
Invitrogen™
eBioscience™ 1-step Fix/Lyse Solution (10X)
The eBioscience 1-step Fix/Lyse Solution enables lysis of red blood cells after staining peripheral blood cells with fluorochrome-conjugated antibodies. ThisRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-5333-57Promo Image200 mL
00-5333-54Promo Image50 mL
Catalog number 00-5333-57
Price (USD)
538.00
Each
Add to cart
Quantity:
200 mL
Price (USD)
538.00
Each
Add to cart
The eBioscience 1-step Fix/Lyse Solution enables lysis of red blood cells after staining peripheral blood cells with fluorochrome-conjugated antibodies. This solution has been specially formulated to lyse non-nucleated erythrocytes while maintaining a fixed and labeled leukocyte population. Therefore, whole blood samples can be stained for the appropriate markers, RBC-lysed and fixed, washed, and then analyzed by flow cytometry. Isolation of mononuclear cells by gradient centrifugation (e.g. Ficoll-Paque density gradient separation) is not required when using this solution. The eBioscience 1-step Fix/Lyse Solution is compatible with antibodies conjugated to organic dyes and tandem dyes.
Reported Application
Flow Cytometric Analysis
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineeBioscience™
Quantity200 mL
Type1-step Fixation/Lyse Solution
Unit SizeEach
Frequently asked questions (FAQs)
Can I use homemade fixation buffers (or other commercially available fixatives) on my cells after staining with eVolve-conjugated antibodies?
No. For optimal results, we recommend that you use the IC Fixation Buffer (Cat. No. 00-8222-49) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54 or 00-5333-57). Impurities in fixation buffers can severely affect fluorochrome brightness.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can I use homemade fixation buffers (or other commercially available fixative) on my cells followed by flow cytometric analysis?
No. We do not recommend the use of other fixatives. We validate our studies using eBioscience's IC Fixation Buffer (Cat. Nos. 00-8222-49, 00-8222-52) and 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Other recipes or manufacturer's products may give inferior results.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I can not run my samples for flow cytometric analysis immediately. What do I do to preserve the best intracellular staining?
For optimal results, we recommend that you analyze stained cells immediately.
However, if you do need to store your samples that were stained with antibodies conjugated to organic fluorochromes, we recommend you complete your staining protocol and fix your samples with IC Fixation Buffer (Cat. Nos. 00-8222-49, 00-8222-52) (100 µL sample with 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Cells can be stored in these buffers for up to 3 days in the dark at 4 degrees C.
We have observed minimal impact on brightness or FRET efficiency/compensation when using the IC Fixation (Cat. Nos. 00-8222-49, 00-8222-52) or 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Differences in fixation buffer quality can affect fluorochrome brightness or FRET efficiency. Fixation of tandem dyes, such as APC-eFluor 780 and PE-Cy7, does not increase the amount of compensation required from the APC or PE detector, respectively. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can samples stained with antibodies conjugates to tandem dyes (i.e., APC-eFluor 780, PE-Cy7, etc.) be fixed and stored?
Yes, samples that are stained with tandem dye conjugated antibodies can be stored in IC Fixation Buffer (Cat. No. 00-8222-49, 00-8222-52) (100 µL sample with 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) for up to 3 days in the dark at 4 degrees C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link