eBioscience™ Permeabilization Buffer (10X)
Product Image
Invitrogen™
eBioscience™ Permeabilization Buffer (10X)
The Permeabilization Buffer (10X), in conjunction with the Intracellular (IC) Fixation Buffer (cat. 00-8222) or Foxp3 Transcription Factor Fixation/Permeabilization ConcentrateRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Catalog NumberQuantity
00-8333-56Promo Image100 mL
Catalog number 00-8333-56
Price (USD)
105.65
Online Exclusive
141.00
Save 35.35 (25%)
Each
Add to cart
Quantity:
100 mL
Price (USD)
105.65
Online Exclusive
141.00
Save 35.35 (25%)
Each
Add to cart
The Permeabilization Buffer (10X), in conjunction with the Intracellular (IC) Fixation Buffer (cat. 00-8222) or Foxp3 Transcription Factor Fixation/Permeabilization Concentrate and Diluent (cat. 00-5521), can be used to fix and permeabilize cells prior to performing intracellular staining of cytokines and other cytoplasmic and/or nuclear antigens. These solutions have been specially formulated for reducing non-specific staining of fluorochrome-labeled antibodies and increasing fluorescence signal to noise ratios.
In intracellular staining protocols, fixation is performed first, followed by permeabilization. Subsequent washing steps, antibody dilutions, and incubations after permeabilization should use the Permeabilization Buffer. The final washing step and cell resuspension prior to running the sample is done using Flow Cytometry Staining Buffer.
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 mL
Reagent TypePermeabilization Reagent
Product LineeBioscience™
Product TypePermeabilization Buffer
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection