Thermo Scientific Chemicals

Cadmium foil, 0.025mm (0.001 in.) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 000064.FQ
25 x 100 mm, Each
Thermo Scientific Chemicals

Cadmium foil, 0.025mm (0.001 in.) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 000064.FQ
25 x 100 mm, Each
Quantity
Catalog number: 000064.FQ
also known as 000064-FQ
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-43-9
IUPAC Name
cadmium
Molecular Formula
Cd
InChI Key
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES
[Cd]
Form
Foil
Appearance (Color)
Silver white
Comment
0.023 - 0.028 mm thick
Total Metal Impurities
0.001% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text