Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis), Thermo Scientific Chemicals
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
50 x 250 mm
Catalog number 000095.FI
also known as 000095-FI
Price (USD)/ Each
108.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
108.00
Add to cart
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis), Thermo Scientific Chemicals
Catalog number000095.FI
Price (USD)/ Each
108.00
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Copper
Comment0.114 - 0.140mm thick
Total Metal Impurities<0.01%
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only